Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 237088502-237088763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS8
Entrez Gene ID
10920
Gene Name
COP9 signalosome subunit 8
Gene Aliases
COP9, CSN8, SGN8
UniGene
Hs.531713
Gene Chromosome Location
Chr.2: 237085441-237098846 on Build GRCh38
Transcripts
NM_006710.4 NM_198189.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6434782 c.195A>T p.K65N Chr.2: 237088650-237088650 on Build GRCh38
3051208 c.150-9G>A p.? Chr.2: 237088596-237088596 on Build GRCh38
4674197 c.158C>T p.A53V Chr.2: 237088613-237088613 on Build GRCh38
6427167 c.181C>A p.P61T Chr.2: 237088636-237088636 on Build GRCh38
6010732 c.197C>T p.S66F Chr.2: 237088652-237088652 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs942475172 Chr.2: 237088579-237088579 on Build GRCh38
rs776223966 Chr.2: 237088596-237088596 on Build GRCh38
rs779023176 Chr.2: 237088578-237088578 on Build GRCh38
rs756550400 Chr.2: 237088699-237088699 on Build GRCh38
rs747000438 Chr.2: 237088591-237088591 on Build GRCh38
rs765906015 Chr.2: 237088689-237088689 on Build GRCh38
rs910989685 Chr.2: 237088543-237088543 on Build GRCh38
rs764905331 Chr.2: 237088613-237088613 on Build GRCh38
rs115111928 Chr.2: 237088645-237088645 on Build GRCh38
rs775047015 Chr.2: 237088616-237088616 on Build GRCh38
rs768861355 Chr.2: 237088577-237088577 on Build GRCh38
rs369320947 Chr.2: 237088595-237088595 on Build GRCh38
rs73999614 Chr.2: 237088726-237088726 on Build GRCh38
rs377666250 Chr.2: 237088682-237088682 on Build GRCh38
rs34316977 Chr.2: 237088566-237088567 on Build GRCh38
rs989199237 Chr.2: 237088662-237088662 on Build GRCh38
rs144623210 Chr.2: 237088607-237088607 on Build GRCh38
rs771917751 Chr.2: 237088575-237088575 on Build GRCh38
rs113198349 Chr.2: 237088556-237088556 on Build GRCh38
rs749370827 Chr.2: 237088562-237088562 on Build GRCh38
rs771801300 Chr.2: 237088584-237088584 on Build GRCh38
rs183141618 Chr.2: 237088581-237088581 on Build GRCh38
rs750929496 Chr.2: 237088686-237088686 on Build GRCh38
rs746337967 Chr.2: 237088555-237088557 on Build GRCh38
rs758990973 Chr.2: 237088606-237088606 on Build GRCh38
rs373635523 Chr.2: 237088640-237088640 on Build GRCh38
rs57864418 Chr.2: 237088587-237088587 on Build GRCh38

View Full Product Details