Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 75947311-75947576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IMPG1
Entrez Gene ID
3617
Gene Name
interphotoreceptor matrix proteoglycan 1
Gene Aliases
GP147, IPM150, SPACR, VMD4
UniGene
Hs.590893
Gene Chromosome Location
Chr.6: 75921114-76072678 on Build GRCh38
Transcripts
NM_001282368.1 NM_001563.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6471389 c.1923T>A p.F641L Chr.6: 75947435-75947435 on Build GRCh38
1634940 c.1975G>T p.D659Y Chr.6: 75947383-75947383 on Build GRCh38
3875627 c.2004C>T p.L668L Chr.6: 75947354-75947354 on Build GRCh38
1644190 c.1975G>A p.D659N Chr.6: 75947383-75947383 on Build GRCh38
6488639 c.2041C>A p.P681T Chr.6: 75947317-75947317 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762387361 Chr.6: 75947393-75947393 on Build GRCh38
rs749953054 Chr.6: 75947392-75947392 on Build GRCh38
rs984685022 Chr.6: 75947515-75947515 on Build GRCh38
rs771309602 Chr.6: 75947349-75947349 on Build GRCh38
rs145955265 Chr.6: 75947352-75947352 on Build GRCh38
rs773080033 Chr.6: 75947441-75947441 on Build GRCh38
rs556959981 Chr.6: 75947516-75947516 on Build GRCh38
rs776335869 Chr.6: 75947421-75947421 on Build GRCh38
rs148615908 Chr.6: 75947399-75947399 on Build GRCh38
rs376390542 Chr.6: 75947509-75947509 on Build GRCh38
rs573887374 Chr.6: 75947401-75947401 on Build GRCh38
rs774647095 Chr.6: 75947363-75947363 on Build GRCh38
rs75117418 Chr.6: 75947457-75947457 on Build GRCh38
rs78305682 Chr.6: 75947402-75947402 on Build GRCh38
rs749382585 Chr.6: 75947414-75947414 on Build GRCh38
rs1037821641 Chr.6: 75947451-75947451 on Build GRCh38
rs746872890 Chr.6: 75947466-75947474 on Build GRCh38
rs749332855 Chr.6: 75947340-75947340 on Build GRCh38
rs147614663 Chr.6: 75947522-75947522 on Build GRCh38
rs756434628 Chr.6: 75947476-75947476 on Build GRCh38
rs145267469 Chr.6: 75947468-75947468 on Build GRCh38
rs779171435 Chr.6: 75947513-75947513 on Build GRCh38
rs371118175 Chr.6: 75947517-75947517 on Build GRCh38
rs770835713 Chr.6: 75947544-75947544 on Build GRCh38
rs760558589 Chr.6: 75947373-75947373 on Build GRCh38
rs777740210 Chr.6: 75947337-75947340 on Build GRCh38
rs1008603589 Chr.6: 75947434-75947434 on Build GRCh38
rs118155926 Chr.6: 75947413-75947413 on Build GRCh38
rs764710111 Chr.6: 75947489-75947489 on Build GRCh38
rs139192386 Chr.6: 75947426-75947426 on Build GRCh38
rs181256434 Chr.6: 75947497-75947497 on Build GRCh38
rs202200012 Chr.6: 75947418-75947418 on Build GRCh38
rs761616758 Chr.6: 75947422-75947422 on Build GRCh38
rs371197189 Chr.6: 75947408-75947408 on Build GRCh38
rs376165008 Chr.6: 75947427-75947427 on Build GRCh38
rs751040344 Chr.6: 75947461-75947461 on Build GRCh38
rs780784630 Chr.6: 75947479-75947480 on Build GRCh38
rs201013836 Chr.6: 75947420-75947420 on Build GRCh38
rs766036273 Chr.6: 75947394-75947394 on Build GRCh38
rs906664476 Chr.6: 75947375-75947375 on Build GRCh38
rs151138934 Chr.6: 75947525-75947525 on Build GRCh38
rs759162476 Chr.6: 75947465-75947465 on Build GRCh38
rs769638626 Chr.6: 75947521-75947521 on Build GRCh38
rs754657889 Chr.6: 75947396-75947396 on Build GRCh38
rs200128845 Chr.6: 75947376-75947376 on Build GRCh38
rs940063319 Chr.6: 75947353-75947353 on Build GRCh38
rs745936741 Chr.6: 75947518-75947518 on Build GRCh38
rs1051579797 Chr.6: 75947482-75947482 on Build GRCh38
rs767150099 Chr.6: 75947467-75947467 on Build GRCh38
rs267601131 Chr.6: 75947409-75947409 on Build GRCh38
rs746117801 Chr.6: 75947482-75947486 on Build GRCh38
rs765195875 Chr.6: 75947390-75947390 on Build GRCh38
rs757107031 Chr.6: 75947477-75947478 on Build GRCh38
rs761693136 Chr.6: 75947377-75947377 on Build GRCh38
rs751273411 Chr.6: 75947395-75947395 on Build GRCh38

View Full Product Details