Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 79277873-79278120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JMY
Entrez Gene ID
133746
Gene Name
junction mediating and regulatory protein, p53 cofactor
Gene Aliases
WHAMM2, WHDC1L3
UniGene
Hs.482605
Gene Chromosome Location
Chr.5: 79236102-79327215 on Build GRCh38
Transcripts
NM_152405.4 XM_005248430.2 XM_011543155.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1595653 c.1132G>A p.E378K Chr.5: 79278009-79278009 on Build GRCh38
1595654 c.1110C>T p.S370S Chr.5: 79277987-79277987 on Build GRCh38
6689662 c.144+4delT p.? Chr.5: 79278087-79278087 on Build GRCh38
390251 c.25C>G p.Q9E Chr.5: 79277964-79277964 on Build GRCh38
6689666 c.101G>A p.R34Q Chr.5: 79278040-79278040 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368041668 Chr.5: 79278003-79278003 on Build GRCh38
rs774133482 Chr.5: 79278016-79278016 on Build GRCh38
rs200019667 Chr.5: 79277926-79277926 on Build GRCh38
rs1031034611 Chr.5: 79277952-79277952 on Build GRCh38
rs202056721 Chr.5: 79277904-79277904 on Build GRCh38
rs776896603 Chr.5: 79277987-79277987 on Build GRCh38
rs371768539 Chr.5: 79278007-79278007 on Build GRCh38
rs778644205 Chr.5: 79278053-79278053 on Build GRCh38
rs766722714 Chr.5: 79277976-79277976 on Build GRCh38
rs773986761 Chr.5: 79277949-79277949 on Build GRCh38
rs750324553 Chr.5: 79278013-79278013 on Build GRCh38
rs759301275 Chr.5: 79278067-79278067 on Build GRCh38
rs759535147 Chr.5: 79278019-79278019 on Build GRCh38
rs756046634 Chr.5: 79278014-79278014 on Build GRCh38
rs13182512 Chr.5: 79277967-79277967 on Build GRCh38
rs373711492 Chr.5: 79278008-79278008 on Build GRCh38
rs367813106 Chr.5: 79278026-79278026 on Build GRCh38
rs373904471 Chr.5: 79277995-79277995 on Build GRCh38
rs747819265 Chr.5: 79278066-79278066 on Build GRCh38
rs527437715 Chr.5: 79278029-79278029 on Build GRCh38
rs199532823 Chr.5: 79277951-79277951 on Build GRCh38
rs747515769 Chr.5: 79278076-79278076 on Build GRCh38
rs768474540 Chr.5: 79277915-79277915 on Build GRCh38
rs748919353 Chr.5: 79277906-79277906 on Build GRCh38
rs772625753 Chr.5: 79278070-79278070 on Build GRCh38
rs764095029 Chr.5: 79278010-79278010 on Build GRCh38
rs866544367 Chr.5: 79278017-79278017 on Build GRCh38
rs747404238 Chr.5: 79278086-79278087 on Build GRCh38
rs752768348 Chr.5: 79278006-79278006 on Build GRCh38
rs771388300 Chr.5: 79278093-79278093 on Build GRCh38
rs942882588 Chr.5: 79277983-79277983 on Build GRCh38
rs754674187 Chr.5: 79278047-79278047 on Build GRCh38
rs61730052 Chr.5: 79277961-79277961 on Build GRCh38
rs35345418 Chr.5: 79277955-79277955 on Build GRCh38

View Full Product Details