Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 33129830-33130055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTAP
Entrez Gene ID
10491
Gene Name
cartilage associated protein
Gene Aliases
CASP, LEPREL3, OI7, P3H5
UniGene
Hs.517888
Gene Chromosome Location
Chr.3: 33113958-33147773 on Build GRCh38
Transcripts
NM_006371.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
122366 c.832G>A p.E278K Chr.3: 33129977-33129977 on Build GRCh38
4323647 c.830G>A p.C277Y Chr.3: 33129975-33129975 on Build GRCh38
1202328 c.866C>T p.P289L Chr.3: 33130011-33130011 on Build GRCh38
3592514 c.847C>T p.P283S Chr.3: 33129992-33129992 on Build GRCh38
6307186 c.805G>T p.E269* Chr.3: 33129950-33129950 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746374230 Chr.3: 33130012-33130012 on Build GRCh38
rs767792216 Chr.3: 33129953-33129953 on Build GRCh38
rs775037541 Chr.3: 33129938-33129938 on Build GRCh38
rs748772443 Chr.3: 33130000-33130000 on Build GRCh38
rs780858493 Chr.3: 33129905-33129905 on Build GRCh38
rs137853946 Chr.3: 33129967-33129971 on Build GRCh38
rs746800696 Chr.3: 33129894-33129894 on Build GRCh38
rs72659361 Chr.3: 33129971-33129971 on Build GRCh38
rs747772727 Chr.3: 33129996-33129996 on Build GRCh38
rs780866141 Chr.3: 33129993-33129993 on Build GRCh38
rs200208631 Chr.3: 33129899-33129899 on Build GRCh38
rs137853945 Chr.3: 33129949-33129954 on Build GRCh38
rs775719763 Chr.3: 33129954-33129954 on Build GRCh38
rs72659362 Chr.3: 33130024-33130024 on Build GRCh38
rs147140948 Chr.3: 33130011-33130011 on Build GRCh38
rs754756497 Chr.3: 33129991-33129991 on Build GRCh38
rs916529347 Chr.3: 33129884-33129884 on Build GRCh38
rs748864250 Chr.3: 33129927-33129927 on Build GRCh38
rs759900212 Chr.3: 33129941-33129941 on Build GRCh38
rs751486898 Chr.3: 33129985-33129985 on Build GRCh38
rs202206745 Chr.3: 33129936-33129936 on Build GRCh38
rs760809534 Chr.3: 33129962-33129962 on Build GRCh38
rs772842929 Chr.3: 33129926-33129926 on Build GRCh38
rs992456389 Chr.3: 33129867-33129867 on Build GRCh38
rs923457111 Chr.3: 33129982-33129982 on Build GRCh38
rs573928986 Chr.3: 33129920-33129920 on Build GRCh38
rs758330236 Chr.3: 33129891-33129891 on Build GRCh38
rs777614682 Chr.3: 33129999-33129999 on Build GRCh38
rs952340977 Chr.3: 33129864-33129864 on Build GRCh38
rs947704311 Chr.3: 33129910-33129910 on Build GRCh38
rs984458663 Chr.3: 33129946-33129946 on Build GRCh38
rs766277607 Chr.3: 33129975-33129975 on Build GRCh38
rs560255943 Chr.3: 33129892-33129892 on Build GRCh38
rs764370024 Chr.3: 33129970-33129970 on Build GRCh38
rs770592109 Chr.3: 33130005-33130005 on Build GRCh38
rs747779936 Chr.3: 33129909-33129909 on Build GRCh38
rs112423184 Chr.3: 33129947-33129947 on Build GRCh38
rs755811700 Chr.3: 33129998-33129998 on Build GRCh38
rs757381053 Chr.3: 33129973-33129973 on Build GRCh38
rs770107661 Chr.3: 33129922-33129923 on Build GRCh38

View Full Product Details