Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 62353172-62353445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF2A
Entrez Gene ID
3796
Gene Name
kinesin family member 2A
Gene Aliases
CDCBM3, HK2, KIF2
UniGene
Hs.558351
Gene Chromosome Location
Chr.5: 62306162-62387184 on Build GRCh38
Transcripts
NM_001098511.2 NM_001243952.1 NM_001243953.1 NM_004520.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
482942 c.458-2A>G p.? Chr.5: 62353273-62353273 on Build GRCh38
5925177 c.558+4C>T p.? Chr.5: 62353379-62353379 on Build GRCh38
7093831 c.475T>G p.C159G Chr.5: 62353292-62353292 on Build GRCh38
6419713 c.448C>A p.Q150K Chr.5: 62353346-62353346 on Build GRCh38
1069239 c.501A>C p.Q167H Chr.5: 62353318-62353318 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771690532 Chr.5: 62353202-62353202 on Build GRCh38
rs767921213 Chr.5: 62353247-62353247 on Build GRCh38
rs759122457 Chr.5: 62353391-62353392 on Build GRCh38
rs777911286 Chr.5: 62353339-62353339 on Build GRCh38
rs151192663 Chr.5: 62353379-62353379 on Build GRCh38
rs754680482 Chr.5: 62353260-62353260 on Build GRCh38
rs778387516 Chr.5: 62353262-62353262 on Build GRCh38
rs757923895 Chr.5: 62353328-62353328 on Build GRCh38
rs965137816 Chr.5: 62353384-62353384 on Build GRCh38
rs747697385 Chr.5: 62353277-62353277 on Build GRCh38
rs979832337 Chr.5: 62353313-62353313 on Build GRCh38
rs767133148 Chr.5: 62353392-62353396 on Build GRCh38
rs770886614 Chr.5: 62353393-62353393 on Build GRCh38
rs770088962 Chr.5: 62353405-62353405 on Build GRCh38
rs776462516 Chr.5: 62353401-62353401 on Build GRCh38
rs200784447 Chr.5: 62353408-62353408 on Build GRCh38
rs878957393 Chr.5: 62353200-62353200 on Build GRCh38
rs745604102 Chr.5: 62353404-62353404 on Build GRCh38
rs753474693 Chr.5: 62353253-62353253 on Build GRCh38
rs774642235 Chr.5: 62353407-62353407 on Build GRCh38
rs757678056 Chr.5: 62353245-62353245 on Build GRCh38
rs200767176 Chr.5: 62353412-62353412 on Build GRCh38

View Full Product Details