Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96761730-96761971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNNM4
Entrez Gene ID
26504
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 4
Gene Aliases
ACDP4
UniGene
Hs.175043
Gene Chromosome Location
Chr.2: 96760902-96811891 on Build GRCh38
Transcripts
NM_020184.3 XM_005263914.3 XM_005263915.3 XM_011510955.2 XM_011510956.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3799289 c.808A>T p.M270L Chr.2: 96761807-96761807 on Build GRCh38
6092739 c.814G>T p.V272L Chr.2: 96761813-96761813 on Build GRCh38
5172746 c.967C>T p.L323F Chr.2: 96761966-96761966 on Build GRCh38
7294722 c.740T>A p.L247H Chr.2: 96761739-96761739 on Build GRCh38
5397616 c.777C>T p.I259I Chr.2: 96761776-96761776 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs56246512 Chr.2: 96761783-96761783 on Build GRCh38
rs763749179 Chr.2: 96761771-96761771 on Build GRCh38
rs370137332 Chr.2: 96761758-96761758 on Build GRCh38
rs773439754 Chr.2: 96761795-96761795 on Build GRCh38
rs999284391 Chr.2: 96761848-96761848 on Build GRCh38
rs776551771 Chr.2: 96761828-96761828 on Build GRCh38
rs766669677 Chr.2: 96761812-96761812 on Build GRCh38
rs1043975317 Chr.2: 96761801-96761801 on Build GRCh38
rs977087685 Chr.2: 96761776-96761776 on Build GRCh38
rs749703825 Chr.2: 96761762-96761762 on Build GRCh38
rs759468747 Chr.2: 96761833-96761833 on Build GRCh38
rs150269409 Chr.2: 96761787-96761787 on Build GRCh38
rs757875854 Chr.2: 96761869-96761869 on Build GRCh38
rs559372337 Chr.2: 96761809-96761809 on Build GRCh38
rs780881771 Chr.2: 96761938-96761938 on Build GRCh38
rs752693477 Chr.2: 96761866-96761866 on Build GRCh38
rs772062538 Chr.2: 96761794-96761794 on Build GRCh38
rs755344155 Chr.2: 96761948-96761948 on Build GRCh38
rs935793378 Chr.2: 96761796-96761796 on Build GRCh38
rs749650885 Chr.2: 96761941-96761941 on Build GRCh38
rs373657140 Chr.2: 96761926-96761926 on Build GRCh38
rs35310683 Chr.2: 96761805-96761806 on Build GRCh38
rs746662702 Chr.2: 96761749-96761749 on Build GRCh38
rs903670676 Chr.2: 96761845-96761845 on Build GRCh38
rs763543912 Chr.2: 96761891-96761891 on Build GRCh38
rs377727962 Chr.2: 96761755-96761755 on Build GRCh38
rs760772919 Chr.2: 96761802-96761802 on Build GRCh38
rs774907198 Chr.2: 96761773-96761773 on Build GRCh38
rs765198380 Chr.2: 96761842-96761842 on Build GRCh38
rs751120696 Chr.2: 96761919-96761919 on Build GRCh38

View Full Product Details