Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74753113-74753346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP1A2
Entrez Gene ID
1544
Gene Name
cytochrome P450 family 1 subfamily A member 2
Gene Aliases
CP12, P3-450, P450(PA)
UniGene
Hs.1361
Gene Chromosome Location
Chr.15: 74748843-74756600 on Build GRCh38
Transcripts
NM_000761.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3503986 c.1177G>A p.D393N Chr.15: 74753194-74753194 on Build GRCh38
3401917 c.1253+1G>A p.? Chr.15: 74753271-74753271 on Build GRCh38
5079254 c.1253+8C>T p.? Chr.15: 74753278-74753278 on Build GRCh38
6142743 c.1226T>C p.V409A Chr.15: 74753243-74753243 on Build GRCh38
4601735 c.1240G>A p.V414I Chr.15: 74753257-74753257 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750625510 Chr.15: 74753233-74753234 on Build GRCh38
rs771872995 Chr.15: 74753289-74753289 on Build GRCh38
rs547136051 Chr.15: 74753290-74753290 on Build GRCh38
rs748007860 Chr.15: 74753282-74753282 on Build GRCh38
rs768486827 Chr.15: 74753270-74753270 on Build GRCh38
rs760711594 Chr.15: 74753211-74753211 on Build GRCh38
rs376308231 Chr.15: 74753235-74753235 on Build GRCh38
rs748737498 Chr.15: 74753258-74753258 on Build GRCh38
rs758805995 Chr.15: 74753290-74753291 on Build GRCh38
rs765437776 Chr.15: 74753185-74753187 on Build GRCh38
rs199635816 Chr.15: 74753176-74753176 on Build GRCh38
rs149928755 Chr.15: 74753201-74753201 on Build GRCh38
rs774289191 Chr.15: 74753185-74753185 on Build GRCh38
rs760798316 Chr.15: 74753133-74753133 on Build GRCh38
rs915586110 Chr.15: 74753202-74753202 on Build GRCh38
rs946811612 Chr.15: 74753275-74753275 on Build GRCh38
rs778340906 Chr.15: 74753160-74753160 on Build GRCh38
rs56107638 Chr.15: 74753271-74753271 on Build GRCh38
rs752138293 Chr.15: 74753243-74753243 on Build GRCh38
rs779272470 Chr.15: 74753138-74753138 on Build GRCh38
rs755156570 Chr.15: 74753137-74753137 on Build GRCh38
rs546190728 Chr.15: 74753203-74753203 on Build GRCh38
rs776776985 Chr.15: 74753299-74753299 on Build GRCh38
rs911014711 Chr.15: 74753200-74753200 on Build GRCh38
rs766562768 Chr.15: 74753136-74753136 on Build GRCh38
rs866111643 Chr.15: 74753221-74753221 on Build GRCh38
rs745416686 Chr.15: 74753166-74753166 on Build GRCh38
rs2960193 Chr.15: 74753226-74753226 on Build GRCh38
rs753816138 Chr.15: 74753223-74753223 on Build GRCh38
rs528804804 Chr.15: 74753241-74753241 on Build GRCh38
rs377527644 Chr.15: 74753242-74753242 on Build GRCh38
rs748970784 Chr.15: 74753178-74753178 on Build GRCh38
rs150839466 Chr.15: 74753238-74753238 on Build GRCh38
rs759609232 Chr.15: 74753305-74753305 on Build GRCh38
rs201515531 Chr.15: 74753149-74753149 on Build GRCh38
rs1052561630 Chr.15: 74753261-74753261 on Build GRCh38
rs753003400 Chr.15: 74753142-74753142 on Build GRCh38
rs766514583 Chr.15: 74753217-74753217 on Build GRCh38
rs552580628 Chr.15: 74753175-74753175 on Build GRCh38
rs770715935 Chr.15: 74753297-74753297 on Build GRCh38
rs768362038 Chr.15: 74753182-74753182 on Build GRCh38
rs931216566 Chr.15: 74753162-74753162 on Build GRCh38
rs376674911 Chr.15: 74753257-74753257 on Build GRCh38
rs761693521 Chr.15: 74753191-74753191 on Build GRCh38
rs762326858 Chr.15: 74753193-74753193 on Build GRCh38
rs1050518072 Chr.15: 74753285-74753285 on Build GRCh38
rs773181867 Chr.15: 74753205-74753205 on Build GRCh38
rs1027454005 Chr.15: 74753151-74753151 on Build GRCh38
rs769315070 Chr.15: 74753169-74753169 on Build GRCh38
rs55889066 Chr.15: 74753234-74753234 on Build GRCh38
rs759768496 Chr.15: 74753237-74753237 on Build GRCh38
rs746993211 Chr.15: 74753293-74753293 on Build GRCh38

View Full Product Details