Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 146514770-146514995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR151
Entrez Gene ID
134391
Gene Name
G protein-coupled receptor 151
Gene Aliases
GALR4, GALRL, GPCR, PGR7
UniGene
Hs.483732
Gene Chromosome Location
Chr.5: 146514854-146516113 on Build GRCh38
Transcripts
NM_194251.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4600100 c.1228G>T p.E410* Chr.5: 146514886-146514886 on Build GRCh38
1434933 c.1122A>C p.K374N Chr.5: 146514992-146514992 on Build GRCh38
420848 c.1157T>A p.V386E Chr.5: 146514957-146514957 on Build GRCh38
3919254 c.1246G>A p.E416K Chr.5: 146514868-146514868 on Build GRCh38
5402597 c.1222G>A p.E408K Chr.5: 146514892-146514892 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758402328 Chr.5: 146514911-146514911 on Build GRCh38
rs752838386 Chr.5: 146514855-146514858 on Build GRCh38
rs150820698 Chr.5: 146514869-146514869 on Build GRCh38
rs759659177 Chr.5: 146514831-146514833 on Build GRCh38
rs142971995 Chr.5: 146514930-146514930 on Build GRCh38
rs758630788 Chr.5: 146514856-146514857 on Build GRCh38
rs368998814 Chr.5: 146514852-146514852 on Build GRCh38
rs139779292 Chr.5: 146514899-146514899 on Build GRCh38
rs747233688 Chr.5: 146514818-146514818 on Build GRCh38
rs201437088 Chr.5: 146514955-146514955 on Build GRCh38
rs765293386 Chr.5: 146514834-146514834 on Build GRCh38
rs770654388 Chr.5: 146514950-146514950 on Build GRCh38
rs372506032 Chr.5: 146514927-146514927 on Build GRCh38
rs775200644 Chr.5: 146514966-146514966 on Build GRCh38
rs765826115 Chr.5: 146514900-146514900 on Build GRCh38
rs780118239 Chr.5: 146514916-146514916 on Build GRCh38
rs759067277 Chr.5: 146514856-146514856 on Build GRCh38
rs754298872 Chr.5: 146514887-146514887 on Build GRCh38
rs780205679 Chr.5: 146514816-146514816 on Build GRCh38
rs745805652 Chr.5: 146514958-146514958 on Build GRCh38
rs1044614982 Chr.5: 146514971-146514971 on Build GRCh38
rs751926840 Chr.5: 146514929-146514930 on Build GRCh38
rs750955838 Chr.5: 146514903-146514903 on Build GRCh38
rs762019412 Chr.5: 146514808-146514808 on Build GRCh38
rs79548345 Chr.5: 146514924-146514924 on Build GRCh38
rs771739074 Chr.5: 146514860-146514860 on Build GRCh38
rs769518867 Chr.5: 146514823-146514823 on Build GRCh38
rs775217248 Chr.5: 146514867-146514867 on Build GRCh38
rs760419500 Chr.5: 146514868-146514868 on Build GRCh38
rs760403061 Chr.5: 146514969-146514969 on Build GRCh38
rs913559149 Chr.5: 146514861-146514861 on Build GRCh38
rs749176612 Chr.5: 146514832-146514832 on Build GRCh38
rs770940993 Chr.5: 146514850-146514850 on Build GRCh38
rs749156193 Chr.5: 146514934-146514934 on Build GRCh38
rs986284835 Chr.5: 146514806-146514806 on Build GRCh38
rs764446251 Chr.5: 146514871-146514872 on Build GRCh38
rs777679676 Chr.5: 146514831-146514831 on Build GRCh38
rs139276521 Chr.5: 146514959-146514959 on Build GRCh38
rs957774399 Chr.5: 146514819-146514819 on Build GRCh38
rs368315441 Chr.5: 146514926-146514926 on Build GRCh38

View Full Product Details