Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 14845009-14845264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCB
Entrez Gene ID
2187
Gene Name
Fanconi anemia complementation group B
Gene Aliases
FA2, FAAP90, FAAP95, FAB, FACB
UniGene
Hs.554740
Gene Chromosome Location
Chr.X: 14690863-14873255 on Build GRCh38
Transcripts
NM_001018113.2 NM_001324162.1 NM_152633.3 XM_011545470.2 XM_017029355.1 XM_017029356.1

SNP Detail

SNP ID Chromosome Location
rs749342600 Chr.X: 14845201-14845201 on Build GRCh38
rs763091316 Chr.X: 14845081-14845081 on Build GRCh38
rs144205393 Chr.X: 14845165-14845165 on Build GRCh38
rs775375428 Chr.X: 14845057-14845057 on Build GRCh38
rs375901101 Chr.X: 14845095-14845095 on Build GRCh38
rs200303151 Chr.X: 14845063-14845063 on Build GRCh38
rs756880603 Chr.X: 14845146-14845146 on Build GRCh38
rs868761818 Chr.X: 14845159-14845159 on Build GRCh38
rs746962101 Chr.X: 14845172-14845172 on Build GRCh38
rs771950360 Chr.X: 14845047-14845047 on Build GRCh38
rs758665871 Chr.X: 14845202-14845202 on Build GRCh38
rs146157131 Chr.X: 14845125-14845125 on Build GRCh38
rs745690135 Chr.X: 14845149-14845149 on Build GRCh38
rs767952529 Chr.X: 14845124-14845124 on Build GRCh38
rs756628396 Chr.X: 14845137-14845137 on Build GRCh38
rs370248837 Chr.X: 14845213-14845213 on Build GRCh38
rs763699754 Chr.X: 14845143-14845143 on Build GRCh38
rs752804063 Chr.X: 14845173-14845173 on Build GRCh38
rs780024809 Chr.X: 14845169-14845169 on Build GRCh38
rs905186149 Chr.X: 14845197-14845197 on Build GRCh38
rs768004295 Chr.X: 14845234-14845234 on Build GRCh38
rs759926657 Chr.X: 14845100-14845100 on Build GRCh38
rs753355273 Chr.X: 14845145-14845145 on Build GRCh38
rs766587734 Chr.X: 14845082-14845082 on Build GRCh38
rs373632536 Chr.X: 14845231-14845231 on Build GRCh38
rs976300441 Chr.X: 14845120-14845120 on Build GRCh38
rs773221142 Chr.X: 14845064-14845064 on Build GRCh38
rs778604488 Chr.X: 14845147-14845147 on Build GRCh38
rs773313492 Chr.X: 14845177-14845177 on Build GRCh38

View Full Product Details