Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 5894987-5895258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO18
Entrez Gene ID
84893
Gene Name
F-box protein, helicase, 18
Gene Aliases
FBH1, Fbx18, hFBH1
UniGene
Hs.498543
Gene Chromosome Location
Chr.10: 5889572-5937595 on Build GRCh38
Transcripts
NM_001258452.1 NM_001258453.1 NM_032807.4 NM_178150.2 XM_011519748.2 XM_017016842.1 XM_017016843.1 XM_017016844.1 XM_017016845.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2148032 c.88G>A p.G30S Chr.10: 5895121-5895121 on Build GRCh38
427711 c.121G>T p.A41S Chr.10: 5895154-5895154 on Build GRCh38
4514286 c.96C>T p.L32L Chr.10: 5895129-5895129 on Build GRCh38
3978669 c.109C>T p.Q37* Chr.10: 5895142-5895142 on Build GRCh38
5431192 c.40C>T p.Q14* Chr.10: 5895073-5895073 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs893156633 Chr.10: 5895153-5895153 on Build GRCh38
rs745740691 Chr.10: 5895140-5895140 on Build GRCh38
rs770334474 Chr.10: 5895180-5895180 on Build GRCh38
rs749776152 Chr.10: 5895213-5895213 on Build GRCh38
rs984981051 Chr.10: 5895060-5895060 on Build GRCh38
rs774431806 Chr.10: 5895218-5895218 on Build GRCh38
rs1053356980 Chr.10: 5895147-5895147 on Build GRCh38
rs144792722 Chr.10: 5895166-5895166 on Build GRCh38
rs12355290 Chr.10: 5895042-5895042 on Build GRCh38
rs756023348 Chr.10: 5895045-5895045 on Build GRCh38
rs750220283 Chr.10: 5895113-5895113 on Build GRCh38
rs778513834 Chr.10: 5895055-5895055 on Build GRCh38
rs747612824 Chr.10: 5895145-5895145 on Build GRCh38
rs369588567 Chr.10: 5895013-5895013 on Build GRCh38
rs149817075 Chr.10: 5895131-5895131 on Build GRCh38
rs558961654 Chr.10: 5895114-5895114 on Build GRCh38
rs201249047 Chr.10: 5895046-5895046 on Build GRCh38
rs747783372 Chr.10: 5895063-5895063 on Build GRCh38
rs746215865 Chr.10: 5895167-5895167 on Build GRCh38
rs901147646 Chr.10: 5895233-5895233 on Build GRCh38
rs770714076 Chr.10: 5895121-5895121 on Build GRCh38
rs189353543 Chr.10: 5895026-5895026 on Build GRCh38
rs910298282 Chr.10: 5895077-5895077 on Build GRCh38
rs141022398 Chr.10: 5895104-5895104 on Build GRCh38
rs537254391 Chr.10: 5895108-5895108 on Build GRCh38
rs372680304 Chr.10: 5895047-5895047 on Build GRCh38
rs199546997 Chr.10: 5895217-5895217 on Build GRCh38
rs980466162 Chr.10: 5895074-5895074 on Build GRCh38
rs547418764 Chr.10: 5895109-5895109 on Build GRCh38
rs143563006 Chr.10: 5895076-5895076 on Build GRCh38
rs771992819 Chr.10: 5895152-5895152 on Build GRCh38
rs771649953 Chr.10: 5895073-5895073 on Build GRCh38
rs951730771 Chr.10: 5895059-5895059 on Build GRCh38
rs538251763 Chr.10: 5895105-5895105 on Build GRCh38
rs746229177 Chr.10: 5895082-5895082 on Build GRCh38
rs758060873 Chr.10: 5895012-5895013 on Build GRCh38
rs377389822 Chr.10: 5895232-5895232 on Build GRCh38
rs562885579 Chr.10: 5895087-5895088 on Build GRCh38
rs571359384 Chr.10: 5895088-5895088 on Build GRCh38
rs776024864 Chr.10: 5895202-5895202 on Build GRCh38
rs758008740 Chr.10: 5895149-5895149 on Build GRCh38
rs960582054 Chr.10: 5895040-5895040 on Build GRCh38
rs755927988 Chr.10: 5895119-5895119 on Build GRCh38
rs767773588 Chr.10: 5895225-5895225 on Build GRCh38
rs767649230 Chr.10: 5895110-5895110 on Build GRCh38

View Full Product Details