Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89132264-89132507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBP7
Entrez Gene ID
388646
Gene Name
guanylate binding protein 7
Gene Aliases
GBP4L
UniGene
Hs.591450
Gene Chromosome Location
Chr.1: 89131751-89176040 on Build GRCh38
Transcripts
NM_207398.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3977952 c.1762G>T p.E588* Chr.1: 89132304-89132304 on Build GRCh38
176011 c.1729C>T p.R577* Chr.1: 89132337-89132337 on Build GRCh38
5928086 c.1709C>T p.S570L Chr.1: 89132357-89132357 on Build GRCh38
5029358 c.1725T>A p.I575I Chr.1: 89132341-89132341 on Build GRCh38
110496 c.1789C>T p.L597F Chr.1: 89132277-89132277 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764768326 Chr.1: 89132363-89132363 on Build GRCh38
rs750373956 Chr.1: 89132400-89132400 on Build GRCh38
rs113831821 Chr.1: 89132458-89132458 on Build GRCh38
rs776149483 Chr.1: 89132412-89132413 on Build GRCh38
rs773661892 Chr.1: 89132350-89132350 on Build GRCh38
rs749385453 Chr.1: 89132325-89132325 on Build GRCh38
rs777165739 Chr.1: 89132454-89132454 on Build GRCh38
rs748416922 Chr.1: 89132338-89132338 on Build GRCh38
rs759066030 Chr.1: 89132360-89132360 on Build GRCh38
rs147259782 Chr.1: 89132342-89132342 on Build GRCh38
rs373608805 Chr.1: 89132356-89132356 on Build GRCh38
rs773536894 Chr.1: 89132463-89132463 on Build GRCh38
rs770273133 Chr.1: 89132407-89132412 on Build GRCh38
rs771729869 Chr.1: 89132382-89132382 on Build GRCh38
rs112057932 Chr.1: 89132450-89132450 on Build GRCh38
rs777070341 Chr.1: 89132358-89132358 on Build GRCh38
rs1019541743 Chr.1: 89132409-89132409 on Build GRCh38
rs376019280 Chr.1: 89132337-89132337 on Build GRCh38
rs141236032 Chr.1: 89132320-89132320 on Build GRCh38
rs748259642 Chr.1: 89132421-89132421 on Build GRCh38
rs752236823 Chr.1: 89132373-89132373 on Build GRCh38
rs139544379 Chr.1: 89132390-89132390 on Build GRCh38
rs34046813 Chr.1: 89132439-89132440 on Build GRCh38
rs777504872 Chr.1: 89132293-89132293 on Build GRCh38
rs751337694 Chr.1: 89132295-89132295 on Build GRCh38
rs746543380 Chr.1: 89132299-89132300 on Build GRCh38
rs144677260 Chr.1: 89132357-89132357 on Build GRCh38
rs778145039 Chr.1: 89132435-89132435 on Build GRCh38
rs746687041 Chr.1: 89132401-89132401 on Build GRCh38
rs763754284 Chr.1: 89132379-89132379 on Build GRCh38
rs943303770 Chr.1: 89132424-89132424 on Build GRCh38
rs781178171 Chr.1: 89132391-89132391 on Build GRCh38
rs148490720 Chr.1: 89132377-89132377 on Build GRCh38
rs772362115 Chr.1: 89132423-89132423 on Build GRCh38
rs1047972130 Chr.1: 89132464-89132464 on Build GRCh38
rs746321263 Chr.1: 89132455-89132455 on Build GRCh38
rs1049436723 Chr.1: 89132395-89132395 on Build GRCh38
rs757164210 Chr.1: 89132297-89132297 on Build GRCh38
rs747393711 Chr.1: 89132449-89132449 on Build GRCh38
rs200147635 Chr.1: 89132408-89132408 on Build GRCh38
rs138254173 Chr.1: 89132336-89132336 on Build GRCh38
rs768673834 Chr.1: 89132331-89132331 on Build GRCh38

View Full Product Details