Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 92514589-92514816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPP
Entrez Gene ID
401541
Gene Name
centromere protein P
Gene Aliases
CENP-P
UniGene
Hs.713775
Gene Chromosome Location
Chr.9: 92325457-92615164 on Build GRCh38
Transcripts
NM_001012267.2 NM_001286969.1 NM_001197295.1 NM_001197296.1 NM_001393.3 XM_017014376.1 XM_017014377.1

Gene Symbol
ECM2
Entrez Gene ID
1842
Gene Name
extracellular matrix protein 2
Gene Aliases
-
UniGene
Hs.117060
Gene Chromosome Location
Chr.9: 92493547-92536092 on Build GRCh38
Transcripts
NM_001012267.2 NM_001286969.1 NM_001197295.1 NM_001197296.1 NM_001393.3 XM_017014376.1 XM_017014377.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6651431 c.986G>A p.C329Y Chr.9: 92514699-92514699 on Build GRCh38
6651430 c.994G>A p.A332T Chr.9: 92514691-92514691 on Build GRCh38
7365824 c.808C>A p.P270T Chr.9: 92514811-92514811 on Build GRCh38
5876561 c.926C>T p.P309L Chr.9: 92514759-92514759 on Build GRCh38
391370 c.809delC p.P270fs*2 Chr.9: 92514810-92514810 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774142752 Chr.9: 92514681-92514681 on Build GRCh38
rs777841346 Chr.9: 92514749-92514749 on Build GRCh38
rs143789386 Chr.9: 92514777-92514777 on Build GRCh38
rs776302657 Chr.9: 92514695-92514695 on Build GRCh38
rs372997619 Chr.9: 92514789-92514789 on Build GRCh38
rs775195975 Chr.9: 92514651-92514651 on Build GRCh38
rs775294587 Chr.9: 92514691-92514691 on Build GRCh38
rs746772392 Chr.9: 92514769-92514769 on Build GRCh38
rs763693210 Chr.9: 92514694-92514694 on Build GRCh38
rs372699831 Chr.9: 92514638-92514638 on Build GRCh38
rs761287103 Chr.9: 92514724-92514724 on Build GRCh38
rs756258771 Chr.9: 92514745-92514745 on Build GRCh38
rs532812407 Chr.9: 92514730-92514730 on Build GRCh38
rs771907543 Chr.9: 92514688-92514688 on Build GRCh38
rs1012998452 Chr.9: 92514696-92514696 on Build GRCh38
rs41280067 Chr.9: 92514770-92514770 on Build GRCh38
rs758674355 Chr.9: 92514640-92514640 on Build GRCh38
rs138357265 Chr.9: 92514790-92514790 on Build GRCh38
rs201012391 Chr.9: 92514669-92514669 on Build GRCh38
rs755206191 Chr.9: 92514656-92514656 on Build GRCh38
rs148975367 Chr.9: 92514731-92514731 on Build GRCh38
rs770043271 Chr.9: 92514675-92514675 on Build GRCh38
rs142608064 Chr.9: 92514750-92514750 on Build GRCh38
rs749910227 Chr.9: 92514728-92514728 on Build GRCh38
rs756788903 Chr.9: 92514633-92514633 on Build GRCh38
rs952168998 Chr.9: 92514761-92514761 on Build GRCh38
rs560017678 Chr.9: 92514741-92514741 on Build GRCh38
rs760308704 Chr.9: 92514622-92514622 on Build GRCh38
rs147083553 Chr.9: 92514655-92514655 on Build GRCh38
rs534479751 Chr.9: 92514719-92514731 on Build GRCh38
rs138567683 Chr.9: 92514742-92514742 on Build GRCh38
rs781597574 Chr.9: 92514668-92514668 on Build GRCh38
rs764812951 Chr.9: 92514778-92514778 on Build GRCh38
rs752287112 Chr.9: 92514619-92514619 on Build GRCh38
rs765700180 Chr.9: 92514637-92514637 on Build GRCh38
rs775319533 Chr.9: 92514766-92514766 on Build GRCh38
rs776249213 Chr.9: 92514771-92514771 on Build GRCh38
rs763748198 Chr.9: 92514624-92514624 on Build GRCh38
rs964668563 Chr.9: 92514781-92514781 on Build GRCh38
rs753295603 Chr.9: 92514627-92514627 on Build GRCh38
rs764782321 Chr.9: 92514725-92514725 on Build GRCh38
rs760484059 Chr.9: 92514692-92514692 on Build GRCh38

View Full Product Details