Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31727474-31727743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDAH2
Entrez Gene ID
23564
Gene Name
dimethylarginine dimethylaminohydrolase 2
Gene Aliases
DDAH, DDAHII, G6a, HEL-S-277, NG30
UniGene
Hs.247362
Gene Chromosome Location
Chr.6: 31727037-31730265 on Build GRCh38
Transcripts
NM_001303007.1 NM_001303008.1 NM_013974.2 XM_011514448.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7113421 c.667C>T p.L223F Chr.6: 31727617-31727617 on Build GRCh38
291547 c.601G>A p.V201M Chr.6: 31727683-31727683 on Build GRCh38
5529618 c.624C>T p.A208A Chr.6: 31727660-31727660 on Build GRCh38
4902968 c.670C>T p.R224C Chr.6: 31727614-31727614 on Build GRCh38
5671421 c.729C>T p.P243P Chr.6: 31727555-31727555 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11540321 Chr.6: 31727591-31727591 on Build GRCh38
rs765073923 Chr.6: 31727559-31727559 on Build GRCh38
rs188475802 Chr.6: 31727669-31727669 on Build GRCh38
rs773811465 Chr.6: 31727677-31727677 on Build GRCh38
rs374303021 Chr.6: 31727702-31727702 on Build GRCh38
rs939267157 Chr.6: 31727714-31727714 on Build GRCh38
rs539164684 Chr.6: 31727667-31727667 on Build GRCh38
rs774241250 Chr.6: 31727625-31727625 on Build GRCh38
rs142513856 Chr.6: 31727550-31727550 on Build GRCh38
rs1027552517 Chr.6: 31727589-31727589 on Build GRCh38
rs756788367 Chr.6: 31727574-31727574 on Build GRCh38
rs768902016 Chr.6: 31727606-31727606 on Build GRCh38
rs762632383 Chr.6: 31727567-31727567 on Build GRCh38
rs748083507 Chr.6: 31727568-31727568 on Build GRCh38
rs770896162 Chr.6: 31727545-31727545 on Build GRCh38
rs909611209 Chr.6: 31727632-31727632 on Build GRCh38
rs750874237 Chr.6: 31727580-31727580 on Build GRCh38
rs752568510 Chr.6: 31727565-31727565 on Build GRCh38
rs376436685 Chr.6: 31727525-31727525 on Build GRCh38
rs749466786 Chr.6: 31727500-31727500 on Build GRCh38
rs756583073 Chr.6: 31727720-31727720 on Build GRCh38
rs549964070 Chr.6: 31727651-31727651 on Build GRCh38
rs749656173 Chr.6: 31727597-31727597 on Build GRCh38
rs771987270 Chr.6: 31727635-31727635 on Build GRCh38
rs780689374 Chr.6: 31727575-31727575 on Build GRCh38
rs138907940 Chr.6: 31727660-31727660 on Build GRCh38
rs759664191 Chr.6: 31727638-31727638 on Build GRCh38
rs567987180 Chr.6: 31727595-31727595 on Build GRCh38
rs748406821 Chr.6: 31727627-31727627 on Build GRCh38
rs751063270 Chr.6: 31727569-31727569 on Build GRCh38
rs781181032 Chr.6: 31727532-31727532 on Build GRCh38
rs749971034 Chr.6: 31727715-31727715 on Build GRCh38
rs200126204 Chr.6: 31727614-31727614 on Build GRCh38
rs776643748 Chr.6: 31727549-31727549 on Build GRCh38
rs767622416 Chr.6: 31727639-31727639 on Build GRCh38
rs959383272 Chr.6: 31727722-31727722 on Build GRCh38
rs768794217 Chr.6: 31727509-31727509 on Build GRCh38
rs761543179 Chr.6: 31727679-31727679 on Build GRCh38
rs111810350 Chr.6: 31727708-31727708 on Build GRCh38

View Full Product Details