Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 168030686-168030941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF25
Entrez Gene ID
3834
Gene Name
kinesin family member 25
Gene Aliases
KNSL3
UniGene
Hs.150013
Gene Chromosome Location
Chr.6: 168002543-168045106 on Build GRCh38
Transcripts
NM_005355.3 NM_030615.2 XM_011535802.2 XM_011535803.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6489967 c.136G>A p.V46M Chr.6: 168030816-168030816 on Build GRCh38
85018 c.138G>A p.V46V Chr.6: 168030818-168030818 on Build GRCh38
595369 c.122C>T p.A41V Chr.6: 168030802-168030802 on Build GRCh38
1076069 c.133G>T p.D45Y Chr.6: 168030813-168030813 on Build GRCh38
6440576 c.131G>A p.G44E Chr.6: 168030811-168030811 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199956268 Chr.6: 168030802-168030802 on Build GRCh38
rs34981094 Chr.6: 168030818-168030818 on Build GRCh38
rs767006965 Chr.6: 168030819-168030819 on Build GRCh38
rs375805319 Chr.6: 168030858-168030858 on Build GRCh38
rs754246982 Chr.6: 168030762-168030762 on Build GRCh38
rs775332458 Chr.6: 168030803-168030803 on Build GRCh38
rs766112291 Chr.6: 168030830-168030830 on Build GRCh38
rs770979239 Chr.6: 168030761-168030762 on Build GRCh38
rs781373756 Chr.6: 168030821-168030821 on Build GRCh38
rs771795716 Chr.6: 168030814-168030814 on Build GRCh38
rs753197430 Chr.6: 168030839-168030839 on Build GRCh38
rs758419602 Chr.6: 168030850-168030850 on Build GRCh38
rs536226852 Chr.6: 168030869-168030869 on Build GRCh38
rs748215968 Chr.6: 168030786-168030786 on Build GRCh38
rs774245188 Chr.6: 168030816-168030816 on Build GRCh38
rs752770376 Chr.6: 168030705-168030705 on Build GRCh38
rs747786149 Chr.6: 168030726-168030726 on Build GRCh38
rs754299875 Chr.6: 168030831-168030831 on Build GRCh38
rs779540288 Chr.6: 168030782-168030782 on Build GRCh38
rs372985926 Chr.6: 168030760-168030760 on Build GRCh38
rs146495006 Chr.6: 168030793-168030793 on Build GRCh38
rs34049091 Chr.6: 168030801-168030801 on Build GRCh38
rs762723763 Chr.6: 168030809-168030809 on Build GRCh38
rs780497629 Chr.6: 168030752-168030752 on Build GRCh38
rs149411206 Chr.6: 168030795-168030795 on Build GRCh38
rs767119808 Chr.6: 168030724-168030724 on Build GRCh38
rs757534908 Chr.6: 168030876-168030876 on Build GRCh38
rs202244742 Chr.6: 168030823-168030823 on Build GRCh38
rs34344055 Chr.6: 168030779-168030779 on Build GRCh38
rs781369846 Chr.6: 168030887-168030887 on Build GRCh38
rs1056095052 Chr.6: 168030774-168030774 on Build GRCh38
rs750056333 Chr.6: 168030725-168030725 on Build GRCh38

View Full Product Details