Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 67511472-67511719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPA6
Entrez Gene ID
57094
Gene Name
carboxypeptidase A6
Gene Aliases
CPAH, ETL5, FEB11
UniGene
Hs.658850
Gene Chromosome Location
Chr.8: 67422170-67747114 on Build GRCh38
Transcripts
NM_020361.4 XM_011517570.2 XM_017013646.1 XM_017013647.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5874913 c.332A>C p.D111A Chr.8: 67511641-67511641 on Build GRCh38
4859346 c.326T>C p.I109T Chr.8: 67511647-67511647 on Build GRCh38
7362441 c.349G>T p.E117* Chr.8: 67511624-67511624 on Build GRCh38
6292245 c.340A>T p.K114* Chr.8: 67511633-67511633 on Build GRCh38
5584896 c.409G>A p.E137K Chr.8: 67511564-67511564 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754831920 Chr.8: 67511530-67511530 on Build GRCh38
rs985518200 Chr.8: 67511587-67511587 on Build GRCh38
rs200555424 Chr.8: 67511537-67511537 on Build GRCh38
rs751616256 Chr.8: 67511518-67511518 on Build GRCh38
rs751562943 Chr.8: 67511670-67511670 on Build GRCh38
rs781238569 Chr.8: 67511690-67511691 on Build GRCh38
rs139120666 Chr.8: 67511553-67511553 on Build GRCh38
rs760990950 Chr.8: 67511605-67511605 on Build GRCh38
rs781081618 Chr.8: 67511684-67511684 on Build GRCh38
rs777807877 Chr.8: 67511547-67511547 on Build GRCh38
rs776114445 Chr.8: 67511626-67511626 on Build GRCh38
rs377667807 Chr.8: 67511658-67511658 on Build GRCh38
rs369797680 Chr.8: 67511600-67511600 on Build GRCh38
rs769090245 Chr.8: 67511614-67511614 on Build GRCh38
rs1038563125 Chr.8: 67511628-67511628 on Build GRCh38
rs201329180 Chr.8: 67511506-67511506 on Build GRCh38
rs751332117 Chr.8: 67511522-67511522 on Build GRCh38
rs151119622 Chr.8: 67511647-67511647 on Build GRCh38
rs757403448 Chr.8: 67511590-67511590 on Build GRCh38
rs994260236 Chr.8: 67511675-67511675 on Build GRCh38
rs780154258 Chr.8: 67511507-67511507 on Build GRCh38
rs886063081 Chr.8: 67511655-67511655 on Build GRCh38
rs958262379 Chr.8: 67511546-67511546 on Build GRCh38
rs199809258 Chr.8: 67511554-67511554 on Build GRCh38
rs918736085 Chr.8: 67511673-67511673 on Build GRCh38
rs762556121 Chr.8: 67511651-67511651 on Build GRCh38
rs754003057 Chr.8: 67511690-67511690 on Build GRCh38
rs886044613 Chr.8: 67511606-67511606 on Build GRCh38
rs1005535087 Chr.8: 67511601-67511601 on Build GRCh38
rs557186136 Chr.8: 67511574-67511574 on Build GRCh38
rs764635474 Chr.8: 67511641-67511641 on Build GRCh38
rs911302349 Chr.8: 67511598-67511598 on Build GRCh38
rs757557217 Chr.8: 67511691-67511691 on Build GRCh38
rs754708382 Chr.8: 67511674-67511674 on Build GRCh38
rs201966963 Chr.8: 67511637-67511637 on Build GRCh38
rs200114239 Chr.8: 67511599-67511599 on Build GRCh38
rs776473519 Chr.8: 67511611-67511611 on Build GRCh38
rs746164595 Chr.8: 67511693-67511694 on Build GRCh38
rs768188762 Chr.8: 67511591-67511591 on Build GRCh38
rs746521547 Chr.8: 67511576-67511576 on Build GRCh38
rs779117472 Chr.8: 67511590-67511590 on Build GRCh38

View Full Product Details