Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184164216-184164489 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DVL3
Entrez Gene ID
1857
Gene Name
dishevelled segment polarity protein 3
Gene Aliases
DRS3
UniGene
Hs.388116
Gene Chromosome Location
Chr.3: 184155311-184173614 on Build GRCh38
Transcripts
NM_004423.3 XM_005247172.2 XM_011512513.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5864011 c.286C>T p.P96S Chr.3: 184164321-184164321 on Build GRCh38
4115656 c.343C>A p.P115T Chr.3: 184164378-184164378 on Build GRCh38
6650474 c.312G>A p.E104E Chr.3: 184164347-184164347 on Build GRCh38
131315 c.281A>T p.D94V Chr.3: 184164316-184164316 on Build GRCh38
5398744 c.268C>T p.P90S Chr.3: 184164303-184164303 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs934221378 Chr.3: 184164348-184164348 on Build GRCh38
rs1014702318 Chr.3: 184164462-184164462 on Build GRCh38
rs370231258 Chr.3: 184164391-184164391 on Build GRCh38
rs771100860 Chr.3: 184164332-184164332 on Build GRCh38
rs779260249 Chr.3: 184164443-184164443 on Build GRCh38
rs563100903 Chr.3: 184164417-184164417 on Build GRCh38
rs200551245 Chr.3: 184164436-184164436 on Build GRCh38
rs778291939 Chr.3: 184164258-184164258 on Build GRCh38
rs1006370814 Chr.3: 184164441-184164441 on Build GRCh38
rs754780569 Chr.3: 184164407-184164407 on Build GRCh38
rs750073787 Chr.3: 184164302-184164302 on Build GRCh38
rs185415845 Chr.3: 184164418-184164418 on Build GRCh38
rs772666279 Chr.3: 184164464-184164464 on Build GRCh38
rs776921430 Chr.3: 184164449-184164449 on Build GRCh38
rs984215644 Chr.3: 184164250-184164251 on Build GRCh38
rs753241787 Chr.3: 184164251-184164251 on Build GRCh38
rs756458717 Chr.3: 184164352-184164352 on Build GRCh38
rs746071512 Chr.3: 184164338-184164338 on Build GRCh38
rs779764964 Chr.3: 184164314-184164314 on Build GRCh38
rs541068803 Chr.3: 184164290-184164290 on Build GRCh38
rs373066030 Chr.3: 184164395-184164395 on Build GRCh38
rs34900840 Chr.3: 184164443-184164444 on Build GRCh38
rs545608557 Chr.3: 184164305-184164305 on Build GRCh38
rs777165493 Chr.3: 184164363-184164363 on Build GRCh38
rs377048880 Chr.3: 184164465-184164465 on Build GRCh38
rs147153174 Chr.3: 184164320-184164320 on Build GRCh38
rs529577038 Chr.3: 184164371-184164371 on Build GRCh38
rs762252652 Chr.3: 184164451-184164451 on Build GRCh38
rs756773300 Chr.3: 184164256-184164256 on Build GRCh38
rs150288196 Chr.3: 184164326-184164326 on Build GRCh38
rs770288075 Chr.3: 184164460-184164460 on Build GRCh38
rs371894225 Chr.3: 184164420-184164420 on Build GRCh38
rs200959046 Chr.3: 184164246-184164246 on Build GRCh38
rs749950896 Chr.3: 184164377-184164377 on Build GRCh38
rs780884904 Chr.3: 184164409-184164409 on Build GRCh38
rs180953456 Chr.3: 184164373-184164373 on Build GRCh38
rs772430635 Chr.3: 184164340-184164340 on Build GRCh38
rs142016012 Chr.3: 184164341-184164341 on Build GRCh38
rs780300833 Chr.3: 184164424-184164424 on Build GRCh38
rs150659339 Chr.3: 184164362-184164362 on Build GRCh38
rs754830767 Chr.3: 184164321-184164321 on Build GRCh38
rs373870236 Chr.3: 184164335-184164335 on Build GRCh38
rs751262267 Chr.3: 184164400-184164400 on Build GRCh38
rs985707313 Chr.3: 184164438-184164438 on Build GRCh38
rs112889179 Chr.3: 184164265-184164265 on Build GRCh38
rs201100826 Chr.3: 184164353-184164353 on Build GRCh38
rs772372510 Chr.3: 184164423-184164423 on Build GRCh38
rs34033424 Chr.3: 184164436-184164436 on Build GRCh38
rs1049906925 Chr.3: 184164349-184164349 on Build GRCh38
rs764768182 Chr.3: 184164269-184164269 on Build GRCh38
rs779075428 Chr.3: 184164419-184164419 on Build GRCh38
rs781210884 Chr.3: 184164325-184164325 on Build GRCh38

View Full Product Details