Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 72334834-72335107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC6B
Entrez Gene ID
23233
Gene Name
exocyst complex component 6B
Gene Aliases
SEC15B, SEC15L2
UniGene
Hs.303454
Gene Chromosome Location
Chr.2: 72175984-72826042 on Build GRCh38
Transcripts
NM_001321729.1 NM_001321730.1 NM_001321731.1 NM_001321733.1 NM_001321734.1 NM_015189.2 NR_135773.1 NR_135774.1 XM_005264224.1 XM_017003641.1 XM_017003642.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6657213 c.2125T>C p.F709L Chr.2: 72335018-72335018 on Build GRCh38
4095530 c.2166G>A p.T722T Chr.2: 72334977-72334977 on Build GRCh38
6092388 c.782A>T p.Q261L Chr.2: 72334972-72334972 on Build GRCh38
6158931 c.792C>A p.F264L Chr.2: 72334962-72334962 on Build GRCh38
145382 c.748G>T p.G250C Chr.2: 72335006-72335006 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs911392305 Chr.2: 72334872-72334872 on Build GRCh38
rs765695924 Chr.2: 72335054-72335057 on Build GRCh38
rs760093971 Chr.2: 72335049-72335049 on Build GRCh38
rs868146697 Chr.2: 72335014-72335014 on Build GRCh38
rs566348239 Chr.2: 72335032-72335033 on Build GRCh38
rs1021905790 Chr.2: 72334875-72334875 on Build GRCh38
rs746903471 Chr.2: 72334956-72334956 on Build GRCh38
rs774475676 Chr.2: 72334945-72334945 on Build GRCh38
rs577958359 Chr.2: 72334927-72334927 on Build GRCh38
rs749516136 Chr.2: 72334922-72334922 on Build GRCh38
rs191567398 Chr.2: 72334992-72334992 on Build GRCh38
rs763633272 Chr.2: 72334928-72334928 on Build GRCh38
rs770505617 Chr.2: 72335001-72335001 on Build GRCh38
rs768719624 Chr.2: 72334938-72334938 on Build GRCh38
rs960571675 Chr.2: 72335024-72335024 on Build GRCh38
rs375284555 Chr.2: 72334977-72334977 on Build GRCh38
rs762335754 Chr.2: 72335041-72335042 on Build GRCh38
rs776467599 Chr.2: 72334974-72334974 on Build GRCh38
rs371853759 Chr.2: 72335071-72335071 on Build GRCh38
rs376972688 Chr.2: 72335082-72335082 on Build GRCh38
rs183996739 Chr.2: 72335048-72335048 on Build GRCh38
rs775955093 Chr.2: 72335002-72335002 on Build GRCh38
rs780193412 Chr.2: 72334906-72334906 on Build GRCh38
rs768021254 Chr.2: 72335058-72335058 on Build GRCh38
rs775212969 Chr.2: 72334997-72334997 on Build GRCh38
rs969696307 Chr.2: 72335085-72335085 on Build GRCh38
rs562975214 Chr.2: 72334886-72334886 on Build GRCh38
rs111624023 Chr.2: 72334972-72334972 on Build GRCh38
rs763683533 Chr.2: 72335007-72335007 on Build GRCh38

View Full Product Details