Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 54172820-54173091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP24A1
Entrez Gene ID
1591
Gene Name
cytochrome P450 family 24 subfamily A member 1
Gene Aliases
CP24, CYP24, HCAI, HCINF1, P450-CC24
UniGene
Hs.89663
Gene Chromosome Location
Chr.20: 54145731-54173985 on Build GRCh38
Transcripts
NM_000782.4 NM_001128915.1 XM_005260304.4 XM_017027691.1 XM_017027692.1 XM_017027693.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5617027 c.399G>T p.P133P Chr.20: 54172959-54172959 on Build GRCh38
4099607 c.369C>T p.S123S Chr.20: 54172989-54172989 on Build GRCh38
6231332 c.304G>A p.G102S Chr.20: 54173054-54173054 on Build GRCh38
1238632 c.439C>T p.L147L Chr.20: 54172919-54172919 on Build GRCh38
6586610 c.393C>T p.I131I Chr.20: 54172965-54172965 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751690053 Chr.20: 54172950-54172950 on Build GRCh38
rs775180997 Chr.20: 54172983-54172983 on Build GRCh38
rs963093306 Chr.20: 54172889-54172889 on Build GRCh38
rs759176351 Chr.20: 54172927-54172927 on Build GRCh38
rs763297732 Chr.20: 54172996-54172996 on Build GRCh38
rs774042537 Chr.20: 54172919-54172919 on Build GRCh38
rs1004642642 Chr.20: 54172885-54172885 on Build GRCh38
rs755278180 Chr.20: 54173001-54173001 on Build GRCh38
rs780430506 Chr.20: 54172947-54172947 on Build GRCh38
rs764570572 Chr.20: 54172936-54172936 on Build GRCh38
rs200891772 Chr.20: 54172961-54172961 on Build GRCh38
rs778708906 Chr.20: 54172976-54172976 on Build GRCh38
rs148084028 Chr.20: 54172982-54172982 on Build GRCh38
rs993130538 Chr.20: 54173002-54173002 on Build GRCh38
rs370328266 Chr.20: 54172929-54172929 on Build GRCh38
rs773653979 Chr.20: 54173038-54173038 on Build GRCh38
rs750777104 Chr.20: 54172946-54172946 on Build GRCh38
rs147642444 Chr.20: 54173045-54173045 on Build GRCh38
rs776150853 Chr.20: 54173035-54173035 on Build GRCh38
rs747100687 Chr.20: 54172902-54172902 on Build GRCh38
rs760156921 Chr.20: 54172988-54172988 on Build GRCh38
rs770793591 Chr.20: 54172975-54172975 on Build GRCh38
rs777927687 Chr.20: 54172908-54172908 on Build GRCh38
rs368019359 Chr.20: 54172953-54172953 on Build GRCh38
rs746144236 Chr.20: 54172879-54172879 on Build GRCh38
rs768320804 Chr.20: 54173034-54173034 on Build GRCh38
rs368218305 Chr.20: 54173053-54173053 on Build GRCh38
rs769791940 Chr.20: 54172903-54172903 on Build GRCh38
rs1015982035 Chr.20: 54172887-54172887 on Build GRCh38
rs770795964 Chr.20: 54172918-54172918 on Build GRCh38
rs886056789 Chr.20: 54173063-54173063 on Build GRCh38
rs777676129 Chr.20: 54172928-54172930 on Build GRCh38
rs765598689 Chr.20: 54172939-54172939 on Build GRCh38
rs750735792 Chr.20: 54172995-54172995 on Build GRCh38
rs942436487 Chr.20: 54173042-54173051 on Build GRCh38
rs145275834 Chr.20: 54173007-54173007 on Build GRCh38
rs757149824 Chr.20: 54173017-54173017 on Build GRCh38
rs758238180 Chr.20: 54173025-54173025 on Build GRCh38
rs766805229 Chr.20: 54173057-54173057 on Build GRCh38
rs745661961 Chr.20: 54173021-54173021 on Build GRCh38
rs139763321 Chr.20: 54172915-54172915 on Build GRCh38
rs746625209 Chr.20: 54173032-54173032 on Build GRCh38
rs374094543 Chr.20: 54172930-54172930 on Build GRCh38
rs867031815 Chr.20: 54173040-54173040 on Build GRCh38
rs149806586 Chr.20: 54172973-54172973 on Build GRCh38
rs918512715 Chr.20: 54173019-54173019 on Build GRCh38
rs114334215 Chr.20: 54172989-54172989 on Build GRCh38
rs114476330 Chr.20: 54172999-54172999 on Build GRCh38
rs549216457 Chr.20: 54173052-54173052 on Build GRCh38
rs371481742 Chr.20: 54172990-54172990 on Build GRCh38
rs149417481 Chr.20: 54172856-54172856 on Build GRCh38
rs376248780 Chr.20: 54172891-54172891 on Build GRCh38
rs59262760 Chr.20: 54172878-54172878 on Build GRCh38
rs780236119 Chr.20: 54172899-54172899 on Build GRCh38
rs771581144 Chr.20: 54172928-54172928 on Build GRCh38
rs762334108 Chr.20: 54172994-54172994 on Build GRCh38
rs757699874 Chr.20: 54172867-54172867 on Build GRCh38
rs748676671 Chr.20: 54173062-54173062 on Build GRCh38
rs116104487 Chr.20: 54172980-54172980 on Build GRCh38
rs760244872 Chr.20: 54172933-54172933 on Build GRCh38
rs762052884 Chr.20: 54172937-54172937 on Build GRCh38

View Full Product Details