Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1685352-1685625 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HN1L
Entrez Gene ID
90861
Gene Name
hematological and neurological expressed 1-like
Gene Aliases
C16orf34, L11
UniGene
Hs.513261
Gene Chromosome Location
Chr.16: 1678277-1702072 on Build GRCh38
Transcripts
NM_144570.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
471379 c.143T>G p.F48C Chr.16: 1685537-1685537 on Build GRCh38
3506512 c.86G>A p.G29E Chr.16: 1685480-1685480 on Build GRCh38
4382778 c.130G>A p.A44T Chr.16: 1685524-1685524 on Build GRCh38
1478560 c.136A>G p.N46D Chr.16: 1685530-1685530 on Build GRCh38
967642 c.179G>T p.R60M Chr.16: 1685573-1685573 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778368450 Chr.16: 1685551-1685551 on Build GRCh38
rs747693820 Chr.16: 1685559-1685559 on Build GRCh38
rs747498016 Chr.16: 1685420-1685420 on Build GRCh38
rs987887233 Chr.16: 1685473-1685473 on Build GRCh38
rs779782264 Chr.16: 1685390-1685390 on Build GRCh38
rs140567985 Chr.16: 1685526-1685526 on Build GRCh38
rs760707336 Chr.16: 1685479-1685479 on Build GRCh38
rs905458693 Chr.16: 1685400-1685400 on Build GRCh38
rs745829023 Chr.16: 1685581-1685581 on Build GRCh38
rs201099964 Chr.16: 1685393-1685393 on Build GRCh38
rs771473482 Chr.16: 1685426-1685426 on Build GRCh38
rs753528213 Chr.16: 1685540-1685540 on Build GRCh38
rs111647888 Chr.16: 1685574-1685574 on Build GRCh38
rs567359396 Chr.16: 1685567-1685567 on Build GRCh38
rs755954033 Chr.16: 1685517-1685517 on Build GRCh38
rs371303987 Chr.16: 1685452-1685452 on Build GRCh38
rs34135815 Chr.16: 1685487-1685487 on Build GRCh38
rs376078833 Chr.16: 1685406-1685406 on Build GRCh38
rs868475727 Chr.16: 1685504-1685505 on Build GRCh38
rs546212158 Chr.16: 1685448-1685448 on Build GRCh38
rs749499308 Chr.16: 1685588-1685588 on Build GRCh38
rs770199129 Chr.16: 1685575-1685575 on Build GRCh38
rs775793002 Chr.16: 1685466-1685466 on Build GRCh38
rs764211542 Chr.16: 1685505-1685506 on Build GRCh38
rs750138673 Chr.16: 1685497-1685497 on Build GRCh38
rs767595374 Chr.16: 1685494-1685494 on Build GRCh38
rs913556945 Chr.16: 1685474-1685474 on Build GRCh38
rs751739235 Chr.16: 1685471-1685471 on Build GRCh38
rs757029446 Chr.16: 1685409-1685409 on Build GRCh38
rs754679163 Chr.16: 1685547-1685547 on Build GRCh38
rs764264367 Chr.16: 1685470-1685470 on Build GRCh38
rs1014006615 Chr.16: 1685376-1685376 on Build GRCh38
rs190193348 Chr.16: 1685402-1685402 on Build GRCh38
rs909750885 Chr.16: 1685563-1685563 on Build GRCh38
rs762874427 Chr.16: 1685468-1685468 on Build GRCh38
rs976340527 Chr.16: 1685530-1685530 on Build GRCh38
rs560011266 Chr.16: 1685465-1685465 on Build GRCh38
rs781732934 Chr.16: 1685569-1685569 on Build GRCh38
rs921166624 Chr.16: 1685542-1685542 on Build GRCh38
rs748196897 Chr.16: 1685410-1685410 on Build GRCh38
rs576721552 Chr.16: 1685381-1685382 on Build GRCh38
rs775783585 Chr.16: 1685576-1685576 on Build GRCh38

View Full Product Details