Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 121993058-121993311 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ILDR1
Entrez Gene ID
286676
Gene Name
immunoglobulin like domain containing receptor 1
Gene Aliases
DFNB42, ILDR1alpha, ILDR1alpha', ILDR1beta
UniGene
Hs.98484
Gene Chromosome Location
Chr.3: 121987323-122060554 on Build GRCh38
Transcripts
NM_001199799.1 NM_001199800.1 NM_175924.3 XM_005247389.4 XM_011512738.2 XM_011512739.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3586107 c.1518G>A p.E506E Chr.3: 121993231-121993231 on Build GRCh38
5430826 c.1392C>A p.R464R Chr.3: 121993261-121993261 on Build GRCh38
3586101 c.1572G>A p.R524R Chr.3: 121993177-121993177 on Build GRCh38
3586102 c.1440G>A p.R480R Chr.3: 121993177-121993177 on Build GRCh38
1693297 c.1370C>T p.S457F Chr.3: 121993247-121993247 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745614639 Chr.3: 121993242-121993242 on Build GRCh38
rs919839015 Chr.3: 121993173-121993173 on Build GRCh38
rs756634716 Chr.3: 121993218-121993218 on Build GRCh38
rs757845401 Chr.3: 121993153-121993153 on Build GRCh38
rs142243054 Chr.3: 121993168-121993168 on Build GRCh38
rs774472194 Chr.3: 121993247-121993247 on Build GRCh38
rs374884043 Chr.3: 121993223-121993223 on Build GRCh38
rs189004426 Chr.3: 121993288-121993288 on Build GRCh38
rs866906195 Chr.3: 121993230-121993230 on Build GRCh38
rs912861659 Chr.3: 121993246-121993246 on Build GRCh38
rs199802916 Chr.3: 121993144-121993144 on Build GRCh38
rs367565247 Chr.3: 121993258-121993258 on Build GRCh38
rs781753189 Chr.3: 121993154-121993154 on Build GRCh38
rs773155662 Chr.3: 121993183-121993183 on Build GRCh38
rs727503095 Chr.3: 121993167-121993167 on Build GRCh38
rs149884253 Chr.3: 121993222-121993222 on Build GRCh38
rs748441415 Chr.3: 121993284-121993284 on Build GRCh38
rs201639358 Chr.3: 121993249-121993249 on Build GRCh38
rs149188951 Chr.3: 121993271-121993271 on Build GRCh38
rs773467432 Chr.3: 121993289-121993289 on Build GRCh38
rs773678744 Chr.3: 121993104-121993104 on Build GRCh38
rs763291150 Chr.3: 121993203-121993203 on Build GRCh38
rs144649620 Chr.3: 121993252-121993252 on Build GRCh38
rs368881679 Chr.3: 121993147-121993147 on Build GRCh38
rs769485101 Chr.3: 121993273-121993273 on Build GRCh38
rs767668911 Chr.3: 121993212-121993212 on Build GRCh38
rs748215825 Chr.3: 121993260-121993260 on Build GRCh38
rs771335457 Chr.3: 121993264-121993264 on Build GRCh38
rs770839837 Chr.3: 121993238-121993238 on Build GRCh38
rs146620043 Chr.3: 121993259-121993259 on Build GRCh38
rs371286984 Chr.3: 121993260-121993260 on Build GRCh38
rs747787444 Chr.3: 121993232-121993232 on Build GRCh38
rs746514835 Chr.3: 121993265-121993265 on Build GRCh38
rs748280061 Chr.3: 121993171-121993171 on Build GRCh38
rs781450878 Chr.3: 121993257-121993258 on Build GRCh38
rs766113693 Chr.3: 121993134-121993134 on Build GRCh38
rs764244695 Chr.3: 121993150-121993150 on Build GRCh38
rs766963403 Chr.3: 121993257-121993257 on Build GRCh38
rs200077106 Chr.3: 121993233-121993233 on Build GRCh38
rs745654608 Chr.3: 121993168-121993168 on Build GRCh38
rs34284625 Chr.3: 121993251-121993251 on Build GRCh38
rs747580467 Chr.3: 121993176-121993176 on Build GRCh38
rs879369909 Chr.3: 121993163-121993163 on Build GRCh38
rs555873964 Chr.3: 121993170-121993170 on Build GRCh38
rs772095981 Chr.3: 121993178-121993178 on Build GRCh38
rs770715527 Chr.3: 121993191-121993191 on Build GRCh38
rs751697424 Chr.3: 121993210-121993210 on Build GRCh38
rs752946322 Chr.3: 121993138-121993138 on Build GRCh38
rs201511787 Chr.3: 121993155-121993155 on Build GRCh38
rs960093595 Chr.3: 121993139-121993139 on Build GRCh38
rs761772837 Chr.3: 121993211-121993211 on Build GRCh38
rs777321422 Chr.3: 121993263-121993263 on Build GRCh38
rs2877561 Chr.3: 121993204-121993204 on Build GRCh38
rs556882881 Chr.3: 121993272-121993272 on Build GRCh38
rs374231194 Chr.3: 121993105-121993105 on Build GRCh38
rs775302314 Chr.3: 121993276-121993276 on Build GRCh38
rs778369601 Chr.3: 121993245-121993245 on Build GRCh38
rs757986659 Chr.3: 121993262-121993262 on Build GRCh38
rs776350294 Chr.3: 121993192-121993192 on Build GRCh38
rs773031345 Chr.3: 121993115-121993115 on Build GRCh38
rs146413045 Chr.3: 121993241-121993241 on Build GRCh38
rs755248446 Chr.3: 121993244-121993244 on Build GRCh38
rs778553516 Chr.3: 121993172-121993172 on Build GRCh38
rs761333724 Chr.3: 121993253-121993253 on Build GRCh38
rs757224204 Chr.3: 121993268-121993268 on Build GRCh38
rs369318229 Chr.3: 121993189-121993189 on Build GRCh38
rs746959964 Chr.3: 121993234-121993234 on Build GRCh38
rs151225236 Chr.3: 121993174-121993174 on Build GRCh38
rs751575285 Chr.3: 121993151-121993151 on Build GRCh38
rs181264416 Chr.3: 121993084-121993084 on Build GRCh38
rs778409709 Chr.3: 121993225-121993225 on Build GRCh38
rs771901142 Chr.3: 121993113-121993113 on Build GRCh38

View Full Product Details