Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81688631-81688874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HGS
Entrez Gene ID
9146
Gene Name
hepatocyte growth factor-regulated tyrosine kinase substrate
Gene Aliases
HRS
UniGene
Hs.661056
Gene Chromosome Location
Chr.17: 81683932-81702121 on Build GRCh38
Transcripts
NM_004712.4 XM_011525463.2 XM_017025297.1 XM_017025298.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6867356 c.313C>A p.R105S Chr.17: 81688725-81688725 on Build GRCh38
985904 c.342C>T p.A114A Chr.17: 81688754-81688754 on Build GRCh38
7179663 c.324C>G p.I108M Chr.17: 81688736-81688736 on Build GRCh38
1387337 c.347C>T p.A116V Chr.17: 81688759-81688759 on Build GRCh38
7425938 c.296A>C p.Q99P Chr.17: 81688708-81688708 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142636777 Chr.17: 81688775-81688775 on Build GRCh38
rs147708106 Chr.17: 81688684-81688684 on Build GRCh38
rs374543980 Chr.17: 81688715-81688715 on Build GRCh38
rs764845907 Chr.17: 81688698-81688698 on Build GRCh38
rs745647846 Chr.17: 81688841-81688851 on Build GRCh38
rs773683815 Chr.17: 81688790-81688790 on Build GRCh38
rs528097559 Chr.17: 81688687-81688687 on Build GRCh38
rs370688379 Chr.17: 81688659-81688659 on Build GRCh38
rs763927305 Chr.17: 81688709-81688709 on Build GRCh38
rs367575782 Chr.17: 81688737-81688737 on Build GRCh38
rs749457767 Chr.17: 81688772-81688772 on Build GRCh38
rs761259276 Chr.17: 81688652-81688652 on Build GRCh38
rs58432187 Chr.17: 81688671-81688671 on Build GRCh38
rs774753883 Chr.17: 81688690-81688690 on Build GRCh38
rs779178497 Chr.17: 81688788-81688788 on Build GRCh38
rs767586999 Chr.17: 81688726-81688726 on Build GRCh38
rs780115266 Chr.17: 81688683-81688683 on Build GRCh38
rs758987034 Chr.17: 81688696-81688696 on Build GRCh38
rs769176696 Chr.17: 81688814-81688814 on Build GRCh38
rs61156089 Chr.17: 81688661-81688661 on Build GRCh38
rs781354351 Chr.17: 81688677-81688677 on Build GRCh38
rs147753915 Chr.17: 81688721-81688721 on Build GRCh38
rs762728512 Chr.17: 81688705-81688705 on Build GRCh38
rs745977457 Chr.17: 81688679-81688679 on Build GRCh38
rs757274153 Chr.17: 81688675-81688675 on Build GRCh38
rs772517024 Chr.17: 81688789-81688789 on Build GRCh38
rs780245369 Chr.17: 81688761-81688761 on Build GRCh38
rs1027272769 Chr.17: 81688686-81688686 on Build GRCh38
rs371451697 Chr.17: 81688742-81688742 on Build GRCh38
rs376034736 Chr.17: 81688834-81688834 on Build GRCh38
rs368611553 Chr.17: 81688674-81688674 on Build GRCh38
rs756312239 Chr.17: 81688682-81688682 on Build GRCh38
rs925370066 Chr.17: 81688651-81688651 on Build GRCh38
rs372376003 Chr.17: 81688694-81688694 on Build GRCh38
rs949600050 Chr.17: 81688704-81688704 on Build GRCh38
rs768898450 Chr.17: 81688685-81688685 on Build GRCh38
rs754943325 Chr.17: 81688745-81688745 on Build GRCh38
rs751405607 Chr.17: 81688716-81688716 on Build GRCh38
rs1055975556 Chr.17: 81688808-81688808 on Build GRCh38
rs750436289 Chr.17: 81688734-81688734 on Build GRCh38
rs199674528 Chr.17: 81688802-81688802 on Build GRCh38
rs764826895 Chr.17: 81688658-81688658 on Build GRCh38
rs764020724 Chr.17: 81688670-81688670 on Build GRCh38
rs757220910 Chr.17: 81688725-81688725 on Build GRCh38
rs768316762 Chr.17: 81688760-81688760 on Build GRCh38

View Full Product Details