Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219374790-219375063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNPEP
Entrez Gene ID
23549
Gene Name
aspartyl aminopeptidase
Gene Aliases
ASPEP, DAP
UniGene
Hs.258551
Gene Chromosome Location
Chr.2: 219372029-219400022 on Build GRCh38
Transcripts
NM_001319116.1 NM_001319117.1 NM_001319118.1 NM_001319119.1 NM_001319120.1 NM_001319121.1 NM_001319122.1 NM_012100.3 NR_134970.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3577972 c.1386C>T p.L462L Chr.2: 219374876-219374876 on Build GRCh38
6296097 c.1348C>A p.H450N Chr.2: 219374914-219374914 on Build GRCh38
476950 c.1249G>T p.V417F Chr.2: 219375013-219375013 on Build GRCh38
3577973 c.1263C>T p.T421T Chr.2: 219374999-219374999 on Build GRCh38
244107 c.1288A>G p.I430V Chr.2: 219374974-219374974 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770374331 Chr.2: 219374848-219374848 on Build GRCh38
rs969674682 Chr.2: 219374951-219374951 on Build GRCh38
rs960930067 Chr.2: 219374856-219374856 on Build GRCh38
rs994059748 Chr.2: 219375023-219375023 on Build GRCh38
rs375490557 Chr.2: 219374827-219374827 on Build GRCh38
rs752574382 Chr.2: 219374894-219374894 on Build GRCh38
rs935366120 Chr.2: 219374898-219374898 on Build GRCh38
rs780726561 Chr.2: 219374945-219374945 on Build GRCh38
rs373191349 Chr.2: 219374891-219374891 on Build GRCh38
rs746617109 Chr.2: 219374834-219374834 on Build GRCh38
rs376715559 Chr.2: 219374874-219374874 on Build GRCh38
rs775997334 Chr.2: 219374854-219374854 on Build GRCh38
rs752486593 Chr.2: 219374830-219374830 on Build GRCh38
rs772183526 Chr.2: 219374963-219374963 on Build GRCh38
rs200988234 Chr.2: 219374997-219374997 on Build GRCh38
rs758125703 Chr.2: 219374833-219374833 on Build GRCh38
rs746924170 Chr.2: 219375029-219375029 on Build GRCh38
rs746444691 Chr.2: 219374840-219374840 on Build GRCh38
rs1801950 Chr.2: 219374866-219374866 on Build GRCh38
rs375968327 Chr.2: 219375010-219375010 on Build GRCh38
rs750042262 Chr.2: 219375041-219375041 on Build GRCh38
rs763926345 Chr.2: 219375015-219375015 on Build GRCh38
rs376806983 Chr.2: 219374910-219374910 on Build GRCh38
rs758213714 Chr.2: 219374904-219374904 on Build GRCh38
rs1035596590 Chr.2: 219374843-219374843 on Build GRCh38
rs3731905 Chr.2: 219374839-219374839 on Build GRCh38
rs372394076 Chr.2: 219374976-219374976 on Build GRCh38
rs769179092 Chr.2: 219374948-219374948 on Build GRCh38
rs748443197 Chr.2: 219374950-219374950 on Build GRCh38
rs925673660 Chr.2: 219375003-219375003 on Build GRCh38
rs750460008 Chr.2: 219374879-219374879 on Build GRCh38
rs3731904 Chr.2: 219374812-219374812 on Build GRCh38
rs774743002 Chr.2: 219374863-219374863 on Build GRCh38
rs773419574 Chr.2: 219374964-219374964 on Build GRCh38
rs775439147 Chr.2: 219375009-219375009 on Build GRCh38
rs200051333 Chr.2: 219374985-219374985 on Build GRCh38
rs751350910 Chr.2: 219375024-219375024 on Build GRCh38
rs769089494 Chr.2: 219374857-219374857 on Build GRCh38
rs368253446 Chr.2: 219374829-219374829 on Build GRCh38
rs369820270 Chr.2: 219374917-219374917 on Build GRCh38
rs749769954 Chr.2: 219374947-219374947 on Build GRCh38
rs774307286 Chr.2: 219374855-219374855 on Build GRCh38
rs186512962 Chr.2: 219374905-219374905 on Build GRCh38
rs201087297 Chr.2: 219374924-219374924 on Build GRCh38
rs775322717 Chr.2: 219375030-219375030 on Build GRCh38
rs369409896 Chr.2: 219374949-219374949 on Build GRCh38
rs780003042 Chr.2: 219374918-219374918 on Build GRCh38
rs376780714 Chr.2: 219374928-219374928 on Build GRCh38
rs527605210 Chr.2: 219375038-219375038 on Build GRCh38
rs897009498 Chr.2: 219374946-219374946 on Build GRCh38
rs756845724 Chr.2: 219374916-219374916 on Build GRCh38
rs368993821 Chr.2: 219374881-219374881 on Build GRCh38
rs370272784 Chr.2: 219375036-219375036 on Build GRCh38
rs762092049 Chr.2: 219374871-219374871 on Build GRCh38

View Full Product Details