Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 59464002-59464227 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR135
Entrez Gene ID
64582
Gene Name
G protein-coupled receptor 135
Gene Aliases
HUMNPIIY20
UniGene
Hs.647573
Gene Chromosome Location
Chr.14: 59452773-59465356 on Build GRCh38
Transcripts
NM_022571.5 XM_017021596.1 XM_017021597.1 XM_017021598.1 XM_017021599.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6673689 c.1150C>T p.P384S Chr.14: 59464077-59464077 on Build GRCh38
3496619 c.1219C>T p.R407W Chr.14: 59464008-59464008 on Build GRCh38
4933367 c.1165A>G p.I389V Chr.14: 59464062-59464062 on Build GRCh38
250969 c.1184C>T p.S395L Chr.14: 59464043-59464043 on Build GRCh38
6521536 c.1088A>T p.Q363L Chr.14: 59464139-59464139 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555173053 Chr.14: 59464031-59464031 on Build GRCh38
rs557349018 Chr.14: 59464168-59464168 on Build GRCh38
rs147790202 Chr.14: 59464065-59464065 on Build GRCh38
rs775956103 Chr.14: 59464077-59464077 on Build GRCh38
rs779740853 Chr.14: 59464024-59464024 on Build GRCh38
rs765102007 Chr.14: 59464096-59464096 on Build GRCh38
rs201546415 Chr.14: 59464058-59464058 on Build GRCh38
rs780796096 Chr.14: 59464120-59464120 on Build GRCh38
rs761103485 Chr.14: 59464158-59464158 on Build GRCh38
rs988853928 Chr.14: 59464072-59464072 on Build GRCh38
rs754063556 Chr.14: 59464084-59464084 on Build GRCh38
rs751542766 Chr.14: 59464190-59464190 on Build GRCh38
rs776476442 Chr.14: 59464043-59464043 on Build GRCh38
rs758182115 Chr.14: 59464185-59464185 on Build GRCh38
rs772413471 Chr.14: 59464139-59464139 on Build GRCh38
rs747655724 Chr.14: 59464199-59464199 on Build GRCh38
rs370580579 Chr.14: 59464110-59464110 on Build GRCh38
rs372770365 Chr.14: 59464128-59464128 on Build GRCh38
rs965014120 Chr.14: 59464025-59464025 on Build GRCh38
rs754922190 Chr.14: 59464194-59464194 on Build GRCh38
rs534909955 Chr.14: 59464020-59464020 on Build GRCh38
rs574844181 Chr.14: 59464136-59464136 on Build GRCh38
rs575016635 Chr.14: 59464161-59464161 on Build GRCh38
rs766268712 Chr.14: 59464187-59464187 on Build GRCh38
rs760122894 Chr.14: 59464066-59464066 on Build GRCh38
rs1011978393 Chr.14: 59464040-59464040 on Build GRCh38
rs771854331 Chr.14: 59464023-59464023 on Build GRCh38
rs553178682 Chr.14: 59464135-59464135 on Build GRCh38
rs754598348 Chr.14: 59464112-59464112 on Build GRCh38
rs777367181 Chr.14: 59464074-59464096 on Build GRCh38
rs370248219 Chr.14: 59464183-59464183 on Build GRCh38
rs764155585 Chr.14: 59464080-59464080 on Build GRCh38
rs200868185 Chr.14: 59464071-59464071 on Build GRCh38
rs570954198 Chr.14: 59464022-59464022 on Build GRCh38
rs755630618 Chr.14: 59464200-59464200 on Build GRCh38
rs767952221 Chr.14: 59464076-59464076 on Build GRCh38
rs753681727 Chr.14: 59464131-59464132 on Build GRCh38
rs375777007 Chr.14: 59464166-59464166 on Build GRCh38
rs750143137 Chr.14: 59464099-59464099 on Build GRCh38
rs143521018 Chr.14: 59464198-59464199 on Build GRCh38
rs965622280 Chr.14: 59464091-59464091 on Build GRCh38
rs760803078 Chr.14: 59464078-59464078 on Build GRCh38
rs376196672 Chr.14: 59464105-59464105 on Build GRCh38
rs780884246 Chr.14: 59464195-59464195 on Build GRCh38
rs757363787 Chr.14: 59464089-59464089 on Build GRCh38
rs768255812 Chr.14: 59464041-59464041 on Build GRCh38
rs756813042 Chr.14: 59464177-59464201 on Build GRCh38
rs776044097 Chr.14: 59464157-59464157 on Build GRCh38
rs764171954 Chr.14: 59464165-59464165 on Build GRCh38
rs771343887 Chr.14: 59464060-59464060 on Build GRCh38
rs113897239 Chr.14: 59464200-59464201 on Build GRCh38

View Full Product Details