Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 36991884-36992139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPM2AIP1
Entrez Gene ID
9852
Gene Name
EPM2A interacting protein 1
Gene Aliases
-
UniGene
Hs.28020
Gene Chromosome Location
Chr.3: 36985866-36993304 on Build GRCh38
Transcripts
NM_014805.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7160736 c.1060C>G p.L354V Chr.3: 36992018-36992018 on Build GRCh38
5420734 c.1028G>C p.R343T Chr.3: 36992050-36992050 on Build GRCh38
1043958 c.1062A>G p.L354L Chr.3: 36992016-36992016 on Build GRCh38
1617519 c.1162A>G p.M388V Chr.3: 36991916-36991916 on Build GRCh38
3593039 c.1031G>A p.R344K Chr.3: 36992047-36992047 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs4647201 Chr.3: 36992000-36992000 on Build GRCh38
rs760982762 Chr.3: 36992019-36992019 on Build GRCh38
rs1043950209 Chr.3: 36992076-36992076 on Build GRCh38
rs752692722 Chr.3: 36991995-36991995 on Build GRCh38
rs546128727 Chr.3: 36991919-36991919 on Build GRCh38
rs754083403 Chr.3: 36991954-36991954 on Build GRCh38
rs752602885 Chr.3: 36992092-36992092 on Build GRCh38
rs771175042 Chr.3: 36992097-36992097 on Build GRCh38
rs766437860 Chr.3: 36992091-36992091 on Build GRCh38
rs946938072 Chr.3: 36992046-36992046 on Build GRCh38
rs4647202 Chr.3: 36992081-36992081 on Build GRCh38
rs747584787 Chr.3: 36992027-36992027 on Build GRCh38
rs890066895 Chr.3: 36992035-36992035 on Build GRCh38
rs933664832 Chr.3: 36992026-36992026 on Build GRCh38
rs750770992 Chr.3: 36991953-36991953 on Build GRCh38
rs778457036 Chr.3: 36992016-36992016 on Build GRCh38
rs781130377 Chr.3: 36991977-36991977 on Build GRCh38
rs770313148 Chr.3: 36991912-36991912 on Build GRCh38
rs762176158 Chr.3: 36991955-36991955 on Build GRCh38
rs773323023 Chr.3: 36992089-36992089 on Build GRCh38
rs764609675 Chr.3: 36991951-36991951 on Build GRCh38
rs765401720 Chr.3: 36991960-36991960 on Build GRCh38
rs769121875 Chr.3: 36992029-36992029 on Build GRCh38
rs775865024 Chr.3: 36991931-36991931 on Build GRCh38
rs755900593 Chr.3: 36991999-36991999 on Build GRCh38
rs756881536 Chr.3: 36992009-36992009 on Build GRCh38
rs4647200 Chr.3: 36991971-36991971 on Build GRCh38
rs762116131 Chr.3: 36992071-36992071 on Build GRCh38
rs772590334 Chr.3: 36991921-36991921 on Build GRCh38
rs983941435 Chr.3: 36991950-36991950 on Build GRCh38
rs763078547 Chr.3: 36992090-36992090 on Build GRCh38
rs776125272 Chr.3: 36992024-36992024 on Build GRCh38
rs777089380 Chr.3: 36992066-36992066 on Build GRCh38
rs543497247 Chr.3: 36992110-36992110 on Build GRCh38
rs761061915 Chr.3: 36991934-36991934 on Build GRCh38
rs750476497 Chr.3: 36991961-36991961 on Build GRCh38
rs756091009 Chr.3: 36992109-36992109 on Build GRCh38
rs765434479 Chr.3: 36992083-36992083 on Build GRCh38

View Full Product Details