Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97599950-97600223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOGA1
Entrez Gene ID
112817
Gene Name
4-hydroxy-2-oxoglutarate aldolase 1
Gene Aliases
C10orf65, DHDPS2, DHDPSL, HP3, NPL2
UniGene
Hs.180346
Gene Chromosome Location
Chr.10: 97584345-97612802 on Build GRCh38
Transcripts
NM_001134670.1 NM_138413.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3442434 c.646G>A p.D216N Chr.10: 97600109-97600109 on Build GRCh38
5999160 c.692A>G p.Y231C Chr.10: 97600155-97600155 on Build GRCh38
3442433 c.636C>T p.T212T Chr.10: 97600099-97600099 on Build GRCh38
1162922 c.669G>A p.S223S Chr.10: 97600132-97600132 on Build GRCh38
7261078 c.662C>T p.A221V Chr.10: 97600125-97600125 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs957797762 Chr.10: 97600009-97600009 on Build GRCh38
rs758122412 Chr.10: 97600096-97600096 on Build GRCh38
rs199604936 Chr.10: 97600023-97600023 on Build GRCh38
rs779492256 Chr.10: 97600097-97600097 on Build GRCh38
rs771246710 Chr.10: 97600025-97600025 on Build GRCh38
rs764104379 Chr.10: 97600053-97600053 on Build GRCh38
rs566004348 Chr.10: 97600000-97600000 on Build GRCh38
rs185803104 Chr.10: 97600168-97600168 on Build GRCh38
rs771334502 Chr.10: 97600132-97600132 on Build GRCh38
rs769361123 Chr.10: 97600130-97600131 on Build GRCh38
rs570257729 Chr.10: 97599986-97599986 on Build GRCh38
rs775016553 Chr.10: 97600193-97600201 on Build GRCh38
rs761816106 Chr.10: 97600193-97600193 on Build GRCh38
rs530968432 Chr.10: 97600006-97600006 on Build GRCh38
rs778797375 Chr.10: 97600086-97600086 on Build GRCh38
rs757147223 Chr.10: 97600057-97600057 on Build GRCh38
rs142766113 Chr.10: 97600131-97600131 on Build GRCh38
rs367698800 Chr.10: 97600041-97600041 on Build GRCh38
rs375091787 Chr.10: 97600172-97600172 on Build GRCh38
rs774579806 Chr.10: 97600026-97600026 on Build GRCh38
rs753833846 Chr.10: 97600178-97600178 on Build GRCh38
rs2297644 Chr.10: 97599982-97599982 on Build GRCh38
rs778937599 Chr.10: 97600038-97600038 on Build GRCh38
rs753648051 Chr.10: 97600055-97600055 on Build GRCh38
rs767987177 Chr.10: 97600105-97600105 on Build GRCh38
rs759819900 Chr.10: 97600161-97600161 on Build GRCh38
rs1020790167 Chr.10: 97600033-97600033 on Build GRCh38
rs752816955 Chr.10: 97600037-97600037 on Build GRCh38
rs746545018 Chr.10: 97600098-97600098 on Build GRCh38
rs778377512 Chr.10: 97600129-97600129 on Build GRCh38
rs368607136 Chr.10: 97600035-97600035 on Build GRCh38
rs551638828 Chr.10: 97600034-97600034 on Build GRCh38
rs57616820 Chr.10: 97600170-97600170 on Build GRCh38
rs776660476 Chr.10: 97600029-97600030 on Build GRCh38
rs967028585 Chr.10: 97600060-97600060 on Build GRCh38
rs996534562 Chr.10: 97599979-97599979 on Build GRCh38
rs371213768 Chr.10: 97600036-97600036 on Build GRCh38
rs760812475 Chr.10: 97600171-97600171 on Build GRCh38
rs774667887 Chr.10: 97600139-97600139 on Build GRCh38
rs990830655 Chr.10: 97600165-97600165 on Build GRCh38
rs375002620 Chr.10: 97600040-97600040 on Build GRCh38
rs750145949 Chr.10: 97600094-97600094 on Build GRCh38
rs759146945 Chr.10: 97600110-97600111 on Build GRCh38

View Full Product Details