Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1398621-1398848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAMT
Entrez Gene ID
2593
Gene Name
guanidinoacetate N-methyltransferase
Gene Aliases
CCDS2, HEL-S-20, PIG2, TP53I2
UniGene
Hs.81131
Gene Chromosome Location
Chr.19: 1397026-1401570 on Build GRCh38
Transcripts
NM_000156.5 NM_138924.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5743918 c.755T>C p.V252A Chr.19: 1398731-1398731 on Build GRCh38
3721892 c.715A>G p.I239V Chr.19: 1398771-1398771 on Build GRCh38
297573 c.639delT p.G214fs*12 Chr.19: 1398847-1398847 on Build GRCh38
1716128 c.806C>T p.A269V Chr.19: 1398680-1398680 on Build GRCh38
4642258 c.734C>T p.A245V Chr.19: 1398752-1398752 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550070759 Chr.19: 1398812-1398812 on Build GRCh38
rs927060832 Chr.19: 1398763-1398763 on Build GRCh38
rs772347859 Chr.19: 1398696-1398696 on Build GRCh38
rs544536911 Chr.19: 1398646-1398646 on Build GRCh38
rs74253480 Chr.19: 1398751-1398751 on Build GRCh38
rs556455640 Chr.19: 1398663-1398663 on Build GRCh38
rs527432115 Chr.19: 1398742-1398742 on Build GRCh38
rs752359312 Chr.19: 1398794-1398794 on Build GRCh38
rs928816046 Chr.19: 1398770-1398770 on Build GRCh38
rs552947620 Chr.19: 1398695-1398695 on Build GRCh38
rs978753587 Chr.19: 1398668-1398668 on Build GRCh38
rs1007476162 Chr.19: 1398649-1398649 on Build GRCh38
rs898542520 Chr.19: 1398801-1398801 on Build GRCh38
rs1007140522 Chr.19: 1398659-1398659 on Build GRCh38
rs760856719 Chr.19: 1398719-1398719 on Build GRCh38
rs964821999 Chr.19: 1398667-1398667 on Build GRCh38
rs755678072 Chr.19: 1398808-1398808 on Build GRCh38
rs758431841 Chr.19: 1398752-1398752 on Build GRCh38
rs774446805 Chr.19: 1398689-1398689 on Build GRCh38
rs866783850 Chr.19: 1398797-1398798 on Build GRCh38
rs1019223297 Chr.19: 1398658-1398658 on Build GRCh38
rs777507971 Chr.19: 1398826-1398826 on Build GRCh38
rs368296913 Chr.19: 1398755-1398755 on Build GRCh38
rs764059052 Chr.19: 1398739-1398739 on Build GRCh38
rs1037987815 Chr.19: 1398825-1398825 on Build GRCh38
rs750994286 Chr.19: 1398764-1398764 on Build GRCh38
rs754469954 Chr.19: 1398773-1398773 on Build GRCh38
rs375165823 Chr.19: 1398796-1398796 on Build GRCh38
rs527268568 Chr.19: 1398707-1398707 on Build GRCh38
rs112200521 Chr.19: 1398675-1398675 on Build GRCh38
rs1033032902 Chr.19: 1398673-1398673 on Build GRCh38
rs114229164 Chr.19: 1398783-1398783 on Build GRCh38
rs560198649 Chr.19: 1398726-1398726 on Build GRCh38
rs775770438 Chr.19: 1398705-1398705 on Build GRCh38
rs760445198 Chr.19: 1398735-1398735 on Build GRCh38
rs950206746 Chr.19: 1398753-1398753 on Build GRCh38

View Full Product Details