Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51746452-51746715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FPR1
Entrez Gene ID
2357
Gene Name
formyl peptide receptor 1
Gene Aliases
FMLP, FPR
UniGene
Hs.753
Gene Chromosome Location
Chr.19: 51745770-51751897 on Build GRCh38
Transcripts
NM_001193306.1 NM_002029.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6307392 c.367C>A p.R123S Chr.19: 51746628-51746628 on Build GRCh38
5596744 c.438C>T p.I146I Chr.19: 51746557-51746557 on Build GRCh38
6233101 c.445C>A p.P149T Chr.19: 51746550-51746550 on Build GRCh38
3537887 c.402G>A p.Q134Q Chr.19: 51746593-51746593 on Build GRCh38
6558213 c.478G>A p.V160I Chr.19: 51746517-51746517 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778341723 Chr.19: 51746548-51746548 on Build GRCh38
rs995333430 Chr.19: 51746683-51746683 on Build GRCh38
rs190373580 Chr.19: 51746643-51746643 on Build GRCh38
rs150860114 Chr.19: 51746479-51746479 on Build GRCh38
rs752024794 Chr.19: 51746590-51746590 on Build GRCh38
rs745998725 Chr.19: 51746585-51746585 on Build GRCh38
rs779324740 Chr.19: 51746667-51746667 on Build GRCh38
rs923151832 Chr.19: 51746558-51746558 on Build GRCh38
rs764598916 Chr.19: 51746621-51746622 on Build GRCh38
rs776424230 Chr.19: 51746617-51746617 on Build GRCh38
rs749737014 Chr.19: 51746664-51746664 on Build GRCh38
rs757978120 Chr.19: 51746641-51746641 on Build GRCh38
rs756459168 Chr.19: 51746559-51746559 on Build GRCh38
rs5030879 Chr.19: 51746647-51746647 on Build GRCh38
rs376781635 Chr.19: 51746550-51746550 on Build GRCh38
rs111768566 Chr.19: 51746507-51746507 on Build GRCh38
rs113777346 Chr.19: 51746516-51746516 on Build GRCh38
rs933017940 Chr.19: 51746580-51746580 on Build GRCh38
rs753449716 Chr.19: 51746521-51746521 on Build GRCh38
rs760570197 Chr.19: 51746518-51746518 on Build GRCh38
rs199764870 Chr.19: 51746695-51746695 on Build GRCh38
rs774585820 Chr.19: 51746489-51746489 on Build GRCh38
rs149931707 Chr.19: 51746627-51746627 on Build GRCh38
rs764940294 Chr.19: 51746628-51746628 on Build GRCh38
rs778269740 Chr.19: 51746656-51746656 on Build GRCh38
rs759532140 Chr.19: 51746500-51746500 on Build GRCh38
rs539229723 Chr.19: 51746665-51746665 on Build GRCh38
rs554681504 Chr.19: 51746482-51746482 on Build GRCh38
rs142367736 Chr.19: 51746555-51746555 on Build GRCh38
rs749458452 Chr.19: 51746576-51746576 on Build GRCh38
rs201110935 Chr.19: 51746483-51746483 on Build GRCh38
rs763553366 Chr.19: 51746524-51746524 on Build GRCh38
rs186613919 Chr.19: 51746616-51746616 on Build GRCh38
rs896034471 Chr.19: 51746675-51746675 on Build GRCh38
rs763517926 Chr.19: 51746529-51746529 on Build GRCh38
rs775451540 Chr.19: 51746588-51746588 on Build GRCh38
rs183314714 Chr.19: 51746556-51746556 on Build GRCh38
rs369354920 Chr.19: 51746631-51746631 on Build GRCh38
rs943366709 Chr.19: 51746624-51746624 on Build GRCh38
rs756907682 Chr.19: 51746535-51746535 on Build GRCh38
rs763819355 Chr.19: 51746519-51746519 on Build GRCh38
rs28930680 Chr.19: 51746689-51746689 on Build GRCh38
rs774567273 Chr.19: 51746582-51746582 on Build GRCh38
rs151277630 Chr.19: 51746581-51746581 on Build GRCh38
rs866791532 Chr.19: 51746480-51746480 on Build GRCh38
rs771064181 Chr.19: 51746484-51746484 on Build GRCh38
rs139132326 Chr.19: 51746586-51746586 on Build GRCh38
rs760563008 Chr.19: 51746614-51746614 on Build GRCh38
rs547228145 Chr.19: 51746646-51746646 on Build GRCh38
rs562631943 Chr.19: 51746565-51746565 on Build GRCh38
rs564702916 Chr.19: 51746508-51746508 on Build GRCh38
rs2070745 Chr.19: 51746694-51746694 on Build GRCh38

View Full Product Details