Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 56060850-56061107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL21A1
Entrez Gene ID
81578
Gene Name
collagen type XXI alpha 1 chain
Gene Aliases
COLA1L, FP633
UniGene
Hs.47629
Gene Chromosome Location
Chr.6: 56056590-56394128 on Build GRCh38
Transcripts
NM_001318751.1 NM_001318752.1 NM_001318753.1 NM_001318754.1 NM_030820.3 NR_134849.1 NR_134850.1 NR_134851.1 XM_006715223.1 XM_011514924.2 XM_011514925.2 XM_011514926.1 XM_011514927.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
382420 c.2311G>C p.D771H Chr.6: 56060932-56060932 on Build GRCh38
3745309 c.2301G>A p.G767G Chr.6: 56060942-56060942 on Build GRCh38
4879704 c.2287G>A p.A763T Chr.6: 56060956-56060956 on Build GRCh38
3745308 c.2301G>A p.G767G Chr.6: 56060942-56060942 on Build GRCh38
3697914 c.2293C>A p.P765T Chr.6: 56060950-56060950 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762424677 Chr.6: 56060983-56060983 on Build GRCh38
rs775055829 Chr.6: 56060956-56060956 on Build GRCh38
rs764396706 Chr.6: 56061010-56061010 on Build GRCh38
rs772755740 Chr.6: 56060979-56060979 on Build GRCh38
rs546785825 Chr.6: 56061046-56061046 on Build GRCh38
rs879360696 Chr.6: 56060968-56060968 on Build GRCh38
rs866248635 Chr.6: 56060932-56060932 on Build GRCh38
rs749375204 Chr.6: 56061043-56061043 on Build GRCh38
rs777944369 Chr.6: 56060942-56060942 on Build GRCh38
rs753612825 Chr.6: 56060893-56060893 on Build GRCh38
rs781322450 Chr.6: 56060935-56060935 on Build GRCh38
rs756332631 Chr.6: 56061008-56061008 on Build GRCh38
rs960997927 Chr.6: 56060980-56060980 on Build GRCh38
rs561341748 Chr.6: 56060995-56060995 on Build GRCh38
rs1053452175 Chr.6: 56061055-56061055 on Build GRCh38
rs1027851782 Chr.6: 56060917-56060917 on Build GRCh38
rs943374535 Chr.6: 56060981-56060981 on Build GRCh38
rs76146749 Chr.6: 56060985-56060985 on Build GRCh38
rs779497331 Chr.6: 56061050-56061050 on Build GRCh38
rs777906725 Chr.6: 56061010-56061010 on Build GRCh38
rs374633200 Chr.6: 56061061-56061061 on Build GRCh38
rs545860371 Chr.6: 56060900-56060900 on Build GRCh38
rs368727301 Chr.6: 56060936-56060936 on Build GRCh38
rs761555786 Chr.6: 56060977-56060977 on Build GRCh38
rs774826764 Chr.6: 56060887-56060887 on Build GRCh38
rs760027373 Chr.6: 56060890-56060890 on Build GRCh38
rs375415415 Chr.6: 56060929-56060929 on Build GRCh38
rs761169667 Chr.6: 56060910-56060910 on Build GRCh38
rs764837291 Chr.6: 56060978-56060978 on Build GRCh38
rs746399157 Chr.6: 56061053-56061053 on Build GRCh38
rs373710529 Chr.6: 56060957-56060957 on Build GRCh38
rs369414780 Chr.6: 56061000-56061000 on Build GRCh38
rs750287951 Chr.6: 56060913-56060913 on Build GRCh38
rs749371683 Chr.6: 56060944-56060944 on Build GRCh38
rs200674177 Chr.6: 56060891-56060891 on Build GRCh38
rs751500082 Chr.6: 56060990-56060990 on Build GRCh38
rs763412541 Chr.6: 56060909-56060909 on Build GRCh38
rs772524070 Chr.6: 56061060-56061060 on Build GRCh38
rs201839603 Chr.6: 56060964-56060964 on Build GRCh38
rs368234389 Chr.6: 56060886-56060886 on Build GRCh38
rs994982707 Chr.6: 56060908-56060908 on Build GRCh38
rs934538482 Chr.6: 56061044-56061044 on Build GRCh38
rs990890797 Chr.6: 56060984-56060984 on Build GRCh38
rs772571145 Chr.6: 56060963-56060963 on Build GRCh38
rs375026913 Chr.6: 56061081-56061081 on Build GRCh38
rs972082899 Chr.6: 56060927-56060927 on Build GRCh38
rs371654187 Chr.6: 56060954-56060954 on Build GRCh38
rs750092285 Chr.6: 56061071-56061074 on Build GRCh38
rs754832242 Chr.6: 56060930-56060930 on Build GRCh38
rs191110368 Chr.6: 56060914-56060914 on Build GRCh38
rs747381927 Chr.6: 56061065-56061065 on Build GRCh38
rs867236544 Chr.6: 56060923-56060923 on Build GRCh38
rs868053191 Chr.6: 56060903-56060903 on Build GRCh38
rs9464337 Chr.6: 56061003-56061003 on Build GRCh38
rs752826681 Chr.6: 56061007-56061007 on Build GRCh38
rs769715793 Chr.6: 56060938-56060938 on Build GRCh38

View Full Product Details