Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 89249196-89249467 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPRIN3
Entrez Gene ID
285513
Gene Name
GPRIN family member 3
Gene Aliases
GRIN3
UniGene
Hs.100912
Gene Chromosome Location
Chr.4: 89237666-89308010 on Build GRCh38
Transcripts
NM_198281.2 XM_005262936.3 XM_005262937.4 XM_017008043.1 XM_017008044.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4939139 c.689C>A p.S230Y Chr.4: 89249422-89249422 on Build GRCh38
5939250 c.668G>A p.R223K Chr.4: 89249443-89249443 on Build GRCh38
4920165 c.716C>T p.T239I Chr.4: 89249395-89249395 on Build GRCh38
4126715 c.794C>A p.P265H Chr.4: 89249317-89249317 on Build GRCh38
1285257 c.766G>A p.G256S Chr.4: 89249345-89249345 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896395010 Chr.4: 89249448-89249448 on Build GRCh38
rs187145478 Chr.4: 89249243-89249243 on Build GRCh38
rs759398973 Chr.4: 89249411-89249411 on Build GRCh38
rs769067227 Chr.4: 89249359-89249359 on Build GRCh38
rs774553483 Chr.4: 89249364-89249364 on Build GRCh38
rs754688869 Chr.4: 89249270-89249270 on Build GRCh38
rs928754177 Chr.4: 89249329-89249329 on Build GRCh38
rs775511774 Chr.4: 89249221-89249221 on Build GRCh38
rs17825627 Chr.4: 89249346-89249346 on Build GRCh38
rs758148879 Chr.4: 89249428-89249428 on Build GRCh38
rs1010509170 Chr.4: 89249369-89249369 on Build GRCh38
rs772088938 Chr.4: 89249384-89249384 on Build GRCh38
rs765449217 Chr.4: 89249302-89249303 on Build GRCh38
rs765125086 Chr.4: 89249413-89249413 on Build GRCh38
rs763048759 Chr.4: 89249295-89249295 on Build GRCh38
rs763254226 Chr.4: 89249222-89249222 on Build GRCh38
rs746314788 Chr.4: 89249349-89249349 on Build GRCh38
rs747636181 Chr.4: 89249277-89249277 on Build GRCh38
rs767651932 Chr.4: 89249305-89249305 on Build GRCh38
rs552873132 Chr.4: 89249361-89249361 on Build GRCh38
rs200125932 Chr.4: 89249430-89249430 on Build GRCh38
rs937788863 Chr.4: 89249405-89249405 on Build GRCh38
rs151008108 Chr.4: 89249291-89249291 on Build GRCh38
rs190865809 Chr.4: 89249255-89249255 on Build GRCh38
rs748375047 Chr.4: 89249376-89249376 on Build GRCh38
rs372625903 Chr.4: 89249350-89249350 on Build GRCh38
rs766047507 Chr.4: 89249324-89249324 on Build GRCh38
rs148496084 Chr.4: 89249242-89249242 on Build GRCh38
rs141124181 Chr.4: 89249392-89249392 on Build GRCh38
rs777441239 Chr.4: 89249347-89249347 on Build GRCh38
rs1019160886 Chr.4: 89249230-89249230 on Build GRCh38
rs965871581 Chr.4: 89249235-89249235 on Build GRCh38
rs142220976 Chr.4: 89249300-89249300 on Build GRCh38
rs754866029 Chr.4: 89249334-89249334 on Build GRCh38
rs764965600 Chr.4: 89249335-89249335 on Build GRCh38
rs999299966 Chr.4: 89249218-89249218 on Build GRCh38
rs776486811 Chr.4: 89249402-89249402 on Build GRCh38
rs35079289 Chr.4: 89249275-89249275 on Build GRCh38
rs142702692 Chr.4: 89249287-89249287 on Build GRCh38
rs760707593 Chr.4: 89249393-89249393 on Build GRCh38
rs760682673 Chr.4: 89249307-89249307 on Build GRCh38
rs764113267 Chr.4: 89249229-89249229 on Build GRCh38
rs780469670 Chr.4: 89249354-89249354 on Build GRCh38
rs768836419 Chr.4: 89249299-89249299 on Build GRCh38
rs755857974 Chr.4: 89249253-89249253 on Build GRCh38
rs185199861 Chr.4: 89249306-89249306 on Build GRCh38
rs376391358 Chr.4: 89249357-89249357 on Build GRCh38
rs944996023 Chr.4: 89249312-89249312 on Build GRCh38
rs753380768 Chr.4: 89249262-89249262 on Build GRCh38
rs902189232 Chr.4: 89249434-89249434 on Build GRCh38
rs1034902848 Chr.4: 89249240-89249240 on Build GRCh38
rs752343609 Chr.4: 89249425-89249425 on Build GRCh38
rs937547876 Chr.4: 89249333-89249333 on Build GRCh38
rs761869927 Chr.4: 89249303-89249303 on Build GRCh38
rs751672527 Chr.4: 89249233-89249233 on Build GRCh38
rs570754033 Chr.4: 89249431-89249431 on Build GRCh38
rs1015371733 Chr.4: 89249256-89249256 on Build GRCh38
rs753637474 Chr.4: 89249325-89249325 on Build GRCh38
rs200813010 Chr.4: 89249286-89249286 on Build GRCh38
rs750175055 Chr.4: 89249378-89249378 on Build GRCh38
rs952015054 Chr.4: 89249252-89249252 on Build GRCh38
rs756767970 Chr.4: 89249438-89249438 on Build GRCh38
rs771761398 Chr.4: 89249284-89249284 on Build GRCh38
rs766314211 Chr.4: 89249397-89249397 on Build GRCh38
rs752462257 Chr.4: 89249337-89249337 on Build GRCh38

View Full Product Details