Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 123381436-123381707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND1A
Entrez Gene ID
57706
Gene Name
DENN domain containing 1A
Gene Aliases
FAM31A, KIAA1608
UniGene
Hs.744995
Gene Chromosome Location
Chr.9: 123379654-123930158 on Build GRCh38
Transcripts
NM_020946.1 XM_005252109.2 XM_005252111.4 XM_006717195.3 XM_011518882.2 XM_011518883.2 XM_011518884.2 XM_011518885.2 XM_011518886.2 XM_017014947.1 XM_017014948.1 XM_017014949.1 XM_017014950.1 XM_017014951.1 XM_017014952.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3788168 c.2960C>T p.P987L Chr.9: 123381502-123381502 on Build GRCh38
3779617 c.2937G>A p.Q979Q Chr.9: 123381525-123381525 on Build GRCh38
7364075 c.2756C>A p.S919* Chr.9: 123381706-123381706 on Build GRCh38
7364076 c.2789C>A p.S930* Chr.9: 123381706-123381706 on Build GRCh38
6439709 c.2983G>A p.D995N Chr.9: 123381512-123381512 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757571699 Chr.9: 123381507-123381507 on Build GRCh38
rs779124894 Chr.9: 123381509-123381509 on Build GRCh38
rs911086434 Chr.9: 123381628-123381628 on Build GRCh38
rs142598714 Chr.9: 123381471-123381471 on Build GRCh38
rs762174811 Chr.9: 123381618-123381618 on Build GRCh38
rs781003360 Chr.9: 123381645-123381645 on Build GRCh38
rs758597456 Chr.9: 123381620-123381620 on Build GRCh38
rs758753385 Chr.9: 123381457-123381461 on Build GRCh38
rs746917881 Chr.9: 123381479-123381479 on Build GRCh38
rs955180567 Chr.9: 123381571-123381571 on Build GRCh38
rs781687359 Chr.9: 123381478-123381479 on Build GRCh38
rs769654098 Chr.9: 123381549-123381549 on Build GRCh38
rs754315383 Chr.9: 123381602-123381602 on Build GRCh38
rs373157481 Chr.9: 123381580-123381580 on Build GRCh38
rs376350165 Chr.9: 123381472-123381472 on Build GRCh38
rs764535042 Chr.9: 123381598-123381598 on Build GRCh38
rs1055858477 Chr.9: 123381488-123381488 on Build GRCh38
rs747017882 Chr.9: 123381527-123381527 on Build GRCh38
rs547365850 Chr.9: 123381483-123381483 on Build GRCh38
rs780444178 Chr.9: 123381560-123381562 on Build GRCh38
rs751587146 Chr.9: 123381477-123381478 on Build GRCh38
rs766859439 Chr.9: 123381543-123381543 on Build GRCh38
rs751624191 Chr.9: 123381629-123381629 on Build GRCh38
rs777692695 Chr.9: 123381668-123381668 on Build GRCh38
rs973757286 Chr.9: 123381646-123381646 on Build GRCh38
rs758541150 Chr.9: 123381510-123381510 on Build GRCh38
rs748993355 Chr.9: 123381679-123381679 on Build GRCh38
rs764565454 Chr.9: 123381502-123381502 on Build GRCh38
rs371138017 Chr.9: 123381503-123381503 on Build GRCh38
rs748926539 Chr.9: 123381624-123381625 on Build GRCh38
rs765919260 Chr.9: 123381496-123381496 on Build GRCh38
rs150591766 Chr.9: 123381551-123381551 on Build GRCh38
rs761567995 Chr.9: 123381490-123381490 on Build GRCh38
rs765585771 Chr.9: 123381619-123381619 on Build GRCh38
rs780224992 Chr.9: 123381623-123381623 on Build GRCh38
rs776590199 Chr.9: 123381528-123381528 on Build GRCh38
rs753309845 Chr.9: 123381499-123381499 on Build GRCh38
rs756438059 Chr.9: 123381485-123381490 on Build GRCh38
rs773709278 Chr.9: 123381563-123381563 on Build GRCh38
rs745972611 Chr.9: 123381454-123381454 on Build GRCh38
rs755035286 Chr.9: 123381636-123381636 on Build GRCh38
rs746084954 Chr.9: 123381501-123381501 on Build GRCh38
rs761281711 Chr.9: 123381588-123381588 on Build GRCh38
rs773002960 Chr.9: 123381552-123381552 on Build GRCh38
rs201942010 Chr.9: 123381491-123381491 on Build GRCh38
rs199678734 Chr.9: 123381495-123381495 on Build GRCh38
rs942435778 Chr.9: 123381639-123381639 on Build GRCh38
rs754188975 Chr.9: 123381506-123381506 on Build GRCh38
rs376419805 Chr.9: 123381680-123381680 on Build GRCh38
rs746390585 Chr.9: 123381484-123381485 on Build GRCh38
rs774009185 Chr.9: 123381686-123381686 on Build GRCh38
rs766040661 Chr.9: 123381558-123381558 on Build GRCh38
rs779985177 Chr.9: 123381525-123381525 on Build GRCh38
rs764268652 Chr.9: 123381475-123381476 on Build GRCh38
rs747915223 Chr.9: 123381546-123381546 on Build GRCh38
rs776270301 Chr.9: 123381487-123381487 on Build GRCh38
rs769777771 Chr.9: 123381659-123381659 on Build GRCh38

View Full Product Details