Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 2988650-2988909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRK4
Entrez Gene ID
2868
Gene Name
G protein-coupled receptor kinase 4
Gene Aliases
GPRK2L, GPRK4, GRK4a, IT11
UniGene
Hs.32959
Gene Chromosome Location
Chr.4: 2963504-3040760 on Build GRCh38
Transcripts
NM_001004056.1 NM_001004057.1 NM_005307.2 NM_182982.2 XM_005247962.3 XM_006713880.3 XM_011513447.2 XM_011513448.2 XM_011513449.2 XM_011513450.2 XM_011513451.2 XM_011513452.2 XM_011513453.2 XM_011513454.2 XM_011513455.2 XM_011513456.2 XM_011513457.2 XM_017008052.1 XM_017008053.1 XM_017008054.1 XM_017008055.1 XM_017008056.1 XM_017008057.1 XM_017008058.1 XM_017008059.1 XM_017008060.1 XM_017008061.1 XM_017008062.1 XM_017008063.1 XM_017008064.1 XM_017008065.1 XM_017008066.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
21205 c.123C>G p.T41T Chr.4: 2988797-2988797 on Build GRCh38
6535265 c.166C>G p.L56V Chr.4: 2988744-2988744 on Build GRCh38
3760571 c.194G>T p.R65L Chr.4: 2988772-2988772 on Build GRCh38
1737704 c.261+3G>A p.? Chr.4: 2988842-2988842 on Build GRCh38
382320 c.153_154GG>TT p.K51>? Chr.4: 2988731-2988732 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771478086 Chr.4: 2988685-2988685 on Build GRCh38
rs770235170 Chr.4: 2988693-2988693 on Build GRCh38
rs745441938 Chr.4: 2988684-2988684 on Build GRCh38
rs775718373 Chr.4: 2988820-2988820 on Build GRCh38
rs376621448 Chr.4: 2988853-2988853 on Build GRCh38
rs900204981 Chr.4: 2988735-2988735 on Build GRCh38
rs959207808 Chr.4: 2988748-2988748 on Build GRCh38
rs768758317 Chr.4: 2988834-2988834 on Build GRCh38
rs140178959 Chr.4: 2988760-2988760 on Build GRCh38
rs778190769 Chr.4: 2988798-2988798 on Build GRCh38
rs936911191 Chr.4: 2988829-2988829 on Build GRCh38
rs374998800 Chr.4: 2988720-2988720 on Build GRCh38
rs188171696 Chr.4: 2988860-2988860 on Build GRCh38
rs747895921 Chr.4: 2988801-2988801 on Build GRCh38
rs1037381067 Chr.4: 2988734-2988734 on Build GRCh38
rs777235073 Chr.4: 2988687-2988687 on Build GRCh38
rs753824468 Chr.4: 2988848-2988848 on Build GRCh38
rs749707867 Chr.4: 2988802-2988802 on Build GRCh38
rs748630336 Chr.4: 2988692-2988692 on Build GRCh38
rs774603898 Chr.4: 2988780-2988780 on Build GRCh38
rs761707075 Chr.4: 2988852-2988852 on Build GRCh38
rs369672282 Chr.4: 2988764-2988764 on Build GRCh38
rs990679341 Chr.4: 2988837-2988837 on Build GRCh38
rs763232692 Chr.4: 2988718-2988718 on Build GRCh38
rs137979610 Chr.4: 2988745-2988745 on Build GRCh38
rs764058067 Chr.4: 2988755-2988755 on Build GRCh38
rs752779015 Chr.4: 2988743-2988743 on Build GRCh38
rs902601191 Chr.4: 2988781-2988781 on Build GRCh38
rs756976226 Chr.4: 2988676-2988676 on Build GRCh38
rs778692636 Chr.4: 2988761-2988761 on Build GRCh38
rs555775139 Chr.4: 2988716-2988716 on Build GRCh38
rs370041000 Chr.4: 2988864-2988864 on Build GRCh38
rs773595759 Chr.4: 2988713-2988713 on Build GRCh38
rs142548315 Chr.4: 2988819-2988819 on Build GRCh38
rs778372965 Chr.4: 2988683-2988683 on Build GRCh38
rs753699504 Chr.4: 2988758-2988758 on Build GRCh38
rs144843822 Chr.4: 2988805-2988805 on Build GRCh38
rs142938182 Chr.4: 2988827-2988827 on Build GRCh38
rs770288052 Chr.4: 2988793-2988793 on Build GRCh38
rs372027080 Chr.4: 2988771-2988771 on Build GRCh38
rs774435006 Chr.4: 2988730-2988730 on Build GRCh38
rs759706175 Chr.4: 2988731-2988731 on Build GRCh38
rs767475510 Chr.4: 2988732-2988732 on Build GRCh38
rs1039266109 Chr.4: 2988882-2988882 on Build GRCh38
rs764184921 Chr.4: 2988842-2988842 on Build GRCh38
rs759702586 Chr.4: 2988818-2988818 on Build GRCh38

View Full Product Details