Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49728076-49728307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IP6K1
Entrez Gene ID
9807
Gene Name
inositol hexakisphosphate kinase 1
Gene Aliases
IHPK1, PiUS
UniGene
Hs.386168
Gene Chromosome Location
Chr.3: 49724295-49786540 on Build GRCh38
Transcripts
NM_001006115.2 NM_001242829.1 NM_153273.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5694267 c.745G>T p.E249* Chr.3: 49728150-49728150 on Build GRCh38
1285557 c.707C>T p.A236V Chr.3: 49728188-49728188 on Build GRCh38
271321 c.690G>A p.R230R Chr.3: 49728205-49728205 on Build GRCh38
6686588 c.636C>T p.N212N Chr.3: 49728259-49728259 on Build GRCh38
2850478 c.626T>A p.L209Q Chr.3: 49728269-49728269 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756392946 Chr.3: 49728244-49728244 on Build GRCh38
rs758950455 Chr.3: 49728189-49728189 on Build GRCh38
rs577495317 Chr.3: 49728100-49728100 on Build GRCh38
rs34964130 Chr.3: 49728232-49728232 on Build GRCh38
rs1009022129 Chr.3: 49728254-49728254 on Build GRCh38
rs773336102 Chr.3: 49728115-49728115 on Build GRCh38
rs374592396 Chr.3: 49728196-49728196 on Build GRCh38
rs775142562 Chr.3: 49728159-49728159 on Build GRCh38
rs749222004 Chr.3: 49728123-49728123 on Build GRCh38
rs34675186 Chr.3: 49728124-49728124 on Build GRCh38
rs759330339 Chr.3: 49728151-49728151 on Build GRCh38
rs147379936 Chr.3: 49728190-49728190 on Build GRCh38
rs897856671 Chr.3: 49728277-49728277 on Build GRCh38
rs137918392 Chr.3: 49728259-49728259 on Build GRCh38
rs370259789 Chr.3: 49728167-49728167 on Build GRCh38
rs774114345 Chr.3: 49728130-49728130 on Build GRCh38
rs375536774 Chr.3: 49728199-49728199 on Build GRCh38
rs748386503 Chr.3: 49728208-49728208 on Build GRCh38
rs149764153 Chr.3: 49728112-49728112 on Build GRCh38
rs964116005 Chr.3: 49728148-49728148 on Build GRCh38
rs753436945 Chr.3: 49728188-49728188 on Build GRCh38
rs760260548 Chr.3: 49728187-49728187 on Build GRCh38
rs749419986 Chr.3: 49728262-49728262 on Build GRCh38
rs774305021 Chr.3: 49728282-49728282 on Build GRCh38
rs1054069867 Chr.3: 49728172-49728172 on Build GRCh38
rs771021875 Chr.3: 49728271-49728271 on Build GRCh38
rs375665750 Chr.3: 49728158-49728158 on Build GRCh38
rs781420994 Chr.3: 49728202-49728202 on Build GRCh38
rs151229839 Chr.3: 49728246-49728246 on Build GRCh38

View Full Product Details