Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 61867237-61867510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM19A2
Entrez Gene ID
338811
Gene Name
family with sequence similarity 19 member A2, C-C motif chemokine like
Gene Aliases
TAFA-2, TAFA2
UniGene
Hs.269745
Gene Chromosome Location
Chr.12: 61708248-62192839 on Build GRCh38
Transcripts
NM_178539.4 XM_006719370.2 XM_011538272.1 XM_011538274.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1284838 c.82G>T p.V28L Chr.12: 61867344-61867344 on Build GRCh38
1990551 c.39A>G p.K13K Chr.12: 61867387-61867387 on Build GRCh38
1990550 c.75delG p.V27fs*32 Chr.12: 61867351-61867351 on Build GRCh38
5375751 c.74G>A p.G25E Chr.12: 61867352-61867352 on Build GRCh38
280978 c.21G>T p.Q7H Chr.12: 61867405-61867405 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773048100 Chr.12: 61867343-61867343 on Build GRCh38
rs776010666 Chr.12: 61867328-61867328 on Build GRCh38
rs567891464 Chr.12: 61867264-61867264 on Build GRCh38
rs888897143 Chr.12: 61867452-61867452 on Build GRCh38
rs770963002 Chr.12: 61867313-61867313 on Build GRCh38
rs186809868 Chr.12: 61867405-61867405 on Build GRCh38
rs374262785 Chr.12: 61867468-61867468 on Build GRCh38
rs754766419 Chr.12: 61867274-61867274 on Build GRCh38
rs751468007 Chr.12: 61867361-61867361 on Build GRCh38
rs752482086 Chr.12: 61867378-61867378 on Build GRCh38
rs757319279 Chr.12: 61867411-61867411 on Build GRCh38
rs780249098 Chr.12: 61867310-61867310 on Build GRCh38
rs34255534 Chr.12: 61867296-61867297 on Build GRCh38
rs922767184 Chr.12: 61867420-61867420 on Build GRCh38
rs779440808 Chr.12: 61867315-61867315 on Build GRCh38
rs373409433 Chr.12: 61867457-61867457 on Build GRCh38
rs746328289 Chr.12: 61867450-61867450 on Build GRCh38
rs755957485 Chr.12: 61867388-61867388 on Build GRCh38
rs772431122 Chr.12: 61867325-61867325 on Build GRCh38
rs775869315 Chr.12: 61867370-61867370 on Build GRCh38
rs377625494 Chr.12: 61867301-61867301 on Build GRCh38
rs201007901 Chr.12: 61867312-61867312 on Build GRCh38
rs959899719 Chr.12: 61867267-61867267 on Build GRCh38
rs1022649708 Chr.12: 61867473-61867473 on Build GRCh38
rs778033321 Chr.12: 61867308-61867308 on Build GRCh38
rs752002082 Chr.12: 61867372-61867374 on Build GRCh38
rs114325167 Chr.12: 61867463-61867463 on Build GRCh38
rs758978895 Chr.12: 61867451-61867451 on Build GRCh38
rs765416705 Chr.12: 61867297-61867297 on Build GRCh38
rs777947032 Chr.12: 61867393-61867393 on Build GRCh38
rs201610781 Chr.12: 61867365-61867365 on Build GRCh38
rs752860655 Chr.12: 61867297-61867297 on Build GRCh38
rs373370504 Chr.12: 61867329-61867329 on Build GRCh38
rs765838841 Chr.12: 61867347-61867347 on Build GRCh38
rs932789581 Chr.12: 61867430-61867430 on Build GRCh38
rs181558760 Chr.12: 61867337-61867337 on Build GRCh38
rs372001500 Chr.12: 61867474-61867474 on Build GRCh38
rs780991892 Chr.12: 61867283-61867283 on Build GRCh38
rs192011039 Chr.12: 61867453-61867453 on Build GRCh38
rs386376727 Chr.12: 61867300-61867301 on Build GRCh38
rs377373344 Chr.12: 61867346-61867346 on Build GRCh38
rs368021999 Chr.12: 61867306-61867307 on Build GRCh38
rs778879950 Chr.12: 61867417-61867417 on Build GRCh38
rs767236977 Chr.12: 61867372-61867372 on Build GRCh38
rs984499976 Chr.12: 61867330-61867330 on Build GRCh38
rs775069295 Chr.12: 61867271-61867271 on Build GRCh38
rs368142177 Chr.12: 61867469-61867469 on Build GRCh38

View Full Product Details