Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52292842-52293083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLYCTK
Entrez Gene ID
132158
Gene Name
glycerate kinase
Gene Aliases
HBEBP2, HBEBP4, HBeAgBP4A
UniGene
Hs.415312
Gene Chromosome Location
Chr.3: 52287558-52295256 on Build GRCh38
Transcripts
NM_001144951.1 NM_145262.3 NR_026699.1 NR_026700.1 NR_026701.1 NR_026702.1 XM_017005730.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6570106 c.1466C>G p.S489* Chr.3: 52293020-52293020 on Build GRCh38
1309324 c.1310G>T p.W437L Chr.3: 52292864-52292864 on Build GRCh38
6499687 c.1364A>T p.D455V Chr.3: 52292918-52292918 on Build GRCh38
5016008 c.1356_1357insG p.Q454fs*15 Chr.3: 52292910-52292911 on Build GRCh38
6671937 c.1345G>A p.G449S Chr.3: 52292899-52292899 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766538412 Chr.3: 52292924-52292924 on Build GRCh38
rs144669248 Chr.3: 52293055-52293055 on Build GRCh38
rs970569662 Chr.3: 52293020-52293020 on Build GRCh38
rs764539112 Chr.3: 52293025-52293025 on Build GRCh38
rs747079334 Chr.3: 52293010-52293010 on Build GRCh38
rs777622209 Chr.3: 52292948-52292949 on Build GRCh38
rs199610904 Chr.3: 52292869-52292869 on Build GRCh38
rs765754153 Chr.3: 52293018-52293018 on Build GRCh38
rs78388149 Chr.3: 52293022-52293022 on Build GRCh38
rs1039704518 Chr.3: 52292868-52292868 on Build GRCh38
rs769344021 Chr.3: 52292893-52292893 on Build GRCh38
rs766025190 Chr.3: 52293034-52293034 on Build GRCh38
rs749901380 Chr.3: 52293030-52293030 on Build GRCh38
rs960539661 Chr.3: 52292959-52292959 on Build GRCh38
rs771159734 Chr.3: 52293011-52293011 on Build GRCh38
rs371018021 Chr.3: 52292978-52292978 on Build GRCh38
rs751088590 Chr.3: 52292876-52292876 on Build GRCh38
rs147924619 Chr.3: 52292980-52292980 on Build GRCh38
rs780986541 Chr.3: 52292877-52292877 on Build GRCh38
rs778624214 Chr.3: 52293063-52293063 on Build GRCh38
rs146744702 Chr.3: 52292901-52292901 on Build GRCh38
rs145897189 Chr.3: 52292951-52292951 on Build GRCh38
rs142496523 Chr.3: 52292910-52292910 on Build GRCh38
rs749070406 Chr.3: 52292899-52292899 on Build GRCh38
rs773278880 Chr.3: 52292992-52292992 on Build GRCh38
rs1004368692 Chr.3: 52292953-52292953 on Build GRCh38
rs372098626 Chr.3: 52293040-52293040 on Build GRCh38
rs991718173 Chr.3: 52293013-52293013 on Build GRCh38
rs769748483 Chr.3: 52292889-52292890 on Build GRCh38
rs763485712 Chr.3: 52293023-52293023 on Build GRCh38
rs772013256 Chr.3: 52292987-52292987 on Build GRCh38
rs918720114 Chr.3: 52293054-52293054 on Build GRCh38
rs369555553 Chr.3: 52292988-52292988 on Build GRCh38
rs774322722 Chr.3: 52292907-52292907 on Build GRCh38
rs749904278 Chr.3: 52292962-52292962 on Build GRCh38
rs121909447 Chr.3: 52293002-52293002 on Build GRCh38
rs765804925 Chr.3: 52292945-52292945 on Build GRCh38
rs777002244 Chr.3: 52292941-52292941 on Build GRCh38
rs779582970 Chr.3: 52292872-52292872 on Build GRCh38
rs80017945 Chr.3: 52292968-52292968 on Build GRCh38
rs778822139 Chr.3: 52292984-52292984 on Build GRCh38
rs779829581 Chr.3: 52292898-52292898 on Build GRCh38
rs749088667 Chr.3: 52292979-52292979 on Build GRCh38
rs759555028 Chr.3: 52292883-52292883 on Build GRCh38
rs745590609 Chr.3: 52292882-52292882 on Build GRCh38
rs547441620 Chr.3: 52293027-52293027 on Build GRCh38
rs759970139 Chr.3: 52293016-52293016 on Build GRCh38
rs767047161 Chr.3: 52293014-52293014 on Build GRCh38
rs941592166 Chr.3: 52293037-52293037 on Build GRCh38
rs748144159 Chr.3: 52292986-52292986 on Build GRCh38
rs753322598 Chr.3: 52292949-52292949 on Build GRCh38
rs758823918 Chr.3: 52292867-52292867 on Build GRCh38
rs563928797 Chr.3: 52292943-52292943 on Build GRCh38
rs899837332 Chr.3: 52292881-52292881 on Build GRCh38
rs767077777 Chr.3: 52292952-52292952 on Build GRCh38
rs749082610 Chr.3: 52293027-52293029 on Build GRCh38
rs772211266 Chr.3: 52292908-52292908 on Build GRCh38
rs755753426 Chr.3: 52292970-52292970 on Build GRCh38
rs121909448 Chr.3: 52293032-52293032 on Build GRCh38
rs766827317 Chr.3: 52292866-52292866 on Build GRCh38
rs779596586 Chr.3: 52292977-52292977 on Build GRCh38

View Full Product Details