Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 4679711-4679984 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC37L1
Entrez Gene ID
55664
Gene Name
cell division cycle 37 like 1
Gene Aliases
CDC37B, HARC
UniGene
Hs.666439
Gene Chromosome Location
Chr.9: 4679553-4708399 on Build GRCh38
Transcripts
NM_017913.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3413677 c.120C>G p.G40G Chr.9: 4679887-4679887 on Build GRCh38
5429502 c.61G>A p.E21K Chr.9: 4679828-4679828 on Build GRCh38
4163931 c.71G>A p.S24N Chr.9: 4679838-4679838 on Build GRCh38
5426591 c.132+7A>G p.? Chr.9: 4679906-4679906 on Build GRCh38
6346080 c.18G>T p.P6P Chr.9: 4679785-4679785 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766764411 Chr.9: 4679782-4679782 on Build GRCh38
rs763145533 Chr.9: 4679911-4679911 on Build GRCh38
rs755145028 Chr.9: 4679786-4679786 on Build GRCh38
rs372857019 Chr.9: 4679742-4679742 on Build GRCh38
rs2295964 Chr.9: 4679906-4679906 on Build GRCh38
rs924091869 Chr.9: 4679869-4679869 on Build GRCh38
rs756399810 Chr.9: 4679775-4679775 on Build GRCh38
rs758829078 Chr.9: 4679847-4679847 on Build GRCh38
rs774003377 Chr.9: 4679830-4679830 on Build GRCh38
rs971278408 Chr.9: 4679765-4679765 on Build GRCh38
rs748887162 Chr.9: 4679875-4679875 on Build GRCh38
rs41298190 Chr.9: 4679733-4679733 on Build GRCh38
rs545423938 Chr.9: 4679947-4679947 on Build GRCh38
rs771941234 Chr.9: 4679903-4679903 on Build GRCh38
rs764477927 Chr.9: 4679913-4679913 on Build GRCh38
rs376094853 Chr.9: 4679858-4679858 on Build GRCh38
rs751371954 Chr.9: 4679774-4679774 on Build GRCh38
rs748716544 Chr.9: 4679949-4679949 on Build GRCh38
rs775331635 Chr.9: 4679753-4679753 on Build GRCh38
rs750851747 Chr.9: 4679936-4679936 on Build GRCh38
rs936895865 Chr.9: 4679965-4679965 on Build GRCh38
rs757309428 Chr.9: 4679810-4679810 on Build GRCh38
rs745352605 Chr.9: 4679807-4679807 on Build GRCh38
rs1039637869 Chr.9: 4679952-4679952 on Build GRCh38
rs375840544 Chr.9: 4679882-4679882 on Build GRCh38
rs763737113 Chr.9: 4679768-4679768 on Build GRCh38
rs779804868 Chr.9: 4679945-4679945 on Build GRCh38
rs774466876 Chr.9: 4679884-4679884 on Build GRCh38
rs778406400 Chr.9: 4679850-4679850 on Build GRCh38
rs745476037 Chr.9: 4679859-4679859 on Build GRCh38
rs371845373 Chr.9: 4679856-4679856 on Build GRCh38
rs560377407 Chr.9: 4679793-4679793 on Build GRCh38
rs769490808 Chr.9: 4679865-4679865 on Build GRCh38
rs998424425 Chr.9: 4679741-4679741 on Build GRCh38
rs776117650 Chr.9: 4679738-4679738 on Build GRCh38
rs769148024 Chr.9: 4679811-4679811 on Build GRCh38
rs765565737 Chr.9: 4679908-4679908 on Build GRCh38
rs762730773 Chr.9: 4679825-4679825 on Build GRCh38
rs770578513 Chr.9: 4679734-4679734 on Build GRCh38
rs145366983 Chr.9: 4679797-4679797 on Build GRCh38
rs200494761 Chr.9: 4679838-4679838 on Build GRCh38
rs11540755 Chr.9: 4679868-4679868 on Build GRCh38
rs1016622653 Chr.9: 4679787-4679787 on Build GRCh38
rs764156850 Chr.9: 4679827-4679827 on Build GRCh38
rs780273134 Chr.9: 4679742-4679742 on Build GRCh38
rs751602758 Chr.9: 4679764-4679764 on Build GRCh38
rs747444348 Chr.9: 4679802-4679802 on Build GRCh38
rs763206852 Chr.9: 4679880-4679880 on Build GRCh38
rs761748785 Chr.9: 4679834-4679834 on Build GRCh38
rs775089565 Chr.9: 4679814-4679814 on Build GRCh38
rs761869487 Chr.9: 4679893-4679893 on Build GRCh38
rs901089352 Chr.9: 4679962-4679962 on Build GRCh38
rs149264354 Chr.9: 4679739-4679739 on Build GRCh38
rs755706878 Chr.9: 4679942-4679942 on Build GRCh38
rs770561888 Chr.9: 4679804-4679804 on Build GRCh38
rs142699976 Chr.9: 4679871-4679871 on Build GRCh38
rs765137666 Chr.9: 4679748-4679748 on Build GRCh38
rs191901333 Chr.9: 4679919-4679919 on Build GRCh38
rs759612765 Chr.9: 4679907-4679907 on Build GRCh38
rs765175145 Chr.9: 4679845-4679845 on Build GRCh38
rs146915614 Chr.9: 4679836-4679836 on Build GRCh38
rs757784566 Chr.9: 4679923-4679923 on Build GRCh38
rs753942730 Chr.9: 4679785-4679785 on Build GRCh38
rs781479991 Chr.9: 4679924-4679924 on Build GRCh38
rs138614276 Chr.9: 4679846-4679846 on Build GRCh38
rs758959917 Chr.9: 4679796-4679796 on Build GRCh38
rs374431975 Chr.9: 4679732-4679732 on Build GRCh38

View Full Product Details