Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 61490347-61490608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH20
Entrez Gene ID
28316
Gene Name
cadherin 20
Gene Aliases
CDH7L3, Cdh7
UniGene
Hs.671510
Gene Chromosome Location
Chr.18: 61333582-61555773 on Build GRCh38
Transcripts
NM_031891.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5554813 c.50G>A p.G17D Chr.18: 61490603-61490603 on Build GRCh38
5803298 c.12T>A p.S4S Chr.18: 61490565-61490565 on Build GRCh38
3526865 c.36C>T p.N12N Chr.18: 61490589-61490589 on Build GRCh38
3526864 c.6G>A p.W2* Chr.18: 61490559-61490559 on Build GRCh38
4072827 c.17G>T p.R6I Chr.18: 61490570-61490570 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765637114 Chr.18: 61490453-61490453 on Build GRCh38
rs986823227 Chr.18: 61490517-61490517 on Build GRCh38
rs1030508027 Chr.18: 61490461-61490461 on Build GRCh38
rs564783201 Chr.18: 61490477-61490478 on Build GRCh38
rs910070273 Chr.18: 61490439-61490439 on Build GRCh38
rs147796041 Chr.18: 61490589-61490589 on Build GRCh38
rs369649809 Chr.18: 61490434-61490434 on Build GRCh38
rs371708959 Chr.18: 61490576-61490576 on Build GRCh38
rs918893880 Chr.18: 61490578-61490578 on Build GRCh38
rs956791973 Chr.18: 61490447-61490447 on Build GRCh38
rs1039963539 Chr.18: 61490490-61490490 on Build GRCh38
rs680530 Chr.18: 61490531-61490531 on Build GRCh38
rs900088458 Chr.18: 61490498-61490498 on Build GRCh38
rs564829777 Chr.18: 61490471-61490471 on Build GRCh38
rs1033484718 Chr.18: 61490545-61490545 on Build GRCh38
rs910792052 Chr.18: 61490519-61490519 on Build GRCh38
rs201360046 Chr.18: 61490503-61490503 on Build GRCh38
rs186858310 Chr.18: 61490555-61490555 on Build GRCh38
rs776056053 Chr.18: 61490504-61490504 on Build GRCh38
rs755600247 Chr.18: 61490573-61490573 on Build GRCh38
rs553884016 Chr.18: 61490414-61490414 on Build GRCh38
rs113254321 Chr.18: 61490492-61490492 on Build GRCh38
rs753962275 Chr.18: 61490571-61490571 on Build GRCh38
rs551265370 Chr.18: 61490472-61490473 on Build GRCh38
rs752583729 Chr.18: 61490514-61490514 on Build GRCh38
rs374394371 Chr.18: 61490511-61490511 on Build GRCh38
rs181103378 Chr.18: 61490456-61490456 on Build GRCh38
rs780283426 Chr.18: 61490567-61490567 on Build GRCh38
rs763561765 Chr.18: 61490547-61490548 on Build GRCh38
rs929079479 Chr.18: 61490433-61490433 on Build GRCh38
rs575454805 Chr.18: 61490485-61490485 on Build GRCh38
rs759541624 Chr.18: 61490507-61490507 on Build GRCh38
rs750630687 Chr.18: 61490550-61490550 on Build GRCh38
rs903146581 Chr.18: 61490526-61490526 on Build GRCh38

View Full Product Details