Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 38778914-38779187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GEMIN6
Entrez Gene ID
79833
Gene Name
gem nuclear organelle associated protein 6
Gene Aliases
-
UniGene
Hs.143818
Gene Chromosome Location
Chr.2: 38778185-38781965 on Build GRCh38
Transcripts
NM_024775.9

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5896607 c.115C>T p.P39S Chr.2: 38779105-38779105 on Build GRCh38
4686776 c.128+1G>A p.? Chr.2: 38779119-38779119 on Build GRCh38
1241206 c.110C>G p.T37R Chr.2: 38779100-38779100 on Build GRCh38
5422968 c.51C>A p.Y17* Chr.2: 38779041-38779041 on Build GRCh38
4583493 c.87G>A p.E29E Chr.2: 38779077-38779077 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771676634 Chr.2: 38779113-38779113 on Build GRCh38
rs767671567 Chr.2: 38779156-38779156 on Build GRCh38
rs763381132 Chr.2: 38779013-38779013 on Build GRCh38
rs370876424 Chr.2: 38779026-38779026 on Build GRCh38
rs745516216 Chr.2: 38779044-38779044 on Build GRCh38
rs142049461 Chr.2: 38779064-38779064 on Build GRCh38
rs775003461 Chr.2: 38779004-38779004 on Build GRCh38
rs774926487 Chr.2: 38779047-38779047 on Build GRCh38
rs778573363 Chr.2: 38779106-38779106 on Build GRCh38
rs747156484 Chr.2: 38778954-38778954 on Build GRCh38
rs755048311 Chr.2: 38778956-38778956 on Build GRCh38
rs1039599703 Chr.2: 38779037-38779037 on Build GRCh38
rs201717504 Chr.2: 38779107-38779107 on Build GRCh38
rs770742111 Chr.2: 38779030-38779030 on Build GRCh38
rs201922909 Chr.2: 38778975-38778975 on Build GRCh38
rs199945237 Chr.2: 38779140-38779140 on Build GRCh38
rs749071755 Chr.2: 38778978-38778978 on Build GRCh38
rs759744985 Chr.2: 38779077-38779077 on Build GRCh38
rs771466232 Chr.2: 38779134-38779134 on Build GRCh38
rs778720924 Chr.2: 38779035-38779035 on Build GRCh38
rs748048568 Chr.2: 38778966-38778966 on Build GRCh38
rs777054481 Chr.2: 38779066-38779068 on Build GRCh38
rs1035401944 Chr.2: 38779154-38779154 on Build GRCh38
rs769037175 Chr.2: 38779037-38779037 on Build GRCh38
rs762410973 Chr.2: 38779007-38779007 on Build GRCh38
rs756153830 Chr.2: 38779085-38779085 on Build GRCh38
rs201171759 Chr.2: 38779131-38779131 on Build GRCh38
rs368627031 Chr.2: 38779012-38779012 on Build GRCh38
rs202030757 Chr.2: 38779161-38779161 on Build GRCh38
rs548768553 Chr.2: 38779082-38779082 on Build GRCh38
rs781137959 Chr.2: 38779023-38779023 on Build GRCh38
rs769653000 Chr.2: 38778967-38778967 on Build GRCh38
rs746622382 Chr.2: 38779117-38779117 on Build GRCh38
rs753794665 Chr.2: 38779089-38779089 on Build GRCh38
rs757128366 Chr.2: 38779105-38779105 on Build GRCh38
rs777789159 Chr.2: 38779086-38779086 on Build GRCh38
rs546561131 Chr.2: 38779157-38779157 on Build GRCh38
rs951017586 Chr.2: 38779150-38779150 on Build GRCh38
rs762188224 Chr.2: 38779074-38779074 on Build GRCh38
rs751756949 Chr.2: 38779016-38779016 on Build GRCh38
rs186951733 Chr.2: 38779008-38779008 on Build GRCh38
rs530479807 Chr.2: 38779052-38779052 on Build GRCh38
rs760833787 Chr.2: 38779158-38779158 on Build GRCh38
rs758757405 Chr.2: 38778951-38778951 on Build GRCh38
rs375382710 Chr.2: 38779153-38779153 on Build GRCh38
rs773842266 Chr.2: 38778988-38778988 on Build GRCh38
rs779727449 Chr.2: 38779116-38779116 on Build GRCh38
rs866634574 Chr.2: 38778946-38778946 on Build GRCh38
rs759186850 Chr.2: 38778997-38778997 on Build GRCh38
rs771504738 Chr.2: 38779000-38779000 on Build GRCh38
rs370812620 Chr.2: 38778950-38778950 on Build GRCh38
rs375864812 Chr.2: 38778949-38778949 on Build GRCh38
rs200189789 Chr.2: 38779147-38779147 on Build GRCh38
rs755170271 Chr.2: 38779021-38779021 on Build GRCh38
rs771685081 Chr.2: 38779046-38779046 on Build GRCh38
rs752819069 Chr.2: 38779024-38779024 on Build GRCh38
rs375747029 Chr.2: 38779060-38779060 on Build GRCh38
rs749119944 Chr.2: 38779027-38779027 on Build GRCh38
rs745668104 Chr.2: 38779109-38779109 on Build GRCh38
rs563056973 Chr.2: 38779015-38779015 on Build GRCh38
rs182583282 Chr.2: 38778979-38778979 on Build GRCh38
rs768307045 Chr.2: 38779125-38779125 on Build GRCh38
rs770467240 Chr.2: 38779051-38779051 on Build GRCh38
rs780403773 Chr.2: 38778953-38778953 on Build GRCh38
rs921598759 Chr.2: 38778971-38778971 on Build GRCh38
rs372135552 Chr.2: 38779133-38779133 on Build GRCh38
rs934654472 Chr.2: 38778999-38778999 on Build GRCh38
rs376044156 Chr.2: 38779002-38779002 on Build GRCh38
rs373228867 Chr.2: 38778947-38778947 on Build GRCh38
rs201740651 Chr.2: 38778964-38778964 on Build GRCh38

View Full Product Details