Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 4784136-4784407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP5Z1
Entrez Gene ID
9907
Gene Name
adaptor related protein complex 5 zeta 1 subunit
Gene Aliases
KIAA0415, SPG48, zeta
UniGene
Hs.558440
Gene Chromosome Location
Chr.7: 4775617-4794395 on Build GRCh38
Transcripts
NM_014855.2 XM_017012864.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1090243 c.2850C>T p.N950N Chr.7: 4784298-4784298 on Build GRCh38
4505359 c.2823C>T p.H941H Chr.7: 4784271-4784271 on Build GRCh38
1211909 c.2762C>T p.P921L Chr.7: 4784210-4784210 on Build GRCh38
4640803 c.2851G>A p.V951M Chr.7: 4784299-4784299 on Build GRCh38
1451085 c.2854C>T p.Q952* Chr.7: 4784302-4784302 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747323854 Chr.7: 4784257-4784257 on Build GRCh38
rs759449685 Chr.7: 4784378-4784378 on Build GRCh38
rs750707781 Chr.7: 4784346-4784346 on Build GRCh38
rs771896080 Chr.7: 4784325-4784325 on Build GRCh38
rs201862383 Chr.7: 4784252-4784252 on Build GRCh38
rs760548155 Chr.7: 4784331-4784331 on Build GRCh38
rs568379343 Chr.7: 4784367-4784367 on Build GRCh38
rs766379431 Chr.7: 4784286-4784286 on Build GRCh38
rs774068163 Chr.7: 4784321-4784321 on Build GRCh38
rs763884027 Chr.7: 4784231-4784231 on Build GRCh38
rs749342646 Chr.7: 4784198-4784198 on Build GRCh38
rs781388706 Chr.7: 4784306-4784306 on Build GRCh38
rs888122759 Chr.7: 4784228-4784228 on Build GRCh38
rs763968248 Chr.7: 4784181-4784181 on Build GRCh38
rs757440325 Chr.7: 4784343-4784343 on Build GRCh38
rs770383096 Chr.7: 4784268-4784268 on Build GRCh38
rs370728528 Chr.7: 4784283-4784283 on Build GRCh38
rs535817519 Chr.7: 4784162-4784162 on Build GRCh38
rs774498322 Chr.7: 4784328-4784328 on Build GRCh38
rs770763321 Chr.7: 4784375-4784375 on Build GRCh38
rs572533659 Chr.7: 4784180-4784180 on Build GRCh38
rs761542147 Chr.7: 4784324-4784324 on Build GRCh38
rs576929346 Chr.7: 4784221-4784221 on Build GRCh38
rs773542837 Chr.7: 4784209-4784209 on Build GRCh38
rs748752062 Chr.7: 4784316-4784316 on Build GRCh38
rs747413625 Chr.7: 4784211-4784211 on Build GRCh38
rs377532483 Chr.7: 4784349-4784349 on Build GRCh38
rs1025960741 Chr.7: 4784171-4784172 on Build GRCh38
rs780361298 Chr.7: 4784351-4784351 on Build GRCh38
rs758494090 Chr.7: 4784299-4784299 on Build GRCh38
rs988434253 Chr.7: 4784287-4784287 on Build GRCh38
rs372585325 Chr.7: 4784238-4784238 on Build GRCh38
rs780637816 Chr.7: 4784314-4784314 on Build GRCh38
rs772940437 Chr.7: 4784365-4784365 on Build GRCh38
rs752299234 Chr.7: 4784166-4784166 on Build GRCh38
rs564871742 Chr.7: 4784190-4784190 on Build GRCh38
rs886062351 Chr.7: 4784305-4784305 on Build GRCh38
rs771385792 Chr.7: 4784212-4784212 on Build GRCh38
rs768288736 Chr.7: 4784320-4784320 on Build GRCh38
rs758587330 Chr.7: 4784255-4784255 on Build GRCh38
rs757143181 Chr.7: 4784233-4784233 on Build GRCh38
rs770164019 Chr.7: 4784307-4784307 on Build GRCh38
rs758137709 Chr.7: 4784175-4784175 on Build GRCh38
rs1011767351 Chr.7: 4784247-4784247 on Build GRCh38
rs759021146 Chr.7: 4784218-4784218 on Build GRCh38
rs774163957 Chr.7: 4784278-4784278 on Build GRCh38
rs554159417 Chr.7: 4784168-4784168 on Build GRCh38
rs942559540 Chr.7: 4784176-4784176 on Build GRCh38
rs775124242 Chr.7: 4784226-4784226 on Build GRCh38
rs765101143 Chr.7: 4784208-4784208 on Build GRCh38
rs373689894 Chr.7: 4784355-4784355 on Build GRCh38
rs760503818 Chr.7: 4784284-4784284 on Build GRCh38
rs763554237 Chr.7: 4784384-4784401 on Build GRCh38
rs767507147 Chr.7: 4784237-4784237 on Build GRCh38
rs762490448 Chr.7: 4784227-4784227 on Build GRCh38
rs761636459 Chr.7: 4784280-4784280 on Build GRCh38
rs777143531 Chr.7: 4784217-4784217 on Build GRCh38
rs967221913 Chr.7: 4784167-4784175 on Build GRCh38
rs752679480 Chr.7: 4784298-4784298 on Build GRCh38
rs986640228 Chr.7: 4784270-4784270 on Build GRCh38
rs767413099 Chr.7: 4784282-4784282 on Build GRCh38
rs778539799 Chr.7: 4784359-4784359 on Build GRCh38
rs767812395 Chr.7: 4784345-4784345 on Build GRCh38
rs954372252 Chr.7: 4784188-4784212 on Build GRCh38
rs755024012 Chr.7: 4784200-4784200 on Build GRCh38
rs751448771 Chr.7: 4784186-4784186 on Build GRCh38
rs771584271 Chr.7: 4784363-4784363 on Build GRCh38
rs907448674 Chr.7: 4784222-4784222 on Build GRCh38
rs544081556 Chr.7: 4784244-4784244 on Build GRCh38
rs894028182 Chr.7: 4784360-4784360 on Build GRCh38
rs983896546 Chr.7: 4784337-4784337 on Build GRCh38
rs776469236 Chr.7: 4784377-4784377 on Build GRCh38
rs776086980 Chr.7: 4784160-4784160 on Build GRCh38
rs774832344 Chr.7: 4784272-4784272 on Build GRCh38
rs775348165 Chr.7: 4784383-4784383 on Build GRCh38
rs908638350 Chr.7: 4784334-4784334 on Build GRCh38
rs768378443 Chr.7: 4784275-4784275 on Build GRCh38
rs889288295 Chr.7: 4784357-4784357 on Build GRCh38
rs886062352 Chr.7: 4784386-4784386 on Build GRCh38
rs756140557 Chr.7: 4784241-4784241 on Build GRCh38
rs751362842 Chr.7: 4784232-4784232 on Build GRCh38
rs377620768 Chr.7: 4784266-4784266 on Build GRCh38
rs774375966 Chr.7: 4784387-4784387 on Build GRCh38
rs765199218 Chr.7: 4784380-4784380 on Build GRCh38
rs751778396 Chr.7: 4784302-4784302 on Build GRCh38
rs762521850 Chr.7: 4784382-4784383 on Build GRCh38
rs771296028 Chr.7: 4784259-4784259 on Build GRCh38
rs777854428 Chr.7: 4784159-4784159 on Build GRCh38
rs377507553 Chr.7: 4784361-4784361 on Build GRCh38
rs766178111 Chr.7: 4784332-4784332 on Build GRCh38
rs781670584 Chr.7: 4784264-4784264 on Build GRCh38
rs764892996 Chr.7: 4784165-4784165 on Build GRCh38
rs772392202 Chr.7: 4784205-4784205 on Build GRCh38
rs369755844 Chr.7: 4784253-4784253 on Build GRCh38
rs757535159 Chr.7: 4784303-4784303 on Build GRCh38
rs77790584 Chr.7: 4784383-4784383 on Build GRCh38
rs374167435 Chr.7: 4784256-4784256 on Build GRCh38
rs17135121 Chr.7: 4784340-4784340 on Build GRCh38
rs746349613 Chr.7: 4784265-4784265 on Build GRCh38
rs772318205 Chr.7: 4784341-4784341 on Build GRCh38
rs779121737 Chr.7: 4784204-4784204 on Build GRCh38
rs749326601 Chr.7: 4784273-4784273 on Build GRCh38
rs540320285 Chr.7: 4784189-4784189 on Build GRCh38
rs560484295 Chr.7: 4784301-4784301 on Build GRCh38
rs370552243 Chr.7: 4784354-4784354 on Build GRCh38
rs987017556 Chr.7: 4784169-4784169 on Build GRCh38

View Full Product Details