Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 37439093-37439348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NWD2
Entrez Gene ID
57495
Gene Name
NACHT and WD repeat domain containing 2
Gene Aliases
KIAA1239
UniGene
Hs.4280
Gene Chromosome Location
Chr.4: 37245068-37449465 on Build GRCh38
Transcripts
NM_001144990.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7211302 c.1022G>A p.G341E Chr.4: 37439116-37439116 on Build GRCh38
4609064 c.1009C>A p.H337N Chr.4: 37439103-37439103 on Build GRCh38
3825783 c.1017C>A p.I339I Chr.4: 37439111-37439111 on Build GRCh38
4797319 c.1197C>T p.N399N Chr.4: 37439291-37439291 on Build GRCh38
4135754 c.1027G>A p.G343S Chr.4: 37439121-37439121 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs963824902 Chr.4: 37439193-37439193 on Build GRCh38
rs1018076447 Chr.4: 37439181-37439181 on Build GRCh38
rs182008355 Chr.4: 37439244-37439244 on Build GRCh38
rs200042443 Chr.4: 37439274-37439274 on Build GRCh38
rs766789155 Chr.4: 37439165-37439165 on Build GRCh38
rs1022418879 Chr.4: 37439144-37439144 on Build GRCh38
rs570036994 Chr.4: 37439239-37439239 on Build GRCh38
rs950267368 Chr.4: 37439210-37439210 on Build GRCh38
rs922499612 Chr.4: 37439243-37439243 on Build GRCh38
rs556772563 Chr.4: 37439202-37439202 on Build GRCh38
rs1055967603 Chr.4: 37439151-37439151 on Build GRCh38
rs767912696 Chr.4: 37439201-37439201 on Build GRCh38
rs868075044 Chr.4: 37439164-37439164 on Build GRCh38
rs79390097 Chr.4: 37439191-37439191 on Build GRCh38
rs142483670 Chr.4: 37439150-37439150 on Build GRCh38
rs567644799 Chr.4: 37439148-37439148 on Build GRCh38

View Full Product Details