Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 58146331-58146584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM110B
Entrez Gene ID
90362
Gene Name
family with sequence similarity 110 member B
Gene Aliases
C8orf72
UniGene
Hs.154652
Gene Chromosome Location
Chr.8: 57994444-58149718 on Build GRCh38
Transcripts
NM_147189.2 XM_005251324.2 XM_005251325.3 XM_005251326.2 XM_017013948.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1457613 c.184G>A p.V62I Chr.8: 58146414-58146414 on Build GRCh38
5452174 c.225G>A p.V75V Chr.8: 58146455-58146455 on Build GRCh38
1457615 c.274G>A p.A92T Chr.8: 58146504-58146504 on Build GRCh38
5468116 c.131A>G p.N44S Chr.8: 58146361-58146361 on Build GRCh38
4406074 c.196G>A p.E66K Chr.8: 58146426-58146426 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879112430 Chr.8: 58146464-58146464 on Build GRCh38
rs768685073 Chr.8: 58146438-58146438 on Build GRCh38
rs752411132 Chr.8: 58146504-58146504 on Build GRCh38
rs868676674 Chr.8: 58146554-58146554 on Build GRCh38
rs938698212 Chr.8: 58146426-58146426 on Build GRCh38
rs766614164 Chr.8: 58146561-58146561 on Build GRCh38
rs368594517 Chr.8: 58146533-58146533 on Build GRCh38
rs760719623 Chr.8: 58146428-58146428 on Build GRCh38
rs747760336 Chr.8: 58146367-58146367 on Build GRCh38
rs763429337 Chr.8: 58146490-58146490 on Build GRCh38
rs751067454 Chr.8: 58146353-58146353 on Build GRCh38
rs750795228 Chr.8: 58146483-58146483 on Build GRCh38
rs761969859 Chr.8: 58146462-58146462 on Build GRCh38
rs375902544 Chr.8: 58146503-58146503 on Build GRCh38
rs556320056 Chr.8: 58146520-58146520 on Build GRCh38
rs754729106 Chr.8: 58146361-58146361 on Build GRCh38
rs778876440 Chr.8: 58146521-58146521 on Build GRCh38
rs1056236127 Chr.8: 58146478-58146478 on Build GRCh38
rs779498329 Chr.8: 58146349-58146349 on Build GRCh38
rs749092772 Chr.8: 58146511-58146511 on Build GRCh38
rs770894056 Chr.8: 58146394-58146394 on Build GRCh38
rs539671135 Chr.8: 58146479-58146479 on Build GRCh38
rs776628310 Chr.8: 58146535-58146535 on Build GRCh38
rs865938034 Chr.8: 58146471-58146471 on Build GRCh38
rs755760538 Chr.8: 58146507-58146507 on Build GRCh38
rs369974822 Chr.8: 58146510-58146510 on Build GRCh38
rs776863778 Chr.8: 58146455-58146455 on Build GRCh38
rs745759037 Chr.8: 58146522-58146522 on Build GRCh38
rs894466338 Chr.8: 58146488-58146488 on Build GRCh38
rs768740067 Chr.8: 58146525-58146525 on Build GRCh38
rs769732551 Chr.8: 58146372-58146372 on Build GRCh38
rs770044019 Chr.8: 58146557-58146557 on Build GRCh38
rs751156478 Chr.8: 58146502-58146502 on Build GRCh38
rs1047103 Chr.8: 58146407-58146407 on Build GRCh38
rs777565277 Chr.8: 58146380-58146380 on Build GRCh38
rs748440229 Chr.8: 58146536-58146536 on Build GRCh38
rs773498874 Chr.8: 58146558-58146558 on Build GRCh38
rs763347134 Chr.8: 58146559-58146559 on Build GRCh38
rs780962054 Chr.8: 58146362-58146362 on Build GRCh38
rs775487158 Chr.8: 58146398-58146398 on Build GRCh38
rs866732145 Chr.8: 58146392-58146392 on Build GRCh38

View Full Product Details