Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3655321-3655578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNASE1
Entrez Gene ID
1773
Gene Name
deoxyribonuclease I
Gene Aliases
DNL1, DRNI
UniGene
Hs.629638
Gene Chromosome Location
Chr.16: 3611705-3665472 on Build GRCh38
Transcripts
NM_005223.3 XM_005255148.3 XM_006720854.3 XM_011522393.2 XM_017022992.1 XM_017022993.1 XM_017022994.1 XM_017022995.1 XM_017022996.1 XM_017022997.1 XM_017022998.1 XM_017022999.1 XM_017023000.1 XM_017023001.1 XM_017023002.1 XM_017023003.1 XM_017023004.1 XM_017023005.1 XM_017023007.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7365126 c.105G>C p.E35D Chr.16: 3655478-3655478 on Build GRCh38
4590090 c.119A>G p.N40S Chr.16: 3655492-3655492 on Build GRCh38
4985752 c.6G>T p.R2S Chr.16: 3655379-3655379 on Build GRCh38
6647442 c.59C>G p.A20G Chr.16: 3655432-3655432 on Build GRCh38
6647443 c.54delG p.A20fs*4 Chr.16: 3655427-3655427 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1020893783 Chr.16: 3655360-3655360 on Build GRCh38
rs914092304 Chr.16: 3655396-3655396 on Build GRCh38
rs961223901 Chr.16: 3655347-3655347 on Build GRCh38
rs769521038 Chr.16: 3655513-3655513 on Build GRCh38
rs780596998 Chr.16: 3655377-3655377 on Build GRCh38
rs759107799 Chr.16: 3655392-3655392 on Build GRCh38
rs747491349 Chr.16: 3655429-3655429 on Build GRCh38
rs367607486 Chr.16: 3655391-3655391 on Build GRCh38
rs199822100 Chr.16: 3655543-3655543 on Build GRCh38
rs1029409874 Chr.16: 3655355-3655355 on Build GRCh38
rs760546693 Chr.16: 3655410-3655410 on Build GRCh38
rs371362495 Chr.16: 3655433-3655433 on Build GRCh38
rs535831887 Chr.16: 3655537-3655537 on Build GRCh38
rs941339218 Chr.16: 3655554-3655554 on Build GRCh38
rs979279136 Chr.16: 3655375-3655375 on Build GRCh38
rs8176927 Chr.16: 3655379-3655379 on Build GRCh38
rs757085573 Chr.16: 3655417-3655417 on Build GRCh38
rs140187838 Chr.16: 3655510-3655510 on Build GRCh38
rs778247219 Chr.16: 3655550-3655550 on Build GRCh38
rs1049068889 Chr.16: 3655435-3655435 on Build GRCh38
rs201161491 Chr.16: 3655448-3655448 on Build GRCh38
rs554921505 Chr.16: 3655349-3655350 on Build GRCh38
rs938098121 Chr.16: 3655529-3655529 on Build GRCh38
rs141673463 Chr.16: 3655449-3655449 on Build GRCh38
rs370054136 Chr.16: 3655503-3655503 on Build GRCh38
rs578133143 Chr.16: 3655378-3655378 on Build GRCh38
rs775433436 Chr.16: 3655403-3655403 on Build GRCh38
rs750574476 Chr.16: 3655461-3655461 on Build GRCh38
rs34907394 Chr.16: 3655478-3655478 on Build GRCh38
rs758497994 Chr.16: 3655426-3655426 on Build GRCh38
rs771655787 Chr.16: 3655492-3655492 on Build GRCh38
rs376685545 Chr.16: 3655366-3655366 on Build GRCh38
rs768214589 Chr.16: 3655502-3655502 on Build GRCh38
rs766430443 Chr.16: 3655466-3655466 on Build GRCh38
rs756483303 Chr.16: 3655489-3655489 on Build GRCh38
rs778926507 Chr.16: 3655363-3655363 on Build GRCh38
rs774887851 Chr.16: 3655442-3655442 on Build GRCh38
rs953839621 Chr.16: 3655485-3655485 on Build GRCh38
rs773012748 Chr.16: 3655517-3655517 on Build GRCh38
rs143327739 Chr.16: 3655481-3655481 on Build GRCh38
rs121912990 Chr.16: 3655386-3655386 on Build GRCh38
rs61741279 Chr.16: 3655381-3655381 on Build GRCh38
rs368497656 Chr.16: 3655350-3655350 on Build GRCh38
rs147546841 Chr.16: 3655509-3655509 on Build GRCh38
rs201945500 Chr.16: 3655558-3655558 on Build GRCh38
rs770328454 Chr.16: 3655432-3655432 on Build GRCh38
rs776458509 Chr.16: 3655462-3655462 on Build GRCh38
rs8176930 Chr.16: 3655469-3655469 on Build GRCh38
rs925599221 Chr.16: 3655370-3655370 on Build GRCh38
rs982314101 Chr.16: 3655526-3655526 on Build GRCh38
rs779796843 Chr.16: 3655493-3655493 on Build GRCh38
rs945564930 Chr.16: 3655399-3655399 on Build GRCh38
rs770733416 Chr.16: 3655387-3655387 on Build GRCh38
rs369619441 Chr.16: 3655486-3655486 on Build GRCh38
rs199538085 Chr.16: 3655415-3655415 on Build GRCh38
rs754004304 Chr.16: 3655349-3655349 on Build GRCh38
rs77254040 Chr.16: 3655464-3655464 on Build GRCh38
rs767007240 Chr.16: 3655394-3655394 on Build GRCh38
rs8176922 Chr.16: 3655358-3655358 on Build GRCh38
rs746491054 Chr.16: 3655394-3655394 on Build GRCh38
rs8176921 Chr.16: 3655352-3655353 on Build GRCh38
rs755459254 Chr.16: 3655431-3655431 on Build GRCh38
rs931600934 Chr.16: 3655548-3655548 on Build GRCh38
rs780001880 Chr.16: 3655428-3655428 on Build GRCh38
rs759518571 Chr.16: 3655544-3655544 on Build GRCh38
rs755123150 Chr.16: 3655471-3655471 on Build GRCh38
rs779747135 Chr.16: 3655453-3655453 on Build GRCh38
rs374609613 Chr.16: 3655373-3655373 on Build GRCh38
rs551458320 Chr.16: 3655477-3655477 on Build GRCh38
rs750700513 Chr.16: 3655424-3655424 on Build GRCh38
rs565570420 Chr.16: 3655441-3655442 on Build GRCh38
rs144058317 Chr.16: 3655499-3655499 on Build GRCh38
rs745479565 Chr.16: 3655491-3655491 on Build GRCh38
rs373820749 Chr.16: 3655439-3655439 on Build GRCh38
rs763190517 Chr.16: 3655523-3655523 on Build GRCh38
rs148015097 Chr.16: 3655414-3655414 on Build GRCh38
rs761583800 Chr.16: 3655454-3655454 on Build GRCh38
rs758458285 Chr.16: 3655376-3655376 on Build GRCh38
rs200484716 Chr.16: 3655380-3655380 on Build GRCh38
rs745523317 Chr.16: 3655434-3655434 on Build GRCh38
rs200404670 Chr.16: 3655490-3655490 on Build GRCh38
rs768282770 Chr.16: 3655401-3655409 on Build GRCh38
rs758128465 Chr.16: 3655345-3655346 on Build GRCh38
rs562520867 Chr.16: 3655455-3655455 on Build GRCh38
rs370732305 Chr.16: 3655549-3655549 on Build GRCh38
rs928096402 Chr.16: 3655528-3655528 on Build GRCh38

View Full Product Details