Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 236468062-236468325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDARADD
Entrez Gene ID
128178
Gene Name
EDAR associated death domain
Gene Aliases
ECTD11A, ECTD11B, ED3, EDA3
UniGene
Hs.352224
Gene Chromosome Location
Chr.1: 236394380-236484708 on Build GRCh38
Transcripts
NM_080738.3 NM_145861.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6651549 c.226G>C p.E76Q Chr.1: 236468237-236468237 on Build GRCh38
5845932 c.233C>T p.P78L Chr.1: 236468274-236468274 on Build GRCh38
6062127 c.246C>T p.S82S Chr.1: 236468257-236468257 on Build GRCh38
6651550 c.196G>C p.E66Q Chr.1: 236468237-236468237 on Build GRCh38
6062128 c.216C>T p.S72S Chr.1: 236468257-236468257 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376772514 Chr.1: 236468265-236468265 on Build GRCh38
rs747953067 Chr.1: 236468224-236468224 on Build GRCh38
rs567161985 Chr.1: 236468301-236468301 on Build GRCh38
rs1044285599 Chr.1: 236468231-236468231 on Build GRCh38
rs968813348 Chr.1: 236468144-236468144 on Build GRCh38
rs895281473 Chr.1: 236468134-236468134 on Build GRCh38
rs540478996 Chr.1: 236468142-236468142 on Build GRCh38
rs763211638 Chr.1: 236468253-236468253 on Build GRCh38
rs35734689 Chr.1: 236468100-236468101 on Build GRCh38
rs768595907 Chr.1: 236468212-236468212 on Build GRCh38
rs897579878 Chr.1: 236468296-236468296 on Build GRCh38
rs1055330533 Chr.1: 236468260-236468260 on Build GRCh38
rs965116592 Chr.1: 236468185-236468185 on Build GRCh38
rs766731640 Chr.1: 236468257-236468257 on Build GRCh38
rs1011195768 Chr.1: 236468138-236468138 on Build GRCh38
rs778788277 Chr.1: 236468217-236468217 on Build GRCh38
rs372150890 Chr.1: 236468199-236468199 on Build GRCh38
rs138967527 Chr.1: 236468269-236468269 on Build GRCh38
rs886046185 Chr.1: 236468216-236468216 on Build GRCh38
rs758046261 Chr.1: 236468188-236468188 on Build GRCh38
rs1031723003 Chr.1: 236468172-236468174 on Build GRCh38
rs998516716 Chr.1: 236468157-236468157 on Build GRCh38
rs370659008 Chr.1: 236468169-236468169 on Build GRCh38
rs1003601106 Chr.1: 236468094-236468094 on Build GRCh38
rs904060537 Chr.1: 236468244-236468244 on Build GRCh38
rs773065240 Chr.1: 236468242-236468242 on Build GRCh38
rs768049797 Chr.1: 236468279-236468279 on Build GRCh38
rs201611000 Chr.1: 236468299-236468299 on Build GRCh38
rs150720000 Chr.1: 236468193-236468193 on Build GRCh38
rs1033866648 Chr.1: 236468118-236468118 on Build GRCh38
rs367622669 Chr.1: 236468285-236468285 on Build GRCh38
rs776471811 Chr.1: 236468223-236468223 on Build GRCh38
rs4565703 Chr.1: 236468291-236468291 on Build GRCh38
rs878931623 Chr.1: 236468303-236468303 on Build GRCh38
rs769534560 Chr.1: 236468227-236468227 on Build GRCh38
rs1045342641 Chr.1: 236468284-236468284 on Build GRCh38

View Full Product Details