Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 142878945-142879200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP11B1
Entrez Gene ID
1584
Gene Name
cytochrome P450 family 11 subfamily B member 1
Gene Aliases
CPN1, CYP11B, FHI, P450C11
UniGene
Hs.184927
Gene Chromosome Location
Chr.8: 142872354-142879839 on Build GRCh38
Transcripts
NM_000497.3 NM_001026213.1 XM_011516874.2 XM_011516875.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3924769 c.367C>T p.R123C Chr.8: 142879060-142879060 on Build GRCh38
5781600 c.364C>A p.H122N Chr.8: 142879063-142879063 on Build GRCh38
3645963 c.244G>A p.D82N Chr.8: 142879183-142879183 on Build GRCh38
3645961 c.345C>T p.P115P Chr.8: 142879082-142879082 on Build GRCh38
3645962 c.344C>T p.P115L Chr.8: 142879083-142879083 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756289827 Chr.8: 142879148-142879148 on Build GRCh38
rs764087240 Chr.8: 142879051-142879051 on Build GRCh38
rs61751140 Chr.8: 142879023-142879023 on Build GRCh38
rs774981688 Chr.8: 142879169-142879169 on Build GRCh38
rs961164148 Chr.8: 142879032-142879032 on Build GRCh38
rs200867786 Chr.8: 142879138-142879138 on Build GRCh38
rs780016868 Chr.8: 142879017-142879017 on Build GRCh38
rs375349633 Chr.8: 142878988-142878988 on Build GRCh38
rs193922539 Chr.8: 142879163-142879163 on Build GRCh38
rs778601992 Chr.8: 142879157-142879157 on Build GRCh38
rs749143255 Chr.8: 142879019-142879019 on Build GRCh38
rs372183481 Chr.8: 142878982-142878982 on Build GRCh38
rs537178555 Chr.8: 142878974-142878974 on Build GRCh38
rs763402949 Chr.8: 142879129-142879129 on Build GRCh38
rs5284 Chr.8: 142879139-142879139 on Build GRCh38
rs5286 Chr.8: 142879029-142879029 on Build GRCh38
rs377423817 Chr.8: 142879042-142879042 on Build GRCh38
rs769450046 Chr.8: 142879168-142879168 on Build GRCh38
rs750620653 Chr.8: 142879145-142879145 on Build GRCh38
rs779342369 Chr.8: 142879002-142879002 on Build GRCh38
rs764418169 Chr.8: 142879083-142879110 on Build GRCh38
rs770400476 Chr.8: 142879093-142879093 on Build GRCh38
rs61751141 Chr.8: 142878992-142878992 on Build GRCh38
rs767045873 Chr.8: 142878991-142878991 on Build GRCh38
rs772003869 Chr.8: 142879079-142879079 on Build GRCh38
rs560011017 Chr.8: 142879013-142879013 on Build GRCh38
rs780624046 Chr.8: 142879165-142879165 on Build GRCh38
rs373778481 Chr.8: 142879160-142879160 on Build GRCh38
rs753202905 Chr.8: 142879043-142879043 on Build GRCh38
rs760355042 Chr.8: 142879036-142879036 on Build GRCh38
rs570031684 Chr.8: 142878965-142878965 on Build GRCh38
rs376893751 Chr.8: 142879171-142879171 on Build GRCh38
rs104894070 Chr.8: 142879146-142879146 on Build GRCh38
rs61751139 Chr.8: 142879092-142879092 on Build GRCh38
rs746948389 Chr.8: 142879162-142879162 on Build GRCh38
rs113340222 Chr.8: 142879066-142879066 on Build GRCh38
rs575153163 Chr.8: 142879101-142879101 on Build GRCh38
rs201137503 Chr.8: 142879052-142879052 on Build GRCh38
rs768465089 Chr.8: 142879022-142879022 on Build GRCh38
rs751694043 Chr.8: 142879086-142879086 on Build GRCh38
rs553707049 Chr.8: 142879026-142879026 on Build GRCh38
rs758884429 Chr.8: 142879044-142879044 on Build GRCh38
rs757389720 Chr.8: 142879053-142879053 on Build GRCh38
rs935479041 Chr.8: 142879106-142879106 on Build GRCh38
rs758387119 Chr.8: 142879062-142879064 on Build GRCh38
rs768426617 Chr.8: 142878986-142878986 on Build GRCh38
rs371431889 Chr.8: 142879097-142879097 on Build GRCh38
rs748036339 Chr.8: 142879076-142879076 on Build GRCh38
rs757489916 Chr.8: 142879016-142879016 on Build GRCh38
rs369926988 Chr.8: 142879025-142879025 on Build GRCh38
rs145050906 Chr.8: 142879060-142879060 on Build GRCh38
rs761269721 Chr.8: 142878990-142878990 on Build GRCh38
rs1029395325 Chr.8: 142879019-142879020 on Build GRCh38
rs143211108 Chr.8: 142879133-142879133 on Build GRCh38
rs150354099 Chr.8: 142879058-142879058 on Build GRCh38
rs745512458 Chr.8: 142879167-142879167 on Build GRCh38
rs776337078 Chr.8: 142879094-142879094 on Build GRCh38
rs777895488 Chr.8: 142879009-142879009 on Build GRCh38
rs5283 Chr.8: 142879181-142879181 on Build GRCh38
rs755503539 Chr.8: 142879001-142879001 on Build GRCh38
rs868587302 Chr.8: 142879147-142879147 on Build GRCh38
rs104894066 Chr.8: 142879080-142879080 on Build GRCh38
rs775055690 Chr.8: 142879182-142879182 on Build GRCh38
rs753221461 Chr.8: 142879006-142879006 on Build GRCh38
rs952727650 Chr.8: 142879048-142879048 on Build GRCh38
rs775858610 Chr.8: 142879120-142879120 on Build GRCh38
rs747249778 Chr.8: 142879014-142879014 on Build GRCh38
rs752490109 Chr.8: 142879158-142879158 on Build GRCh38
rs772733691 Chr.8: 142879081-142879081 on Build GRCh38
rs200993800 Chr.8: 142879135-142879135 on Build GRCh38
rs369117124 Chr.8: 142878981-142878981 on Build GRCh38
rs5285 Chr.8: 142879033-142879033 on Build GRCh38
rs756538930 Chr.8: 142878979-142878979 on Build GRCh38
rs762347776 Chr.8: 142879183-142879183 on Build GRCh38
rs373046641 Chr.8: 142879041-142879041 on Build GRCh38
rs746083604 Chr.8: 142879059-142879059 on Build GRCh38
rs766731958 Chr.8: 142878996-142878996 on Build GRCh38
rs754165418 Chr.8: 142879000-142879000 on Build GRCh38

View Full Product Details