Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 47456552-47456801 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMMD8
Entrez Gene ID
54951
Gene Name
COMM domain containing 8
Gene Aliases
-
UniGene
Hs.23956
Gene Chromosome Location
Chr.4: 47450793-47463702 on Build GRCh38
Transcripts
NM_017845.3 XM_017008330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6430562 c.373A>G p.K125E Chr.4: 47456579-47456579 on Build GRCh38
5812411 c.223-2A>T p.? Chr.4: 47456731-47456731 on Build GRCh38
6891341 c.338C>T p.S113F Chr.4: 47456614-47456614 on Build GRCh38
4482044 c.256C>T p.H86Y Chr.4: 47456696-47456696 on Build GRCh38
260256 c.227T>G p.F76C Chr.4: 47456725-47456725 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184044350 Chr.4: 47456756-47456756 on Build GRCh38
rs762627011 Chr.4: 47456637-47456637 on Build GRCh38
rs776662027 Chr.4: 47456589-47456589 on Build GRCh38
rs540984741 Chr.4: 47456614-47456614 on Build GRCh38
rs756675294 Chr.4: 47456689-47456689 on Build GRCh38
rs762860605 Chr.4: 47456772-47456772 on Build GRCh38
rs763818110 Chr.4: 47456638-47456638 on Build GRCh38
rs759511061 Chr.4: 47456730-47456731 on Build GRCh38
rs772161416 Chr.4: 47456739-47456739 on Build GRCh38
rs775366724 Chr.4: 47456771-47456771 on Build GRCh38
rs770725774 Chr.4: 47456588-47456588 on Build GRCh38
rs773222222 Chr.4: 47456740-47456740 on Build GRCh38
rs759595322 Chr.4: 47456605-47456605 on Build GRCh38
rs886528592 Chr.4: 47456672-47456672 on Build GRCh38
rs756818264 Chr.4: 47456664-47456664 on Build GRCh38
rs1004928509 Chr.4: 47456710-47456710 on Build GRCh38
rs750981707 Chr.4: 47456647-47456647 on Build GRCh38
rs780602871 Chr.4: 47456665-47456665 on Build GRCh38
rs769857271 Chr.4: 47456752-47456752 on Build GRCh38
rs759542122 Chr.4: 47456742-47456742 on Build GRCh38
rs779220775 Chr.4: 47456725-47456725 on Build GRCh38
rs200625536 Chr.4: 47456673-47456673 on Build GRCh38
rs768809837 Chr.4: 47456717-47456717 on Build GRCh38
rs748138078 Chr.4: 47456736-47456736 on Build GRCh38
rs752603657 Chr.4: 47456618-47456618 on Build GRCh38
rs749660918 Chr.4: 47456713-47456713 on Build GRCh38
rs1040980400 Chr.4: 47456729-47456729 on Build GRCh38
rs11541139 Chr.4: 47456655-47456655 on Build GRCh38
rs968832596 Chr.4: 47456628-47456628 on Build GRCh38

View Full Product Details