Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 95428506-95428779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM81B
Entrez Gene ID
153643
Gene Name
family with sequence similarity 81 member B
Gene Aliases
-
UniGene
Hs.276287
Gene Chromosome Location
Chr.5: 95384793-95450441 on Build GRCh38
Transcripts
NM_152548.2 XM_005271905.2 XM_005271906.4 XM_011543207.1 XM_011543208.1 XM_011543209.1 XM_011543210.2 XM_017009118.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7166889 c.736C>G p.Q246E Chr.5: 95428682-95428682 on Build GRCh38
4825346 c.657-1G>C p.? Chr.5: 95428602-95428602 on Build GRCh38
3856947 c.751G>A p.A251T Chr.5: 95428697-95428697 on Build GRCh38
281016 c.744C>T p.F248F Chr.5: 95428690-95428690 on Build GRCh38
4625620 c.784A>G p.K262E Chr.5: 95428730-95428730 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190506251 Chr.5: 95428650-95428650 on Build GRCh38
rs571687149 Chr.5: 95428556-95428556 on Build GRCh38
rs943364618 Chr.5: 95428550-95428550 on Build GRCh38
rs747243303 Chr.5: 95428596-95428596 on Build GRCh38
rs545139648 Chr.5: 95428694-95428694 on Build GRCh38
rs768064943 Chr.5: 95428644-95428644 on Build GRCh38
rs1541797 Chr.5: 95428662-95428662 on Build GRCh38
rs754883002 Chr.5: 95428674-95428674 on Build GRCh38
rs770077907 Chr.5: 95428697-95428697 on Build GRCh38
rs775893980 Chr.5: 95428635-95428635 on Build GRCh38
rs778993418 Chr.5: 95428679-95428679 on Build GRCh38
rs201492691 Chr.5: 95428696-95428696 on Build GRCh38
rs769272282 Chr.5: 95428704-95428704 on Build GRCh38
rs773783618 Chr.5: 95428646-95428646 on Build GRCh38
rs774825070 Chr.5: 95428708-95428708 on Build GRCh38
rs750053234 Chr.5: 95428659-95428659 on Build GRCh38
rs746286929 Chr.5: 95428612-95428612 on Build GRCh38
rs372529698 Chr.5: 95428728-95428728 on Build GRCh38
rs770164622 Chr.5: 95428632-95428632 on Build GRCh38
rs77367879 Chr.5: 95428584-95428584 on Build GRCh38
rs1004161551 Chr.5: 95428614-95428614 on Build GRCh38
rs1039037635 Chr.5: 95428553-95428553 on Build GRCh38
rs558073227 Chr.5: 95428544-95428544 on Build GRCh38
rs547904033 Chr.5: 95428661-95428661 on Build GRCh38
rs376930847 Chr.5: 95428562-95428562 on Build GRCh38
rs759242751 Chr.5: 95428748-95428748 on Build GRCh38
rs779901715 Chr.5: 95428576-95428576 on Build GRCh38
rs529803514 Chr.5: 95428597-95428597 on Build GRCh38
rs776559594 Chr.5: 95428747-95428747 on Build GRCh38
rs150926447 Chr.5: 95428691-95428691 on Build GRCh38
rs946776700 Chr.5: 95428563-95428563 on Build GRCh38
rs1022614847 Chr.5: 95428711-95428711 on Build GRCh38
rs771536970 Chr.5: 95428727-95428727 on Build GRCh38
rs760321217 Chr.5: 95428737-95428737 on Build GRCh38
rs780408975 Chr.5: 95428702-95428702 on Build GRCh38
rs751016445 Chr.5: 95428543-95428543 on Build GRCh38
rs139557479 Chr.5: 95428582-95428582 on Build GRCh38
rs1053780937 Chr.5: 95428591-95428591 on Build GRCh38
rs950576814 Chr.5: 95428735-95428735 on Build GRCh38
rs763467946 Chr.5: 95428637-95428637 on Build GRCh38
rs754284288 Chr.5: 95428658-95428658 on Build GRCh38
rs777857392 Chr.5: 95428590-95428590 on Build GRCh38
rs771350458 Chr.5: 95428604-95428604 on Build GRCh38
rs772880748 Chr.5: 95428733-95428733 on Build GRCh38
rs536458755 Chr.5: 95428698-95428698 on Build GRCh38
rs1026517491 Chr.5: 95428734-95428734 on Build GRCh38
rs145514102 Chr.5: 95428558-95428558 on Build GRCh38
rs753737738 Chr.5: 95428666-95428666 on Build GRCh38
rs751751535 Chr.5: 95428690-95428690 on Build GRCh38
rs374317657 Chr.5: 95428723-95428723 on Build GRCh38
rs781554523 Chr.5: 95428606-95428606 on Build GRCh38

View Full Product Details