Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118590290-118590551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARCN1
Entrez Gene ID
372
Gene Name
archain 1
Gene Aliases
COPD
UniGene
Hs.33642
Gene Chromosome Location
Chr.11: 118572363-118603033 on Build GRCh38
Transcripts
NM_001142281.1 NM_001655.4 XM_005271542.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7186424 c.826A>G p.M276V Chr.11: 118590348-118590348 on Build GRCh38
6251727 c.931G>T p.G311C Chr.11: 118590453-118590453 on Build GRCh38
6602821 c.935G>A p.R312Q Chr.11: 118590457-118590457 on Build GRCh38
1245301 c.895G>A p.G299S Chr.11: 118590417-118590417 on Build GRCh38
6842075 c.893A>G p.H298R Chr.11: 118590415-118590415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148276430 Chr.11: 118590509-118590509 on Build GRCh38
rs149615562 Chr.11: 118590467-118590467 on Build GRCh38
rs781923327 Chr.11: 118590346-118590346 on Build GRCh38
rs782665421 Chr.11: 118590435-118590436 on Build GRCh38
rs782487231 Chr.11: 118590463-118590463 on Build GRCh38
rs773412345 Chr.11: 118590479-118590479 on Build GRCh38
rs1035134433 Chr.11: 118590445-118590445 on Build GRCh38
rs782036972 Chr.11: 118590378-118590378 on Build GRCh38
rs782367570 Chr.11: 118590342-118590342 on Build GRCh38
rs1063124 Chr.11: 118590449-118590449 on Build GRCh38
rs781854385 Chr.11: 118590431-118590432 on Build GRCh38
rs782009352 Chr.11: 118590333-118590333 on Build GRCh38
rs782790694 Chr.11: 118590377-118590377 on Build GRCh38
rs138250193 Chr.11: 118590457-118590457 on Build GRCh38
rs978102572 Chr.11: 118590444-118590444 on Build GRCh38
rs782156810 Chr.11: 118590419-118590419 on Build GRCh38
rs782020909 Chr.11: 118590497-118590497 on Build GRCh38
rs782109325 Chr.11: 118590382-118590382 on Build GRCh38
rs748079967 Chr.11: 118590415-118590415 on Build GRCh38
rs371011063 Chr.11: 118590389-118590389 on Build GRCh38
rs147118477 Chr.11: 118590390-118590390 on Build GRCh38
rs781996712 Chr.11: 118590514-118590514 on Build GRCh38
rs781905158 Chr.11: 118590438-118590439 on Build GRCh38
rs782223696 Chr.11: 118590337-118590337 on Build GRCh38
rs530117599 Chr.11: 118590313-118590313 on Build GRCh38
rs782384548 Chr.11: 118590330-118590330 on Build GRCh38
rs201094957 Chr.11: 118590510-118590510 on Build GRCh38
rs916709319 Chr.11: 118590475-118590475 on Build GRCh38
rs763191104 Chr.11: 118590491-118590491 on Build GRCh38
rs375565238 Chr.11: 118590425-118590425 on Build GRCh38
rs782775362 Chr.11: 118590426-118590426 on Build GRCh38
rs782520198 Chr.11: 118590433-118590434 on Build GRCh38
rs782723334 Chr.11: 118590430-118590431 on Build GRCh38
rs782777693 Chr.11: 118590385-118590385 on Build GRCh38

View Full Product Details