Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 76022636-76022889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ART3
Entrez Gene ID
419
Gene Name
ADP-ribosyltransferase 3
Gene Aliases
ARTC3
UniGene
Hs.24976
Gene Chromosome Location
Chr.4: 76011180-76112802 on Build GRCh38
Transcripts
NM_001130017.2 NM_001565.3

Gene Symbol
CXCL10
Entrez Gene ID
3627
Gene Name
C-X-C motif chemokine ligand 10
Gene Aliases
C7, IFI10, INP10, IP-10, SCYB10, crg-2, gIP-10, mob-1
UniGene
Hs.632586
Gene Chromosome Location
Chr.4: 76021116-76023536 on Build GRCh38
Transcripts
NM_001130017.2 NM_001565.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3826224 c.82G>A p.V28I Chr.4: 76022797-76022797 on Build GRCh38
481513 c.108T>C p.S36S Chr.4: 76022771-76022771 on Build GRCh38
1319316 c.138A>G p.E46E Chr.4: 76022741-76022741 on Build GRCh38
3605907 c.100A>G p.S34G Chr.4: 76022779-76022779 on Build GRCh38
6469685 c.85C>T p.R29C Chr.4: 76022794-76022794 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896618954 Chr.4: 76022836-76022836 on Build GRCh38
rs756375420 Chr.4: 76022820-76022820 on Build GRCh38
rs774798493 Chr.4: 76022728-76022728 on Build GRCh38
rs11548618 Chr.4: 76022794-76022794 on Build GRCh38
rs748534350 Chr.4: 76022732-76022732 on Build GRCh38
rs760437717 Chr.4: 76022776-76022776 on Build GRCh38
rs772534996 Chr.4: 76022737-76022737 on Build GRCh38
rs557248373 Chr.4: 76022782-76022782 on Build GRCh38
rs773414753 Chr.4: 76022751-76022751 on Build GRCh38
rs772450528 Chr.4: 76022853-76022853 on Build GRCh38
rs752574654 Chr.4: 76022674-76022674 on Build GRCh38
rs753948493 Chr.4: 76022832-76022832 on Build GRCh38
rs201830102 Chr.4: 76022790-76022790 on Build GRCh38
rs777837361 Chr.4: 76022841-76022843 on Build GRCh38
rs762956789 Chr.4: 76022796-76022796 on Build GRCh38
rs756286152 Chr.4: 76022704-76022704 on Build GRCh38
rs758220509 Chr.4: 76022675-76022675 on Build GRCh38
rs778254950 Chr.4: 76022854-76022854 on Build GRCh38
rs200825234 Chr.4: 76022722-76022722 on Build GRCh38
rs73829006 Chr.4: 76022858-76022858 on Build GRCh38
rs897559459 Chr.4: 76022718-76022718 on Build GRCh38
rs776216030 Chr.4: 76022791-76022791 on Build GRCh38
rs778277559 Chr.4: 76022705-76022705 on Build GRCh38
rs748693711 Chr.4: 76022850-76022850 on Build GRCh38
rs374869885 Chr.4: 76022834-76022834 on Build GRCh38
rs769298828 Chr.4: 76022852-76022852 on Build GRCh38
rs759079472 Chr.4: 76022793-76022793 on Build GRCh38
rs746142444 Chr.4: 76022703-76022703 on Build GRCh38
rs780075270 Chr.4: 76022821-76022821 on Build GRCh38
rs930323843 Chr.4: 76022720-76022720 on Build GRCh38
rs750468593 Chr.4: 76022677-76022677 on Build GRCh38
rs200829450 Chr.4: 76022706-76022706 on Build GRCh38
rs748077493 Chr.4: 76022804-76022804 on Build GRCh38
rs778941036 Chr.4: 76022845-76022845 on Build GRCh38
rs754904457 Chr.4: 76022679-76022679 on Build GRCh38
rs751298966 Chr.4: 76022808-76022808 on Build GRCh38

View Full Product Details