Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44814607-44814840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAM
Entrez Gene ID
4059
Gene Name
basal cell adhesion molecule (Lutheran blood group)
Gene Aliases
AU, CD239, LU, MSK19
UniGene
Hs.625725
Gene Chromosome Location
Chr.19: 44809059-44821421 on Build GRCh38
Transcripts
NM_001013257.2 NM_005581.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7106288 c.925G>A p.E309K Chr.19: 44814607-44814607 on Build GRCh38
6377671 c.1028A>G p.D343G Chr.19: 44814710-44814710 on Build GRCh38
4665300 c.956_957insG p.N321fs*27 Chr.19: 44814638-44814639 on Build GRCh38
243178 c.986G>A p.R329Q Chr.19: 44814668-44814668 on Build GRCh38
712400 c.976G>T p.G326* Chr.19: 44814658-44814658 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755899513 Chr.19: 44814751-44814751 on Build GRCh38
rs752440604 Chr.19: 44814747-44814747 on Build GRCh38
rs373208429 Chr.19: 44814783-44814783 on Build GRCh38
rs188278431 Chr.19: 44814685-44814685 on Build GRCh38
rs1028802523 Chr.19: 44814816-44814816 on Build GRCh38
rs748839969 Chr.19: 44814695-44814695 on Build GRCh38
rs754292035 Chr.19: 44814720-44814720 on Build GRCh38
rs374759522 Chr.19: 44814726-44814726 on Build GRCh38
rs190232001 Chr.19: 44814637-44814637 on Build GRCh38
rs1000214875 Chr.19: 44814721-44814721 on Build GRCh38
rs369897337 Chr.19: 44814780-44814780 on Build GRCh38
rs770541247 Chr.19: 44814708-44814708 on Build GRCh38
rs532679625 Chr.19: 44814742-44814742 on Build GRCh38
rs148153043 Chr.19: 44814753-44814753 on Build GRCh38
rs139012083 Chr.19: 44814813-44814813 on Build GRCh38
rs956006521 Chr.19: 44814740-44814740 on Build GRCh38
rs777788448 Chr.19: 44814688-44814688 on Build GRCh38
rs578111861 Chr.19: 44814784-44814784 on Build GRCh38
rs758536239 Chr.19: 44814741-44814741 on Build GRCh38
rs776868641 Chr.19: 44814667-44814667 on Build GRCh38
rs756894242 Chr.19: 44814760-44814760 on Build GRCh38
rs142102840 Chr.19: 44814711-44814711 on Build GRCh38
rs975536965 Chr.19: 44814797-44814797 on Build GRCh38
rs955574747 Chr.19: 44814817-44814817 on Build GRCh38
rs777165923 Chr.19: 44814787-44814787 on Build GRCh38
rs373486211 Chr.19: 44814768-44814768 on Build GRCh38
rs762307739 Chr.19: 44814801-44814801 on Build GRCh38
rs143753295 Chr.19: 44814714-44814714 on Build GRCh38
rs10410517 Chr.19: 44814779-44814779 on Build GRCh38
rs371206032 Chr.19: 44814712-44814712 on Build GRCh38
rs367891212 Chr.19: 44814651-44814651 on Build GRCh38
rs369153988 Chr.19: 44814735-44814735 on Build GRCh38
rs375498217 Chr.19: 44814678-44814678 on Build GRCh38
rs368656529 Chr.19: 44814668-44814668 on Build GRCh38
rs368698889 Chr.19: 44814709-44814709 on Build GRCh38
rs374263348 Chr.19: 44814654-44814654 on Build GRCh38
rs182019561 Chr.19: 44814677-44814677 on Build GRCh38
rs201966991 Chr.19: 44814724-44814724 on Build GRCh38
rs764537438 Chr.19: 44814715-44814715 on Build GRCh38
rs777905118 Chr.19: 44814752-44814752 on Build GRCh38
rs908765077 Chr.19: 44814694-44814694 on Build GRCh38
rs769936617 Chr.19: 44814722-44814722 on Build GRCh38
rs767743074 Chr.19: 44814634-44814634 on Build GRCh38
rs940395375 Chr.19: 44814706-44814706 on Build GRCh38
rs147593462 Chr.19: 44814679-44814679 on Build GRCh38
rs903092506 Chr.19: 44814754-44814754 on Build GRCh38
rs775728050 Chr.19: 44814713-44814713 on Build GRCh38
rs778552042 Chr.19: 44814775-44814775 on Build GRCh38
rs753313618 Chr.19: 44814636-44814636 on Build GRCh38
rs957437270 Chr.19: 44814649-44814649 on Build GRCh38

View Full Product Details