Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33038680-33038939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJA1
Entrez Gene ID
3301
Gene Name
DnaJ heat shock protein family (Hsp40) member A1
Gene Aliases
DJ-2, DjA1, HDJ2, HSDJ, HSJ-2, HSJ2, HSPF4, NEDD7, hDJ-2
UniGene
Hs.445203
Gene Chromosome Location
Chr.9: 33025203-33039907 on Build GRCh38
Transcripts
NM_001314039.1 NM_001539.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
232194 c.985C>T p.P329S Chr.9: 33038694-33038694 on Build GRCh38
3906869 c.1063A>G p.T355A Chr.9: 33038772-33038772 on Build GRCh38
5177912 c.1173_1174insGTTCAGTG p.C394fs*>7 Chr.9: 33038882-33038883 on Build GRCh38
5507159 c.1051G>A p.E351K Chr.9: 33038760-33038760 on Build GRCh38
3906870 c.1127A>G p.Y376C Chr.9: 33038836-33038836 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748280590 Chr.9: 33038868-33038868 on Build GRCh38
rs756513491 Chr.9: 33038827-33038827 on Build GRCh38
rs1039624400 Chr.9: 33038765-33038765 on Build GRCh38
rs750767685 Chr.9: 33038823-33038823 on Build GRCh38
rs374710198 Chr.9: 33038830-33038830 on Build GRCh38
rs754394456 Chr.9: 33038863-33038865 on Build GRCh38
rs568893066 Chr.9: 33038865-33038865 on Build GRCh38
rs766703382 Chr.9: 33038796-33038797 on Build GRCh38
rs781143779 Chr.9: 33038912-33038912 on Build GRCh38
rs756282667 Chr.9: 33038705-33038705 on Build GRCh38
rs778818641 Chr.9: 33038861-33038861 on Build GRCh38
rs758245297 Chr.9: 33038872-33038872 on Build GRCh38
rs775074991 Chr.9: 33038796-33038796 on Build GRCh38
rs141132012 Chr.9: 33038829-33038829 on Build GRCh38
rs780305363 Chr.9: 33038706-33038706 on Build GRCh38
rs776490065 Chr.9: 33038780-33038780 on Build GRCh38
rs368226813 Chr.9: 33038838-33038838 on Build GRCh38
rs749336661 Chr.9: 33038712-33038712 on Build GRCh38
rs777895077 Chr.9: 33038885-33038885 on Build GRCh38
rs1803513 Chr.9: 33038884-33038884 on Build GRCh38
rs772077578 Chr.9: 33038758-33038758 on Build GRCh38
rs770904664 Chr.9: 33038904-33038904 on Build GRCh38
rs778896977 Chr.9: 33038750-33038750 on Build GRCh38
rs773091099 Chr.9: 33038761-33038761 on Build GRCh38
rs768094883 Chr.9: 33038800-33038800 on Build GRCh38
rs746793842 Chr.9: 33038894-33038894 on Build GRCh38
rs868488974 Chr.9: 33038786-33038786 on Build GRCh38
rs746848794 Chr.9: 33038772-33038772 on Build GRCh38
rs776768067 Chr.9: 33038909-33038909 on Build GRCh38
rs185238499 Chr.9: 33038837-33038837 on Build GRCh38
rs759099963 Chr.9: 33038789-33038789 on Build GRCh38
rs148382351 Chr.9: 33038756-33038756 on Build GRCh38
rs367858309 Chr.9: 33038787-33038787 on Build GRCh38
rs867748833 Chr.9: 33038751-33038751 on Build GRCh38
rs890716045 Chr.9: 33038747-33038747 on Build GRCh38
rs770673934 Chr.9: 33038776-33038776 on Build GRCh38
rs755081187 Chr.9: 33038718-33038718 on Build GRCh38

View Full Product Details