Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 73736926-73737189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD11
Entrez Gene ID
83451
Gene Name
abhydrolase domain containing 11
Gene Aliases
PP1226, WBSCR21
UniGene
Hs.647045
Gene Chromosome Location
Chr.7: 73736094-73738867 on Build GRCh38
Transcripts
NM_001145364.2 NM_001301058.1 NM_001321382.1 NM_001321383.1 NM_148912.3 NM_148913.3 NR_026912.2 NR_135627.1 XM_006716144.2 XM_011516590.2 XM_011516591.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1451997 c.690C>T p.F230F Chr.7: 73737054-73737054 on Build GRCh38
1451998 c.681C>T p.D227D Chr.7: 73737063-73737063 on Build GRCh38
7151801 c.779C>A p.T260K Chr.7: 73736965-73736965 on Build GRCh38
3087351 c.789C>T p.L263L Chr.7: 73736955-73736955 on Build GRCh38
3087353 c.735G>T p.K245N Chr.7: 73737009-73737009 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782284150 Chr.7: 73737093-73737093 on Build GRCh38
rs373725417 Chr.7: 73737147-73737147 on Build GRCh38
rs150032016 Chr.7: 73737136-73737136 on Build GRCh38
rs371548132 Chr.7: 73737156-73737156 on Build GRCh38
rs782628985 Chr.7: 73737032-73737032 on Build GRCh38
rs994674858 Chr.7: 73737114-73737114 on Build GRCh38
rs150652642 Chr.7: 73737074-73737074 on Build GRCh38
rs782188494 Chr.7: 73737098-73737098 on Build GRCh38
rs782113507 Chr.7: 73737059-73737059 on Build GRCh38
rs112164225 Chr.7: 73737011-73737011 on Build GRCh38
rs901517537 Chr.7: 73737047-73737047 on Build GRCh38
rs1052083837 Chr.7: 73737039-73737039 on Build GRCh38
rs969323144 Chr.7: 73737164-73737164 on Build GRCh38
rs139322195 Chr.7: 73736970-73736970 on Build GRCh38
rs781814593 Chr.7: 73737148-73737148 on Build GRCh38
rs782333441 Chr.7: 73736992-73736992 on Build GRCh38
rs782521802 Chr.7: 73737096-73737096 on Build GRCh38
rs782265860 Chr.7: 73737113-73737113 on Build GRCh38
rs200259168 Chr.7: 73737138-73737138 on Build GRCh38
rs370080737 Chr.7: 73737119-73737119 on Build GRCh38
rs776064862 Chr.7: 73737052-73737052 on Build GRCh38
rs201294310 Chr.7: 73737133-73737133 on Build GRCh38
rs782372341 Chr.7: 73736969-73736969 on Build GRCh38
rs781927551 Chr.7: 73737023-73737023 on Build GRCh38
rs782065664 Chr.7: 73737132-73737132 on Build GRCh38
rs781806554 Chr.7: 73737069-73737069 on Build GRCh38
rs201844573 Chr.7: 73737070-73737070 on Build GRCh38
rs369313640 Chr.7: 73737062-73737062 on Build GRCh38
rs138792415 Chr.7: 73737078-73737078 on Build GRCh38
rs782004814 Chr.7: 73737058-73737058 on Build GRCh38
rs537791221 Chr.7: 73737054-73737054 on Build GRCh38
rs782605297 Chr.7: 73736955-73736955 on Build GRCh38
rs782253011 Chr.7: 73736964-73736964 on Build GRCh38
rs367603645 Chr.7: 73737154-73737154 on Build GRCh38
rs916493649 Chr.7: 73737150-73737150 on Build GRCh38
rs782735052 Chr.7: 73737055-73737055 on Build GRCh38
rs368659985 Chr.7: 73737134-73737134 on Build GRCh38
rs782159271 Chr.7: 73737142-73737142 on Build GRCh38
rs201053538 Chr.7: 73737103-73737103 on Build GRCh38
rs372621687 Chr.7: 73737128-73737128 on Build GRCh38
rs143075789 Chr.7: 73737097-73737097 on Build GRCh38
rs1007002367 Chr.7: 73737084-73737084 on Build GRCh38
rs782364772 Chr.7: 73737101-73737101 on Build GRCh38
rs782288758 Chr.7: 73736972-73736972 on Build GRCh38

View Full Product Details